Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50071222-50071728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
UniGene
Hs.265829
Gene Chromosome Location
Chr.17: 50055968-50090485 on Build GRCh38
Transcripts
NM_002204.3 XM_005257308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135958 c.833T>C p.M278T Chr.17: 50071392-50071392 on Build GRCh38
1384144 c.903G>A p.S301S Chr.17: 50071462-50071462 on Build GRCh38
1384142 c.839C>T p.A280V Chr.17: 50071398-50071398 on Build GRCh38
3819908 c.957T>C p.D319D Chr.17: 50071516-50071516 on Build GRCh38
7299136 c.912C>T p.G304G Chr.17: 50071471-50071471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779778232 Chr.17: 50071471-50071471 on Build GRCh38
rs970462555 Chr.17: 50071683-50071683 on Build GRCh38
rs759973429 Chr.17: 50071436-50071436 on Build GRCh38
rs150491650 Chr.17: 50071248-50071248 on Build GRCh38
rs2230390 Chr.17: 50071361-50071361 on Build GRCh38
rs892666228 Chr.17: 50071667-50071667 on Build GRCh38
rs552718616 Chr.17: 50071556-50071556 on Build GRCh38
rs888545435 Chr.17: 50071577-50071577 on Build GRCh38
rs141746825 Chr.17: 50071320-50071320 on Build GRCh38
rs371908585 Chr.17: 50071527-50071527 on Build GRCh38
rs919564171 Chr.17: 50071391-50071391 on Build GRCh38
rs777946030 Chr.17: 50071493-50071493 on Build GRCh38
rs968442272 Chr.17: 50071699-50071699 on Build GRCh38
rs957092154 Chr.17: 50071321-50071321 on Build GRCh38
rs1981515 Chr.17: 50071559-50071559 on Build GRCh38
rs778440230 Chr.17: 50071362-50071362 on Build GRCh38
rs145459787 Chr.17: 50071396-50071396 on Build GRCh38
rs745505565 Chr.17: 50071380-50071380 on Build GRCh38
rs148050628 Chr.17: 50071397-50071397 on Build GRCh38
rs906497068 Chr.17: 50071594-50071594 on Build GRCh38
rs1031654314 Chr.17: 50071600-50071600 on Build GRCh38
rs779646280 Chr.17: 50071392-50071392 on Build GRCh38
rs757988580 Chr.17: 50071462-50071462 on Build GRCh38
rs756726866 Chr.17: 50071343-50071343 on Build GRCh38
rs1008815419 Chr.17: 50071528-50071528 on Build GRCh38
rs141797951 Chr.17: 50071398-50071398 on Build GRCh38
rs1011646862 Chr.17: 50071257-50071257 on Build GRCh38
rs961612470 Chr.17: 50071379-50071379 on Build GRCh38
rs923767050 Chr.17: 50071692-50071692 on Build GRCh38
rs982360346 Chr.17: 50071688-50071688 on Build GRCh38
rs752451792 Chr.17: 50071434-50071434 on Build GRCh38
rs774533994 Chr.17: 50071427-50071427 on Build GRCh38
rs768088650 Chr.17: 50071325-50071325 on Build GRCh38
rs138622747 Chr.17: 50071318-50071318 on Build GRCh38
rs61730088 Chr.17: 50071488-50071488 on Build GRCh38
rs758967122 Chr.17: 50071339-50071339 on Build GRCh38
rs150595761 Chr.17: 50071461-50071461 on Build GRCh38
rs763327889 Chr.17: 50071315-50071315 on Build GRCh38
rs970984820 Chr.17: 50071299-50071299 on Build GRCh38
rs56320412 Chr.17: 50071465-50071465 on Build GRCh38
rs746916084 Chr.17: 50071271-50071271 on Build GRCh38
rs768640879 Chr.17: 50071287-50071287 on Build GRCh38
rs540714229 Chr.17: 50071544-50071544 on Build GRCh38
rs763203601 Chr.17: 50071420-50071420 on Build GRCh38
rs200751011 Chr.17: 50071426-50071426 on Build GRCh38
rs375561420 Chr.17: 50071564-50071564 on Build GRCh38
rs531492869 Chr.17: 50071267-50071267 on Build GRCh38
rs199801873 Chr.17: 50071399-50071399 on Build GRCh38
rs1017931118 Chr.17: 50071703-50071703 on Build GRCh38
rs761101448 Chr.17: 50071563-50071563 on Build GRCh38
rs765496899 Chr.17: 50071266-50071266 on Build GRCh38
rs569140894 Chr.17: 50071681-50071681 on Build GRCh38
rs764721144 Chr.17: 50071444-50071444 on Build GRCh38
rs535573258 Chr.17: 50071540-50071540 on Build GRCh38
rs1012240480 Chr.17: 50071245-50071245 on Build GRCh38
rs765449610 Chr.17: 50071300-50071300 on Build GRCh38
rs773547087 Chr.17: 50071313-50071313 on Build GRCh38
rs746833221 Chr.17: 50071393-50071393 on Build GRCh38
rs777480064 Chr.17: 50071385-50071385 on Build GRCh38
rs374953336 Chr.17: 50071294-50071294 on Build GRCh38
rs139244482 Chr.17: 50071486-50071486 on Build GRCh38
rs761985021 Chr.17: 50071298-50071298 on Build GRCh38
rs780594058 Chr.17: 50071408-50071408 on Build GRCh38
rs751211130 Chr.17: 50071472-50071472 on Build GRCh38
rs763950453 Chr.17: 50071582-50071582 on Build GRCh38
rs560301275 Chr.17: 50071256-50071256 on Build GRCh38
rs1022396228 Chr.17: 50071693-50071693 on Build GRCh38
rs1019747688 Chr.17: 50071557-50071557 on Build GRCh38
rs749405129 Chr.17: 50071494-50071494 on Build GRCh38
rs773496015 Chr.17: 50071409-50071409 on Build GRCh38
rs776514708 Chr.17: 50071292-50071292 on Build GRCh38
rs761225872 Chr.17: 50071439-50071439 on Build GRCh38
rs780925917 Chr.17: 50071702-50071702 on Build GRCh38
rs113850944 Chr.17: 50071309-50071309 on Build GRCh38
rs549142503 Chr.17: 50071587-50071587 on Build GRCh38
rs771016039 Chr.17: 50071495-50071495 on Build GRCh38
rs367698113 Chr.17: 50071487-50071487 on Build GRCh38
rs753243623 Chr.17: 50071332-50071332 on Build GRCh38
rs1026630972 Chr.17: 50071605-50071605 on Build GRCh38
rs746528236 Chr.17: 50071386-50071386 on Build GRCh38
rs1003831915 Chr.17: 50071595-50071595 on Build GRCh38
rs774597013 Chr.17: 50071522-50071522 on Build GRCh38
rs952367774 Chr.17: 50071617-50071617 on Build GRCh38
rs890000484 Chr.17: 50071510-50071510 on Build GRCh38
rs546685892 Chr.17: 50071437-50071437 on Build GRCh38

View Full Product Details