Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50071906-50072413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
UniGene
Hs.265829
Gene Chromosome Location
Chr.17: 50055968-50090485 on Build GRCh38
Transcripts
NM_002204.3 XM_005257308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2698745 c.1047G>A p.A349A Chr.17: 50072073-50072073 on Build GRCh38
71208 c.1091C>G p.P364R Chr.17: 50072117-50072117 on Build GRCh38
2698743 c.982G>A p.A328T Chr.17: 50072008-50072008 on Build GRCh38
3755647 c.996C>T p.F332F Chr.17: 50072022-50072022 on Build GRCh38
3795758 c.1066C>T p.H356Y Chr.17: 50072092-50072092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755938628 Chr.17: 50072085-50072085 on Build GRCh38
rs61730085 Chr.17: 50072139-50072139 on Build GRCh38
rs536348230 Chr.17: 50072339-50072339 on Build GRCh38
rs897348359 Chr.17: 50072202-50072202 on Build GRCh38
rs140821385 Chr.17: 50072081-50072081 on Build GRCh38
rs777892581 Chr.17: 50071990-50071990 on Build GRCh38
rs757333788 Chr.17: 50072008-50072008 on Build GRCh38
rs760776525 Chr.17: 50072306-50072306 on Build GRCh38
rs765857108 Chr.17: 50071949-50071949 on Build GRCh38
rs1003739759 Chr.17: 50072287-50072287 on Build GRCh38
rs770219180 Chr.17: 50072208-50072208 on Build GRCh38
rs760458341 Chr.17: 50072229-50072229 on Build GRCh38
rs549115710 Chr.17: 50072086-50072086 on Build GRCh38
rs375802918 Chr.17: 50071938-50071938 on Build GRCh38
rs989321555 Chr.17: 50072322-50072322 on Build GRCh38
rs200979989 Chr.17: 50072176-50072176 on Build GRCh38
rs906188131 Chr.17: 50072280-50072280 on Build GRCh38
rs780977081 Chr.17: 50071964-50071964 on Build GRCh38
rs1052867931 Chr.17: 50072154-50072154 on Build GRCh38
rs778982919 Chr.17: 50072130-50072130 on Build GRCh38
rs775772151 Chr.17: 50072043-50072043 on Build GRCh38
rs889305433 Chr.17: 50072250-50072250 on Build GRCh38
rs747397146 Chr.17: 50072045-50072045 on Build GRCh38
rs560139902 Chr.17: 50072054-50072054 on Build GRCh38
rs769110428 Chr.17: 50072052-50072052 on Build GRCh38
rs2269772 Chr.17: 50072022-50072022 on Build GRCh38
rs759172421 Chr.17: 50072072-50072072 on Build GRCh38
rs556246893 Chr.17: 50072347-50072347 on Build GRCh38
rs538397377 Chr.17: 50072143-50072143 on Build GRCh38
rs879189423 Chr.17: 50072218-50072232 on Build GRCh38
rs756071354 Chr.17: 50071980-50071980 on Build GRCh38
rs900936940 Chr.17: 50072278-50072278 on Build GRCh38
rs747413363 Chr.17: 50072204-50072217 on Build GRCh38
rs772187398 Chr.17: 50072026-50072026 on Build GRCh38
rs753810579 Chr.17: 50072115-50072115 on Build GRCh38
rs763974233 Chr.17: 50072234-50072234 on Build GRCh38
rs758596606 Chr.17: 50072145-50072145 on Build GRCh38
rs757205480 Chr.17: 50072116-50072116 on Build GRCh38
rs777049499 Chr.17: 50072196-50072196 on Build GRCh38
rs775258042 Chr.17: 50072224-50072224 on Build GRCh38
rs201297040 Chr.17: 50072073-50072073 on Build GRCh38
rs765910090 Chr.17: 50072062-50072062 on Build GRCh38
rs777078098 Chr.17: 50072300-50072300 on Build GRCh38
rs1041122221 Chr.17: 50072200-50072200 on Build GRCh38
rs530584009 Chr.17: 50072323-50072323 on Build GRCh38
rs188095913 Chr.17: 50072260-50072260 on Build GRCh38
rs748623564 Chr.17: 50072198-50072198 on Build GRCh38
rs369616046 Chr.17: 50071953-50071953 on Build GRCh38
rs773739927 Chr.17: 50072068-50072068 on Build GRCh38
rs35973528 Chr.17: 50072104-50072104 on Build GRCh38
rs780284856 Chr.17: 50072153-50072153 on Build GRCh38
rs1011868262 Chr.17: 50071994-50071994 on Build GRCh38
rs773865164 Chr.17: 50072217-50072217 on Build GRCh38
rs909749394 Chr.17: 50072079-50072079 on Build GRCh38
rs35000551 Chr.17: 50072200-50072201 on Build GRCh38
rs180725800 Chr.17: 50072372-50072372 on Build GRCh38
rs746096651 Chr.17: 50072023-50072023 on Build GRCh38
rs1007668009 Chr.17: 50072253-50072253 on Build GRCh38
rs373498450 Chr.17: 50072189-50072189 on Build GRCh38
rs147295823 Chr.17: 50072007-50072007 on Build GRCh38
rs762285671 Chr.17: 50072061-50072061 on Build GRCh38
rs377253057 Chr.17: 50072218-50072218 on Build GRCh38
rs768974658 Chr.17: 50072203-50072204 on Build GRCh38
rs534253939 Chr.17: 50072308-50072308 on Build GRCh38
rs553788847 Chr.17: 50072321-50072321 on Build GRCh38
rs752556325 Chr.17: 50071967-50071967 on Build GRCh38
rs762420164 Chr.17: 50071941-50071941 on Build GRCh38
rs200202112 Chr.17: 50071958-50071958 on Build GRCh38

View Full Product Details