Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50073835-50074336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
UniGene
Hs.265829
Gene Chromosome Location
Chr.17: 50055968-50090485 on Build GRCh38
Transcripts
NM_002204.3 XM_005257308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755649 c.1230T>C p.L410L Chr.17: 50073989-50073989 on Build GRCh38
4438715 c.1246G>A p.V416I Chr.17: 50074144-50074144 on Build GRCh38
4960163 c.1255G>A p.G419R Chr.17: 50074153-50074153 on Build GRCh38
4392208 c.1202T>A p.I401N Chr.17: 50073961-50073961 on Build GRCh38
2698767 c.1382G>A p.R461Q Chr.17: 50074280-50074280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375217313 Chr.17: 50074053-50074053 on Build GRCh38
rs140325939 Chr.17: 50074017-50074017 on Build GRCh38
rs753007168 Chr.17: 50074008-50074008 on Build GRCh38
rs565778216 Chr.17: 50074167-50074167 on Build GRCh38
rs760837392 Chr.17: 50074291-50074291 on Build GRCh38
rs754265360 Chr.17: 50074098-50074098 on Build GRCh38
rs549725925 Chr.17: 50074041-50074041 on Build GRCh38
rs765120355 Chr.17: 50073943-50073943 on Build GRCh38
rs765387709 Chr.17: 50074039-50074039 on Build GRCh38
rs757788850 Chr.17: 50074099-50074099 on Build GRCh38
rs765107531 Chr.17: 50073867-50073867 on Build GRCh38
rs762902142 Chr.17: 50074156-50074157 on Build GRCh38
rs770756249 Chr.17: 50074158-50074158 on Build GRCh38
rs370182931 Chr.17: 50073895-50073895 on Build GRCh38
rs1002618031 Chr.17: 50074070-50074070 on Build GRCh38
rs769770674 Chr.17: 50074147-50074150 on Build GRCh38
rs201776534 Chr.17: 50074038-50074038 on Build GRCh38
rs201085427 Chr.17: 50074135-50074135 on Build GRCh38
rs754213910 Chr.17: 50074189-50074189 on Build GRCh38
rs777452976 Chr.17: 50074256-50074256 on Build GRCh38
rs761751659 Chr.17: 50073866-50073866 on Build GRCh38
rs565473963 Chr.17: 50073901-50073901 on Build GRCh38
rs368943810 Chr.17: 50074298-50074298 on Build GRCh38
rs772836425 Chr.17: 50074151-50074151 on Build GRCh38
rs780821916 Chr.17: 50074137-50074137 on Build GRCh38
rs1038851543 Chr.17: 50074145-50074145 on Build GRCh38
rs749750178 Chr.17: 50074016-50074016 on Build GRCh38
rs762945257 Chr.17: 50073959-50073959 on Build GRCh38
rs968305603 Chr.17: 50073887-50073887 on Build GRCh38
rs2285524 Chr.17: 50073989-50073989 on Build GRCh38
rs779593402 Chr.17: 50074100-50074100 on Build GRCh38
rs746538302 Chr.17: 50074019-50074019 on Build GRCh38
rs749115741 Chr.17: 50074156-50074156 on Build GRCh38
rs776970761 Chr.17: 50074294-50074294 on Build GRCh38
rs78768954 Chr.17: 50074188-50074188 on Build GRCh38
rs761950316 Chr.17: 50074113-50074114 on Build GRCh38
rs770703080 Chr.17: 50074267-50074267 on Build GRCh38
rs771691787 Chr.17: 50073930-50073930 on Build GRCh38
rs766380198 Chr.17: 50073968-50073968 on Build GRCh38
rs763321144 Chr.17: 50074300-50074300 on Build GRCh38
rs760335644 Chr.17: 50073934-50073934 on Build GRCh38
rs774253538 Chr.17: 50074160-50074160 on Build GRCh38
rs770299309 Chr.17: 50073911-50073911 on Build GRCh38
rs936553754 Chr.17: 50074075-50074075 on Build GRCh38
rs778254852 Chr.17: 50074011-50074011 on Build GRCh38
rs1004901363 Chr.17: 50074202-50074202 on Build GRCh38
rs1005159439 Chr.17: 50073894-50073894 on Build GRCh38
rs755684284 Chr.17: 50074248-50074248 on Build GRCh38
rs144049965 Chr.17: 50074283-50074283 on Build GRCh38
rs754513773 Chr.17: 50074118-50074118 on Build GRCh38
rs759584545 Chr.17: 50074044-50074044 on Build GRCh38
rs554367981 Chr.17: 50074261-50074261 on Build GRCh38
rs909032691 Chr.17: 50073997-50073997 on Build GRCh38
rs750952837 Chr.17: 50074209-50074209 on Build GRCh38
rs755146298 Chr.17: 50073987-50073987 on Build GRCh38
rs764296048 Chr.17: 50074094-50074094 on Build GRCh38
rs538716387 Chr.17: 50074086-50074086 on Build GRCh38
rs761575916 Chr.17: 50074033-50074033 on Build GRCh38
rs117783166 Chr.17: 50073932-50073932 on Build GRCh38
rs61730084 Chr.17: 50074247-50074247 on Build GRCh38
rs752845203 Chr.17: 50074049-50074049 on Build GRCh38
rs776114748 Chr.17: 50074028-50074028 on Build GRCh38
rs569581246 Chr.17: 50074051-50074051 on Build GRCh38
rs765745707 Chr.17: 50074197-50074197 on Build GRCh38
rs768435486 Chr.17: 50073935-50073935 on Build GRCh38
rs775640037 Chr.17: 50074180-50074180 on Build GRCh38
rs968194256 Chr.17: 50074161-50074161 on Build GRCh38
rs769501290 Chr.17: 50074146-50074146 on Build GRCh38
rs1009628995 Chr.17: 50074133-50074133 on Build GRCh38
rs757664252 Chr.17: 50074193-50074193 on Build GRCh38
rs776437753 Chr.17: 50073942-50073942 on Build GRCh38
rs746415954 Chr.17: 50074104-50074104 on Build GRCh38
rs575130011 Chr.17: 50074184-50074184 on Build GRCh38
rs773145172 Chr.17: 50074152-50074153 on Build GRCh38
rs756416844 Chr.17: 50074009-50074009 on Build GRCh38
rs151126162 Chr.17: 50074276-50074276 on Build GRCh38
rs769472839 Chr.17: 50074034-50074034 on Build GRCh38
rs776779986 Chr.17: 50073996-50073996 on Build GRCh38
rs753687314 Chr.17: 50073865-50073865 on Build GRCh38
rs780510840 Chr.17: 50074245-50074245 on Build GRCh38
rs751654002 Chr.17: 50073986-50073986 on Build GRCh38
rs368191702 Chr.17: 50073957-50073957 on Build GRCh38
rs766943378 Chr.17: 50074307-50074307 on Build GRCh38
rs372535143 Chr.17: 50074045-50074045 on Build GRCh38
rs1042313983 Chr.17: 50074031-50074031 on Build GRCh38
rs747690104 Chr.17: 50074142-50074142 on Build GRCh38
rs376879376 Chr.17: 50074297-50074297 on Build GRCh38
rs750476435 Chr.17: 50073879-50073879 on Build GRCh38
rs778115417 Chr.17: 50073913-50073913 on Build GRCh38
rs1016251016 Chr.17: 50074211-50074211 on Build GRCh38
rs758949436 Chr.17: 50074233-50074233 on Build GRCh38
rs749846498 Chr.17: 50073923-50073923 on Build GRCh38
rs543346479 Chr.17: 50074296-50074296 on Build GRCh38
rs779651025 Chr.17: 50074018-50074018 on Build GRCh38
rs896576191 Chr.17: 50074131-50074131 on Build GRCh38
rs758377704 Chr.17: 50073889-50073889 on Build GRCh38
rs976168995 Chr.17: 50074269-50074269 on Build GRCh38
rs963624366 Chr.17: 50074268-50074268 on Build GRCh38
rs370928871 Chr.17: 50073897-50073897 on Build GRCh38
rs200083436 Chr.17: 50073907-50073907 on Build GRCh38
rs768382419 Chr.17: 50074021-50074021 on Build GRCh38
rs768657346 Chr.17: 50074293-50074293 on Build GRCh38
rs772057144 Chr.17: 50074280-50074280 on Build GRCh38

View Full Product Details