Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50462092-50462594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF2
Entrez Gene ID
80221
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
ACSMW, AVYV493
UniGene
Hs.288959
Gene Chromosome Location
Chr.17: 50426158-50474845 on Build GRCh38
Transcripts
NM_001288968.1 NM_001288969.1 NM_001288970.1 NM_001288971.1 NM_001288972.1 NM_025149.5 XM_006722110.3 XM_011525294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4658972 c.610G>C p.A204P Chr.17: 50462286-50462286 on Build GRCh38
1610456 c.765T>G p.H255Q Chr.17: 50462558-50462558 on Build GRCh38
3889885 c.757T>C p.S253P Chr.17: 50462550-50462550 on Build GRCh38
4657172 c.758C>T p.S253F Chr.17: 50462551-50462551 on Build GRCh38
6502565 c.742A>G p.N248D Chr.17: 50462535-50462535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757750250 Chr.17: 50462222-50462222 on Build GRCh38
rs759419044 Chr.17: 50462269-50462269 on Build GRCh38
rs921752135 Chr.17: 50462248-50462248 on Build GRCh38
rs980450421 Chr.17: 50462405-50462405 on Build GRCh38
rs754072414 Chr.17: 50462303-50462303 on Build GRCh38
rs966359002 Chr.17: 50462209-50462209 on Build GRCh38
rs773684392 Chr.17: 50462446-50462446 on Build GRCh38
rs755639012 Chr.17: 50462347-50462347 on Build GRCh38
rs200973283 Chr.17: 50462286-50462286 on Build GRCh38
rs768663196 Chr.17: 50462509-50462509 on Build GRCh38
rs991889499 Chr.17: 50462151-50462151 on Build GRCh38
rs371431770 Chr.17: 50462135-50462135 on Build GRCh38
rs2277631 Chr.17: 50462402-50462402 on Build GRCh38
rs891163835 Chr.17: 50462330-50462330 on Build GRCh38
rs200245405 Chr.17: 50462270-50462270 on Build GRCh38
rs762128850 Chr.17: 50462309-50462309 on Build GRCh38
rs150674486 Chr.17: 50462564-50462564 on Build GRCh38
rs201746826 Chr.17: 50462322-50462322 on Build GRCh38
rs774143302 Chr.17: 50462266-50462266 on Build GRCh38
rs758938078 Chr.17: 50462324-50462324 on Build GRCh38
rs1039951207 Chr.17: 50462231-50462231 on Build GRCh38
rs777479211 Chr.17: 50462388-50462388 on Build GRCh38
rs201927915 Chr.17: 50462168-50462168 on Build GRCh38
rs760910663 Chr.17: 50462206-50462206 on Build GRCh38
rs202105387 Chr.17: 50462542-50462542 on Build GRCh38
rs776012354 Chr.17: 50462147-50462147 on Build GRCh38
rs773627017 Chr.17: 50462522-50462522 on Build GRCh38
rs777340533 Chr.17: 50462257-50462257 on Build GRCh38
rs1016112966 Chr.17: 50462391-50462391 on Build GRCh38
rs201149532 Chr.17: 50462271-50462271 on Build GRCh38
rs940037000 Chr.17: 50462331-50462331 on Build GRCh38
rs753322943 Chr.17: 50462549-50462549 on Build GRCh38
rs1037404482 Chr.17: 50462371-50462371 on Build GRCh38
rs752074448 Chr.17: 50462467-50462467 on Build GRCh38
rs543097710 Chr.17: 50462406-50462406 on Build GRCh38
rs887208807 Chr.17: 50462249-50462249 on Build GRCh38
rs560515941 Chr.17: 50462366-50462366 on Build GRCh38
rs766896767 Chr.17: 50462465-50462465 on Build GRCh38
rs765591411 Chr.17: 50462434-50462434 on Build GRCh38
rs769457997 Chr.17: 50462255-50462255 on Build GRCh38
rs141937528 Chr.17: 50462528-50462528 on Build GRCh38
rs147280032 Chr.17: 50462487-50462487 on Build GRCh38
rs531790351 Chr.17: 50462424-50462424 on Build GRCh38
rs747647647 Chr.17: 50462254-50462254 on Build GRCh38
rs756817776 Chr.17: 50462559-50462559 on Build GRCh38
rs199678954 Chr.17: 50462494-50462494 on Build GRCh38
rs754416313 Chr.17: 50462414-50462414 on Build GRCh38
rs887073462 Chr.17: 50462570-50462570 on Build GRCh38
rs867308871 Chr.17: 50462563-50462563 on Build GRCh38
rs773024266 Chr.17: 50462150-50462150 on Build GRCh38
rs372856581 Chr.17: 50462488-50462488 on Build GRCh38
rs757039909 Chr.17: 50462246-50462246 on Build GRCh38
rs995696513 Chr.17: 50462124-50462124 on Build GRCh38
rs117733857 Chr.17: 50462172-50462172 on Build GRCh38
rs748875428 Chr.17: 50462389-50462389 on Build GRCh38
rs201428015 Chr.17: 50462508-50462508 on Build GRCh38
rs370871048 Chr.17: 50462551-50462551 on Build GRCh38
rs1034232402 Chr.17: 50462126-50462126 on Build GRCh38
rs978475846 Chr.17: 50462476-50462476 on Build GRCh38
rs34091038 Chr.17: 50462315-50462316 on Build GRCh38
rs776815146 Chr.17: 50462517-50462517 on Build GRCh38
rs138434188 Chr.17: 50462521-50462521 on Build GRCh38
rs780707588 Chr.17: 50462241-50462241 on Build GRCh38
rs779554757 Chr.17: 50462224-50462224 on Build GRCh38
rs112547536 Chr.17: 50462527-50462527 on Build GRCh38
rs780469835 Chr.17: 50462343-50462343 on Build GRCh38
rs139906230 Chr.17: 50462573-50462573 on Build GRCh38
rs774332997 Chr.17: 50462171-50462171 on Build GRCh38
rs745644750 Chr.17: 50462495-50462495 on Build GRCh38
rs769358469 Chr.17: 50462148-50462148 on Build GRCh38
rs754988573 Chr.17: 50462421-50462421 on Build GRCh38
rs763371922 Chr.17: 50462526-50462526 on Build GRCh38
rs750843061 Chr.17: 50462323-50462323 on Build GRCh38
rs762777116 Chr.17: 50462154-50462154 on Build GRCh38
rs768549107 Chr.17: 50462423-50462423 on Build GRCh38
rs572336359 Chr.17: 50462284-50462284 on Build GRCh38
rs147483243 Chr.17: 50462497-50462497 on Build GRCh38
rs778638235 Chr.17: 50462395-50462395 on Build GRCh38
rs764360875 Chr.17: 50462287-50462287 on Build GRCh38
rs762080134 Chr.17: 50462427-50462427 on Build GRCh38
rs375794969 Chr.17: 50462531-50462531 on Build GRCh38
rs528153231 Chr.17: 50462520-50462520 on Build GRCh38
rs750891816 Chr.17: 50462233-50462233 on Build GRCh38
rs764263233 Chr.17: 50462207-50462207 on Build GRCh38
rs192023009 Chr.17: 50462409-50462409 on Build GRCh38
rs752230148 Chr.17: 50462344-50462344 on Build GRCh38
rs749071712 Chr.17: 50462260-50462260 on Build GRCh38
rs754204685 Chr.17: 50462213-50462213 on Build GRCh38
rs954544068 Chr.17: 50462562-50462562 on Build GRCh38
rs767525197 Chr.17: 50462189-50462189 on Build GRCh38
rs138994578 Chr.17: 50462447-50462447 on Build GRCh38
rs143964709 Chr.17: 50462240-50462240 on Build GRCh38
rs770590610 Chr.17: 50462390-50462390 on Build GRCh38
rs779900817 Chr.17: 50462499-50462499 on Build GRCh38
rs752740830 Chr.17: 50462204-50462204 on Build GRCh38
rs755514814 Chr.17: 50462483-50462483 on Build GRCh38
rs756870862 Chr.17: 50462490-50462490 on Build GRCh38
rs770780869 Chr.17: 50462261-50462261 on Build GRCh38

View Full Product Details