Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58541500-58542006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf47
Entrez Gene ID
284083
Gene Name
chromosome 17 open reading frame 47
Gene Aliases
-
UniGene
Hs.646574
Gene Chromosome Location
Chr.17: 58541587-58544372 on Build GRCh38
Transcripts
NM_001038704.2 XM_011524637.2 XM_017024498.1 NR_110810.1

Gene Symbol
SEPT4-AS1
Entrez Gene ID
101927688
Gene Name
SEPT4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660034
Gene Chromosome Location
Chr.17: 58519837-58556925 on Build GRCh38
Transcripts
NM_001038704.2 XM_011524637.2 XM_017024498.1 NR_110810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6032990 c.1562-7C>T p.? Chr.17: 58541973-58541973 on Build GRCh38
1630305 c.1586G>A p.R529H Chr.17: 58541942-58541942 on Build GRCh38
473112 c.1663G>T p.G555C Chr.17: 58541865-58541865 on Build GRCh38
6614645 c.1670A>G p.E557G Chr.17: 58541858-58541858 on Build GRCh38
6213086 c.1609_1610insT p.C538fs*19 Chr.17: 58541918-58541919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892109736 Chr.17: 58541604-58541604 on Build GRCh38
rs754010302 Chr.17: 58541937-58541937 on Build GRCh38
rs751986586 Chr.17: 58541963-58541963 on Build GRCh38
rs1010215943 Chr.17: 58541828-58541828 on Build GRCh38
rs748853946 Chr.17: 58541846-58541846 on Build GRCh38
rs777533218 Chr.17: 58541844-58541844 on Build GRCh38
rs892217435 Chr.17: 58541907-58541907 on Build GRCh38
rs545290641 Chr.17: 58541533-58541533 on Build GRCh38
rs951327242 Chr.17: 58541728-58541728 on Build GRCh38
rs747385361 Chr.17: 58541836-58541836 on Build GRCh38
rs760866839 Chr.17: 58541786-58541786 on Build GRCh38
rs779419204 Chr.17: 58541813-58541813 on Build GRCh38
rs753960646 Chr.17: 58541789-58541789 on Build GRCh38
rs770512502 Chr.17: 58541855-58541855 on Build GRCh38
rs1024937929 Chr.17: 58541627-58541627 on Build GRCh38
rs188958081 Chr.17: 58541556-58541556 on Build GRCh38
rs988867786 Chr.17: 58541533-58541534 on Build GRCh38
rs757306154 Chr.17: 58541799-58541799 on Build GRCh38
rs184956913 Chr.17: 58541783-58541783 on Build GRCh38
rs550504476 Chr.17: 58541782-58541782 on Build GRCh38
rs767154507 Chr.17: 58541769-58541769 on Build GRCh38
rs576889733 Chr.17: 58541943-58541943 on Build GRCh38
rs771944448 Chr.17: 58541972-58541972 on Build GRCh38
rs912593290 Chr.17: 58541802-58541802 on Build GRCh38
rs368196409 Chr.17: 58541896-58541896 on Build GRCh38
rs750854905 Chr.17: 58541821-58541821 on Build GRCh38
rs755707373 Chr.17: 58541817-58541817 on Build GRCh38
rs1034503144 Chr.17: 58541690-58541690 on Build GRCh38
rs563881629 Chr.17: 58541542-58541542 on Build GRCh38
rs758831245 Chr.17: 58541975-58541975 on Build GRCh38
rs533801946 Chr.17: 58541909-58541909 on Build GRCh38
rs774354204 Chr.17: 58541859-58541859 on Build GRCh38
rs554947137 Chr.17: 58541864-58541864 on Build GRCh38
rs1013257626 Chr.17: 58541626-58541626 on Build GRCh38
rs376283033 Chr.17: 58541970-58541970 on Build GRCh38
rs546950988 Chr.17: 58541700-58541700 on Build GRCh38
rs748886952 Chr.17: 58541960-58541960 on Build GRCh38
rs765635349 Chr.17: 58541726-58541726 on Build GRCh38
rs201771232 Chr.17: 58541847-58541847 on Build GRCh38
rs181185998 Chr.17: 58541705-58541705 on Build GRCh38
rs761912661 Chr.17: 58541940-58541940 on Build GRCh38
rs1047416466 Chr.17: 58541695-58541695 on Build GRCh38
rs755368105 Chr.17: 58541948-58541948 on Build GRCh38
rs979715107 Chr.17: 58541558-58541558 on Build GRCh38
rs1055012157 Chr.17: 58541595-58541595 on Build GRCh38
rs371706623 Chr.17: 58541942-58541942 on Build GRCh38
rs777586281 Chr.17: 58541950-58541950 on Build GRCh38
rs769635443 Chr.17: 58541842-58541842 on Build GRCh38
rs779503642 Chr.17: 58541915-58541916 on Build GRCh38
rs746382966 Chr.17: 58541965-58541966 on Build GRCh38
rs1002225567 Chr.17: 58541683-58541683 on Build GRCh38
rs558490844 Chr.17: 58541927-58541927 on Build GRCh38
rs141193307 Chr.17: 58541827-58541827 on Build GRCh38
rs751906832 Chr.17: 58541945-58541945 on Build GRCh38
rs936528995 Chr.17: 58541869-58541869 on Build GRCh38
rs775478208 Chr.17: 58541980-58541980 on Build GRCh38
rs539093852 Chr.17: 58541749-58541749 on Build GRCh38
rs752107010 Chr.17: 58541798-58541798 on Build GRCh38
rs139756375 Chr.17: 58541944-58541944 on Build GRCh38
rs16943091 Chr.17: 58541892-58541892 on Build GRCh38
rs780224433 Chr.17: 58541830-58541830 on Build GRCh38
rs991158364 Chr.17: 58541965-58541965 on Build GRCh38
rs528513968 Chr.17: 58541658-58541658 on Build GRCh38
rs561237949 Chr.17: 58541575-58541575 on Build GRCh38
rs949004959 Chr.17: 58541653-58541653 on Build GRCh38
rs1052160860 Chr.17: 58541578-58541578 on Build GRCh38
rs148229730 Chr.17: 58541561-58541561 on Build GRCh38
rs760488667 Chr.17: 58541914-58541914 on Build GRCh38
rs761574969 Chr.17: 58541714-58541714 on Build GRCh38
rs750885770 Chr.17: 58541809-58541809 on Build GRCh38
rs778348497 Chr.17: 58541966-58541966 on Build GRCh38

View Full Product Details