Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58544126-58544629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf47
Entrez Gene ID
284083
Gene Name
chromosome 17 open reading frame 47
Gene Aliases
-
UniGene
Hs.646574
Gene Chromosome Location
Chr.17: 58541587-58544372 on Build GRCh38
Transcripts
NM_001038704.2 XM_011524637.2 XM_017024498.1 NR_110810.1

Gene Symbol
SEPT4-AS1
Entrez Gene ID
101927688
Gene Name
SEPT4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660034
Gene Chromosome Location
Chr.17: 58519837-58556925 on Build GRCh38
Transcripts
NM_001038704.2 XM_011524637.2 XM_017024498.1 NR_110810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5036934 c.15T>G p.N5K Chr.17: 58544172-58544172 on Build GRCh38
6147063 c.50G>C p.R17T Chr.17: 58544137-58544137 on Build GRCh38
3195862 c.44C>A p.A15E Chr.17: 58544143-58544143 on Build GRCh38
3742344 c.9G>A p.K3K Chr.17: 58544178-58544178 on Build GRCh38
4188644 c.40T>A p.S14T Chr.17: 58544147-58544147 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188375864 Chr.17: 58544294-58544294 on Build GRCh38
rs761453119 Chr.17: 58544439-58544441 on Build GRCh38
rs940542792 Chr.17: 58544296-58544296 on Build GRCh38
rs201743249 Chr.17: 58544240-58544240 on Build GRCh38
rs867387868 Chr.17: 58544316-58544316 on Build GRCh38
rs775813794 Chr.17: 58544153-58544153 on Build GRCh38
rs1022516955 Chr.17: 58544519-58544519 on Build GRCh38
rs182919893 Chr.17: 58544255-58544255 on Build GRCh38
rs773887873 Chr.17: 58544184-58544184 on Build GRCh38
rs764041612 Chr.17: 58544209-58544209 on Build GRCh38
rs1040162402 Chr.17: 58544334-58544337 on Build GRCh38
rs747434415 Chr.17: 58544154-58544154 on Build GRCh38
rs961966971 Chr.17: 58544586-58544586 on Build GRCh38
rs9894430 Chr.17: 58544501-58544501 on Build GRCh38
rs768833520 Chr.17: 58544155-58544155 on Build GRCh38
rs779255240 Chr.17: 58544227-58544227 on Build GRCh38
rs901590531 Chr.17: 58544364-58544364 on Build GRCh38
rs766015575 Chr.17: 58544177-58544177 on Build GRCh38
rs892827976 Chr.17: 58544419-58544419 on Build GRCh38
rs114669726 Chr.17: 58544158-58544158 on Build GRCh38
rs930526625 Chr.17: 58544200-58544200 on Build GRCh38
rs1035638779 Chr.17: 58544541-58544541 on Build GRCh38
rs892762728 Chr.17: 58544469-58544469 on Build GRCh38
rs924569924 Chr.17: 58544362-58544362 on Build GRCh38
rs1003067105 Chr.17: 58544568-58544568 on Build GRCh38
rs1040487603 Chr.17: 58544330-58544330 on Build GRCh38
rs368904477 Chr.17: 58544196-58544196 on Build GRCh38
rs967247511 Chr.17: 58544524-58544524 on Build GRCh38
rs944569160 Chr.17: 58544315-58544315 on Build GRCh38
rs916559192 Chr.17: 58544588-58544588 on Build GRCh38
rs1054012808 Chr.17: 58544242-58544242 on Build GRCh38
rs978288115 Chr.17: 58544540-58544540 on Build GRCh38
rs990924131 Chr.17: 58544566-58544566 on Build GRCh38
rs1011091244 Chr.17: 58544433-58544433 on Build GRCh38
rs547137293 Chr.17: 58544383-58544383 on Build GRCh38
rs757597086 Chr.17: 58544218-58544218 on Build GRCh38
rs910397562 Chr.17: 58544308-58544308 on Build GRCh38
rs368473165 Chr.17: 58544190-58544190 on Build GRCh38
rs1028603185 Chr.17: 58544410-58544410 on Build GRCh38
rs772862732 Chr.17: 58544156-58544156 on Build GRCh38
rs1015769617 Chr.17: 58544575-58544575 on Build GRCh38
rs886241956 Chr.17: 58544283-58544283 on Build GRCh38
rs190772133 Chr.17: 58544565-58544565 on Build GRCh38
rs148000539 Chr.17: 58544314-58544315 on Build GRCh38
rs767428778 Chr.17: 58544561-58544561 on Build GRCh38
rs142914239 Chr.17: 58544151-58544151 on Build GRCh38
rs373637384 Chr.17: 58544152-58544152 on Build GRCh38
rs753685230 Chr.17: 58544217-58544217 on Build GRCh38
rs759024358 Chr.17: 58544187-58544188 on Build GRCh38
rs542243827 Chr.17: 58544487-58544487 on Build GRCh38
rs148960491 Chr.17: 58544492-58544492 on Build GRCh38
rs750954817 Chr.17: 58544169-58544172 on Build GRCh38
rs372204884 Chr.17: 58544201-58544201 on Build GRCh38
rs3217043 Chr.17: 58544554-58544554 on Build GRCh38

View Full Product Details