Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37865473-37866011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5B
Entrez Gene ID
3633
Gene Name
inositol polyphosphate-5-phosphatase B
Gene Aliases
5PTase
UniGene
Hs.449942
Gene Chromosome Location
Chr.1: 37860697-37947094 on Build GRCh38
Transcripts
NM_001297434.1 NM_005540.2 XM_006710621.3 XM_011541392.2 XM_011541393.2 XM_017001207.1 XM_017001208.1 XM_017001209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6126324 c.2671G>T p.E891* Chr.1: 37865844-37865844 on Build GRCh38
4635917 c.2722G>C p.E908Q Chr.1: 37865793-37865793 on Build GRCh38
6312665 c.2654C>A p.A885D Chr.1: 37865861-37865861 on Build GRCh38
3489280 c.2658G>A p.L886L Chr.1: 37865857-37865857 on Build GRCh38
3489279 c.2667C>T p.F889F Chr.1: 37865848-37865848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016635484 Chr.1: 37865628-37865628 on Build GRCh38
rs35888275 Chr.1: 37865893-37865894 on Build GRCh38
rs199676247 Chr.1: 37865786-37865786 on Build GRCh38
rs991408220 Chr.1: 37865712-37865712 on Build GRCh38
rs910591046 Chr.1: 37865836-37865836 on Build GRCh38
rs773564879 Chr.1: 37865742-37865742 on Build GRCh38
rs763259527 Chr.1: 37865876-37865876 on Build GRCh38
rs757744184 Chr.1: 37865848-37865849 on Build GRCh38
rs753717967 Chr.1: 37865919-37865919 on Build GRCh38
rs767001325 Chr.1: 37865896-37865896 on Build GRCh38
rs994140625 Chr.1: 37865942-37865942 on Build GRCh38
rs768669533 Chr.1: 37865842-37865842 on Build GRCh38
rs752236380 Chr.1: 37865912-37865912 on Build GRCh38
rs765946094 Chr.1: 37865756-37865756 on Build GRCh38
rs535289020 Chr.1: 37865831-37865831 on Build GRCh38
rs752616092 Chr.1: 37865796-37865796 on Build GRCh38
rs748673634 Chr.1: 37865864-37865864 on Build GRCh38
rs145271992 Chr.1: 37865716-37865716 on Build GRCh38
rs917137918 Chr.1: 37865718-37865718 on Build GRCh38
rs970834799 Chr.1: 37865959-37865959 on Build GRCh38
rs777487082 Chr.1: 37865811-37865811 on Build GRCh38
rs958357988 Chr.1: 37865706-37865706 on Build GRCh38
rs552561325 Chr.1: 37865546-37865546 on Build GRCh38
rs1047923260 Chr.1: 37865958-37865958 on Build GRCh38
rs376188578 Chr.1: 37865513-37865513 on Build GRCh38
rs755205085 Chr.1: 37865810-37865810 on Build GRCh38
rs139225475 Chr.1: 37865569-37865569 on Build GRCh38
rs758700003 Chr.1: 37865823-37865823 on Build GRCh38
rs757069566 Chr.1: 37865930-37865930 on Build GRCh38
rs778830172 Chr.1: 37865936-37865936 on Build GRCh38
rs186262482 Chr.1: 37865620-37865620 on Build GRCh38
rs780383560 Chr.1: 37865826-37865826 on Build GRCh38
rs200324778 Chr.1: 37865937-37865937 on Build GRCh38
rs774037773 Chr.1: 37865892-37865892 on Build GRCh38
rs900984720 Chr.1: 37865626-37865626 on Build GRCh38
rs756072632 Chr.1: 37865800-37865800 on Build GRCh38
rs943333016 Chr.1: 37865865-37865865 on Build GRCh38
rs373731178 Chr.1: 37865913-37865913 on Build GRCh38
rs773583571 Chr.1: 37865869-37865869 on Build GRCh38
rs764475169 Chr.1: 37865712-37865712 on Build GRCh38
rs929800046 Chr.1: 37865684-37865684 on Build GRCh38
rs759193579 Chr.1: 37865895-37865895 on Build GRCh38
rs754433375 Chr.1: 37865776-37865776 on Build GRCh38
rs978263001 Chr.1: 37865978-37865978 on Build GRCh38
rs989200834 Chr.1: 37865584-37865584 on Build GRCh38
rs762656117 Chr.1: 37865743-37865743 on Build GRCh38
rs781303898 Chr.1: 37865843-37865843 on Build GRCh38
rs779352819 Chr.1: 37865672-37865672 on Build GRCh38
rs1025015443 Chr.1: 37865920-37865920 on Build GRCh38
rs769237381 Chr.1: 37865715-37865715 on Build GRCh38
rs761657105 Chr.1: 37865690-37865690 on Build GRCh38
rs910943325 Chr.1: 37865609-37865609 on Build GRCh38
rs574709112 Chr.1: 37865735-37865735 on Build GRCh38
rs376296305 Chr.1: 37865866-37865866 on Build GRCh38
rs942493633 Chr.1: 37865612-37865612 on Build GRCh38
rs778785114 Chr.1: 37865629-37865630 on Build GRCh38
rs369532196 Chr.1: 37865877-37865877 on Build GRCh38
rs902020201 Chr.1: 37865542-37865542 on Build GRCh38
rs1006634750 Chr.1: 37865863-37865863 on Build GRCh38
rs764055861 Chr.1: 37865914-37865914 on Build GRCh38
rs537589796 Chr.1: 37865538-37865538 on Build GRCh38
rs781226511 Chr.1: 37865975-37865975 on Build GRCh38
rs936128173 Chr.1: 37865815-37865815 on Build GRCh38
rs755227301 Chr.1: 37865938-37865938 on Build GRCh38
rs762360490 Chr.1: 37865717-37865717 on Build GRCh38
rs754222392 Chr.1: 37865815-37865816 on Build GRCh38
rs749033817 Chr.1: 37865814-37865814 on Build GRCh38

View Full Product Details