Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37868389-37868849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5B
Entrez Gene ID
3633
Gene Name
inositol polyphosphate-5-phosphatase B
Gene Aliases
5PTase
UniGene
Hs.449942
Gene Chromosome Location
Chr.1: 37860697-37947094 on Build GRCh38
Transcripts
NM_001297434.1 NM_005540.2 XM_006710621.3 XM_011541392.2 XM_011541393.2 XM_017001206.1 XM_017001207.1 XM_017001208.1 XM_017001209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6126323 c.2494G>C p.E832Q Chr.1: 37868548-37868548 on Build GRCh38
358101 c.2466G>T p.Q822H Chr.1: 37868576-37868576 on Build GRCh38
7268641 c.2490delC p.E832fs*4 Chr.1: 37868552-37868552 on Build GRCh38
6867454 c.2469G>T p.L823F Chr.1: 37868573-37868573 on Build GRCh38
3419197 c.2525G>A p.R842Q Chr.1: 37868517-37868517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575969418 Chr.1: 37868648-37868648 on Build GRCh38
rs558356074 Chr.1: 37868740-37868740 on Build GRCh38
rs775160931 Chr.1: 37868496-37868496 on Build GRCh38
rs866467843 Chr.1: 37868592-37868592 on Build GRCh38
rs67006214 Chr.1: 37868646-37868646 on Build GRCh38
rs375670508 Chr.1: 37868618-37868618 on Build GRCh38
rs747427663 Chr.1: 37868771-37868771 on Build GRCh38
rs996055800 Chr.1: 37868435-37868435 on Build GRCh38
rs2306426 Chr.1: 37868646-37868646 on Build GRCh38
rs772891704 Chr.1: 37868534-37868534 on Build GRCh38
rs1052946602 Chr.1: 37868811-37868811 on Build GRCh38
rs746743486 Chr.1: 37868635-37868635 on Build GRCh38
rs753855705 Chr.1: 37868585-37868585 on Build GRCh38
rs369913128 Chr.1: 37868481-37868481 on Build GRCh38
rs912966259 Chr.1: 37868712-37868712 on Build GRCh38
rs761903907 Chr.1: 37868517-37868517 on Build GRCh38
rs753001007 Chr.1: 37868558-37868558 on Build GRCh38
rs939484987 Chr.1: 37868805-37868805 on Build GRCh38
rs866564043 Chr.1: 37868518-37868518 on Build GRCh38
rs746241504 Chr.1: 37868601-37868601 on Build GRCh38
rs961522649 Chr.1: 37868557-37868557 on Build GRCh38
rs927774196 Chr.1: 37868787-37868787 on Build GRCh38
rs768413196 Chr.1: 37868643-37868643 on Build GRCh38
rs771653758 Chr.1: 37868491-37868491 on Build GRCh38
rs1043201680 Chr.1: 37868739-37868739 on Build GRCh38
rs765210866 Chr.1: 37868533-37868533 on Build GRCh38
rs780252910 Chr.1: 37868625-37868625 on Build GRCh38
rs764214869 Chr.1: 37868575-37868575 on Build GRCh38
rs766061567 Chr.1: 37868552-37868552 on Build GRCh38
rs113891260 Chr.1: 37868419-37868419 on Build GRCh38
rs775705986 Chr.1: 37868578-37868582 on Build GRCh38
rs386630429 Chr.1: 37868645-37868646 on Build GRCh38
rs757765937 Chr.1: 37868599-37868599 on Build GRCh38
rs1030272372 Chr.1: 37868461-37868461 on Build GRCh38
rs765355687 Chr.1: 37868756-37868758 on Build GRCh38
rs775892299 Chr.1: 37868703-37868703 on Build GRCh38
rs779260469 Chr.1: 37868476-37868476 on Build GRCh38
rs766077707 Chr.1: 37868676-37868676 on Build GRCh38
rs892456112 Chr.1: 37868462-37868462 on Build GRCh38
rs762773258 Chr.1: 37868546-37868546 on Build GRCh38
rs372406929 Chr.1: 37868512-37868512 on Build GRCh38
rs947123972 Chr.1: 37868729-37868729 on Build GRCh38
rs1043916514 Chr.1: 37868484-37868484 on Build GRCh38
rs540140382 Chr.1: 37868730-37868730 on Build GRCh38
rs773247047 Chr.1: 37868652-37868652 on Build GRCh38
rs746619799 Chr.1: 37868509-37868509 on Build GRCh38
rs199558961 Chr.1: 37868510-37868510 on Build GRCh38
rs891102753 Chr.1: 37868506-37868506 on Build GRCh38
rs756321392 Chr.1: 37868563-37868563 on Build GRCh38
rs1042931082 Chr.1: 37868415-37868415 on Build GRCh38
rs921764460 Chr.1: 37868741-37868741 on Build GRCh38
rs988535612 Chr.1: 37868710-37868710 on Build GRCh38
rs757747530 Chr.1: 37868458-37868458 on Build GRCh38
rs2306425 Chr.1: 37868645-37868645 on Build GRCh38
rs1016075837 Chr.1: 37868535-37868535 on Build GRCh38

View Full Product Details