Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37875218-37875759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5B
Entrez Gene ID
3633
Gene Name
inositol polyphosphate-5-phosphatase B
Gene Aliases
5PTase
UniGene
Hs.449942
Gene Chromosome Location
Chr.1: 37860697-37947094 on Build GRCh38
Transcripts
NM_001297434.1 NM_005540.2 XM_006710621.3 XM_011541392.2 XM_011541393.2 XM_017001206.1 XM_017001207.1 XM_017001208.1 XM_017001209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7422982 c.1983G>C p.K661N Chr.1: 37875651-37875651 on Build GRCh38
321019 c.1970G>T p.R657L Chr.1: 37875664-37875664 on Build GRCh38
5992853 c.1918-10T>C p.? Chr.1: 37875726-37875726 on Build GRCh38
6989029 c.2024G>A p.R675Q Chr.1: 37875610-37875610 on Build GRCh38
426219 c.1975C>G p.L659V Chr.1: 37875659-37875659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769139417 Chr.1: 37875557-37875557 on Build GRCh38
rs946852819 Chr.1: 37875516-37875516 on Build GRCh38
rs573827440 Chr.1: 37875400-37875400 on Build GRCh38
rs1049835728 Chr.1: 37875292-37875292 on Build GRCh38
rs980214586 Chr.1: 37875454-37875454 on Build GRCh38
rs1008319155 Chr.1: 37875308-37875308 on Build GRCh38
rs767442521 Chr.1: 37875607-37875607 on Build GRCh38
rs748620190 Chr.1: 37875649-37875649 on Build GRCh38
rs897316257 Chr.1: 37875723-37875723 on Build GRCh38
rs973525265 Chr.1: 37875338-37875338 on Build GRCh38
rs370747244 Chr.1: 37875564-37875564 on Build GRCh38
rs374018602 Chr.1: 37875568-37875568 on Build GRCh38
rs756014498 Chr.1: 37875610-37875610 on Build GRCh38
rs768734100 Chr.1: 37875326-37875326 on Build GRCh38
rs1050159743 Chr.1: 37875637-37875637 on Build GRCh38
rs776872175 Chr.1: 37875559-37875559 on Build GRCh38
rs550313044 Chr.1: 37875363-37875363 on Build GRCh38
rs539109150 Chr.1: 37875548-37875548 on Build GRCh38
rs377554015 Chr.1: 37875701-37875701 on Build GRCh38
rs1052605263 Chr.1: 37875481-37875481 on Build GRCh38
rs377641126 Chr.1: 37875576-37875576 on Build GRCh38
rs759049604 Chr.1: 37875689-37875689 on Build GRCh38
rs575037660 Chr.1: 37875707-37875707 on Build GRCh38
rs769064205 Chr.1: 37875635-37875635 on Build GRCh38
rs914180515 Chr.1: 37875738-37875738 on Build GRCh38
rs892649560 Chr.1: 37875500-37875500 on Build GRCh38
rs995930526 Chr.1: 37875554-37875554 on Build GRCh38
rs764047712 Chr.1: 37875708-37875708 on Build GRCh38
rs185568405 Chr.1: 37875254-37875254 on Build GRCh38
rs961837770 Chr.1: 37875332-37875332 on Build GRCh38
rs190478359 Chr.1: 37875426-37875426 on Build GRCh38
rs748708214 Chr.1: 37875731-37875731 on Build GRCh38
rs3748804 Chr.1: 37875291-37875291 on Build GRCh38
rs750732621 Chr.1: 37875621-37875621 on Build GRCh38
rs751019223 Chr.1: 37875587-37875587 on Build GRCh38
rs920651660 Chr.1: 37875343-37875343 on Build GRCh38
rs747535557 Chr.1: 37875556-37875556 on Build GRCh38
rs146846053 Chr.1: 37875347-37875347 on Build GRCh38
rs1043009590 Chr.1: 37875249-37875249 on Build GRCh38
rs766573465 Chr.1: 37875714-37875714 on Build GRCh38
rs533782005 Chr.1: 37875636-37875636 on Build GRCh38
rs903845386 Chr.1: 37875286-37875286 on Build GRCh38
rs771042257 Chr.1: 37875605-37875605 on Build GRCh38
rs780003329 Chr.1: 37875394-37875394 on Build GRCh38
rs770189923 Chr.1: 37875660-37875660 on Build GRCh38
rs777690297 Chr.1: 37875619-37875619 on Build GRCh38
rs1043884086 Chr.1: 37875529-37875529 on Build GRCh38
rs980841017 Chr.1: 37875698-37875698 on Build GRCh38
rs780001724 Chr.1: 37875628-37875628 on Build GRCh38
rs765860634 Chr.1: 37875579-37875579 on Build GRCh38
rs758689721 Chr.1: 37875626-37875626 on Build GRCh38
rs146816630 Chr.1: 37875276-37875277 on Build GRCh38
rs954642938 Chr.1: 37875276-37875276 on Build GRCh38
rs371113985 Chr.1: 37875664-37875664 on Build GRCh38
rs755123159 Chr.1: 37875719-37875719 on Build GRCh38
rs891240614 Chr.1: 37875293-37875293 on Build GRCh38
rs753620651 Chr.1: 37875711-37875711 on Build GRCh38
rs762567363 Chr.1: 37875577-37875577 on Build GRCh38
rs960020319 Chr.1: 37875518-37875518 on Build GRCh38
rs1035435991 Chr.1: 37875514-37875514 on Build GRCh38
rs759408466 Chr.1: 37875603-37875603 on Build GRCh38
rs555111201 Chr.1: 37875665-37875665 on Build GRCh38
rs376043866 Chr.1: 37875669-37875669 on Build GRCh38
rs555078900 Chr.1: 37875449-37875449 on Build GRCh38
rs904298468 Chr.1: 37875536-37875536 on Build GRCh38
rs766907538 Chr.1: 37875699-37875699 on Build GRCh38
rs1015673514 Chr.1: 37875336-37875336 on Build GRCh38
rs754745858 Chr.1: 37875505-37875505 on Build GRCh38
rs551043365 Chr.1: 37875563-37875563 on Build GRCh38
rs773760148 Chr.1: 37875688-37875688 on Build GRCh38
rs987381620 Chr.1: 37875303-37875303 on Build GRCh38
rs781107436 Chr.1: 37875728-37875728 on Build GRCh38
rs891129877 Chr.1: 37875676-37875676 on Build GRCh38
rs751620300 Chr.1: 37875717-37875717 on Build GRCh38
rs961890609 Chr.1: 37875602-37875602 on Build GRCh38
rs747052210 Chr.1: 37875633-37875633 on Build GRCh38

View Full Product Details