Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37882728-37883234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5B
Entrez Gene ID
3633
Gene Name
inositol polyphosphate-5-phosphatase B
Gene Aliases
5PTase
UniGene
Hs.449942
Gene Chromosome Location
Chr.1: 37860697-37947094 on Build GRCh38
Transcripts
NM_001297434.1 NM_005540.2 XM_006710621.3 XM_011541392.2 XM_011541393.2 XM_017001206.1 XM_017001207.1 XM_017001208.1 XM_017001209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2167667 c.1638G>T p.E546D Chr.1: 37882840-37882840 on Build GRCh38
6686106 c.1625delA p.K542fs*17 Chr.1: 37882853-37882853 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748383314 Chr.1: 37882862-37882862 on Build GRCh38
rs539406675 Chr.1: 37882791-37882791 on Build GRCh38
rs756308701 Chr.1: 37882970-37882970 on Build GRCh38
rs1002241201 Chr.1: 37883022-37883022 on Build GRCh38
rs79609816 Chr.1: 37883093-37883093 on Build GRCh38
rs925303911 Chr.1: 37883049-37883049 on Build GRCh38
rs969335751 Chr.1: 37882827-37882827 on Build GRCh38
rs775051493 Chr.1: 37882913-37882913 on Build GRCh38
rs895643149 Chr.1: 37883201-37883201 on Build GRCh38
rs897372255 Chr.1: 37883107-37883107 on Build GRCh38
rs557487867 Chr.1: 37882976-37882976 on Build GRCh38
rs947580268 Chr.1: 37882958-37882958 on Build GRCh38
rs2279262 Chr.1: 37883038-37883038 on Build GRCh38
rs911846478 Chr.1: 37882960-37882960 on Build GRCh38
rs375174033 Chr.1: 37882964-37882964 on Build GRCh38
rs753289611 Chr.1: 37882823-37882823 on Build GRCh38
rs1001272448 Chr.1: 37883196-37883196 on Build GRCh38
rs757122115 Chr.1: 37882757-37882757 on Build GRCh38
rs749673005 Chr.1: 37883080-37883080 on Build GRCh38
rs568657228 Chr.1: 37882790-37882790 on Build GRCh38
rs201441741 Chr.1: 37882845-37882845 on Build GRCh38
rs373523453 Chr.1: 37882836-37882836 on Build GRCh38
rs763753917 Chr.1: 37882932-37882932 on Build GRCh38
rs745714523 Chr.1: 37882898-37882899 on Build GRCh38
rs778009368 Chr.1: 37882895-37882895 on Build GRCh38
rs760022941 Chr.1: 37882808-37882808 on Build GRCh38
rs778753592 Chr.1: 37882779-37882779 on Build GRCh38
rs550399102 Chr.1: 37883185-37883185 on Build GRCh38
rs373963229 Chr.1: 37882873-37882873 on Build GRCh38
rs755790002 Chr.1: 37882819-37882819 on Build GRCh38
rs1001303770 Chr.1: 37882971-37882971 on Build GRCh38
rs961236893 Chr.1: 37882869-37882869 on Build GRCh38
rs1044190514 Chr.1: 37882793-37882793 on Build GRCh38
rs566376718 Chr.1: 37882786-37882786 on Build GRCh38
rs528353319 Chr.1: 37883066-37883066 on Build GRCh38
rs925723697 Chr.1: 37883165-37883165 on Build GRCh38
rs772012413 Chr.1: 37882921-37882922 on Build GRCh38
rs376896246 Chr.1: 37882782-37882782 on Build GRCh38
rs979416680 Chr.1: 37883048-37883048 on Build GRCh38
rs914015452 Chr.1: 37882920-37882920 on Build GRCh38
rs368070373 Chr.1: 37882925-37882925 on Build GRCh38
rs147875109 Chr.1: 37882930-37882930 on Build GRCh38
rs147827971 Chr.1: 37883172-37883172 on Build GRCh38
rs561675725 Chr.1: 37883161-37883161 on Build GRCh38
rs761352614 Chr.1: 37882947-37882947 on Build GRCh38
rs114094709 Chr.1: 37883008-37883008 on Build GRCh38
rs78987221 Chr.1: 37882969-37882969 on Build GRCh38
rs768953192 Chr.1: 37882768-37882768 on Build GRCh38
rs915587649 Chr.1: 37882957-37882957 on Build GRCh38
rs1043348697 Chr.1: 37882959-37882959 on Build GRCh38
rs1049158888 Chr.1: 37883018-37883018 on Build GRCh38
rs574719881 Chr.1: 37882905-37882905 on Build GRCh38
rs373774955 Chr.1: 37883034-37883034 on Build GRCh38
rs755177686 Chr.1: 37882892-37882892 on Build GRCh38
rs968186232 Chr.1: 37883134-37883134 on Build GRCh38
rs767906609 Chr.1: 37882814-37882814 on Build GRCh38
rs748378385 Chr.1: 37882780-37882780 on Build GRCh38
rs41307896 Chr.1: 37882834-37882834 on Build GRCh38
rs931561220 Chr.1: 37883128-37883128 on Build GRCh38
rs779884546 Chr.1: 37882852-37882852 on Build GRCh38
rs760077684 Chr.1: 37882922-37882922 on Build GRCh38
rs529086263 Chr.1: 37882760-37882760 on Build GRCh38
rs201236404 Chr.1: 37882898-37882898 on Build GRCh38
rs778105420 Chr.1: 37883070-37883070 on Build GRCh38
rs778692357 Chr.1: 37883200-37883200 on Build GRCh38
rs753378143 Chr.1: 37882934-37882934 on Build GRCh38
rs959884970 Chr.1: 37882975-37882975 on Build GRCh38
rs567929225 Chr.1: 37882758-37882758 on Build GRCh38
rs756716219 Chr.1: 37882974-37882974 on Build GRCh38
rs117123666 Chr.1: 37883054-37883054 on Build GRCh38
rs776757457 Chr.1: 37882773-37882773 on Build GRCh38
rs1037683096 Chr.1: 37883099-37883099 on Build GRCh38
rs960989019 Chr.1: 37883053-37883053 on Build GRCh38
rs189726562 Chr.1: 37882948-37882948 on Build GRCh38
rs905273134 Chr.1: 37883019-37883019 on Build GRCh38
rs752352623 Chr.1: 37882817-37882817 on Build GRCh38
rs749328829 Chr.1: 37882897-37882897 on Build GRCh38
rs985535583 Chr.1: 37883065-37883065 on Build GRCh38
rs757208244 Chr.1: 37882830-37882830 on Build GRCh38
rs149782091 Chr.1: 37882820-37882820 on Build GRCh38
rs978480225 Chr.1: 37882928-37882928 on Build GRCh38
rs774875657 Chr.1: 37882801-37882801 on Build GRCh38
rs897632950 Chr.1: 37883005-37883005 on Build GRCh38
rs979580163 Chr.1: 37883153-37883153 on Build GRCh38

View Full Product Details