Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39625313-39625814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEYL
Entrez Gene ID
26508
Gene Name
hes related family bHLH transcription factor with YRPW motif-like
Gene Aliases
HESR3, HEY3, HRT3, bHLHb33
UniGene
Hs.472566
Gene Chromosome Location
Chr.1: 39623431-39639676 on Build GRCh38
Transcripts
NM_014571.3 XM_005270745.2

SNP Detail

SNP ID Chromosome Location
rs549601918 Chr.1: 39625540-39625540 on Build GRCh38
rs993512141 Chr.1: 39625747-39625747 on Build GRCh38
rs1042152735 Chr.1: 39625347-39625347 on Build GRCh38
rs1010866304 Chr.1: 39625399-39625406 on Build GRCh38
rs536317655 Chr.1: 39625342-39625342 on Build GRCh38
rs1026769472 Chr.1: 39625770-39625770 on Build GRCh38
rs939717328 Chr.1: 39625559-39625559 on Build GRCh38
rs552246769 Chr.1: 39625642-39625642 on Build GRCh38
rs905682328 Chr.1: 39625660-39625660 on Build GRCh38
rs749839999 Chr.1: 39625612-39625612 on Build GRCh38
rs954817971 Chr.1: 39625769-39625769 on Build GRCh38
rs760908899 Chr.1: 39625790-39625790 on Build GRCh38
rs759594991 Chr.1: 39625667-39625667 on Build GRCh38
rs559685317 Chr.1: 39625641-39625641 on Build GRCh38
rs1002716050 Chr.1: 39625410-39625410 on Build GRCh38
rs532688278 Chr.1: 39625594-39625594 on Build GRCh38
rs182773043 Chr.1: 39625343-39625343 on Build GRCh38
rs771391854 Chr.1: 39625758-39625759 on Build GRCh38
rs1029181979 Chr.1: 39625713-39625713 on Build GRCh38
rs937893430 Chr.1: 39625608-39625608 on Build GRCh38
rs11206478 Chr.1: 39625447-39625447 on Build GRCh38
rs147241011 Chr.1: 39625680-39625680 on Build GRCh38
rs1007739955 Chr.1: 39625682-39625682 on Build GRCh38
rs945069388 Chr.1: 39625636-39625636 on Build GRCh38
rs912231052 Chr.1: 39625537-39625537 on Build GRCh38
rs774475980 Chr.1: 39625666-39625666 on Build GRCh38
rs984634727 Chr.1: 39625558-39625558 on Build GRCh38
rs956196233 Chr.1: 39625439-39625439 on Build GRCh38
rs771393922 Chr.1: 39625635-39625635 on Build GRCh38
rs564937424 Chr.1: 39625446-39625446 on Build GRCh38
rs775579059 Chr.1: 39625776-39625776 on Build GRCh38
rs1035673392 Chr.1: 39625433-39625433 on Build GRCh38
rs576115030 Chr.1: 39625353-39625353 on Build GRCh38
rs560538573 Chr.1: 39625344-39625344 on Build GRCh38
rs188708237 Chr.1: 39625645-39625645 on Build GRCh38
rs1019166182 Chr.1: 39625709-39625709 on Build GRCh38
rs1021608771 Chr.1: 39625685-39625685 on Build GRCh38
rs568805127 Chr.1: 39625712-39625712 on Build GRCh38
rs544674309 Chr.1: 39625356-39625356 on Build GRCh38
rs148112227 Chr.1: 39625515-39625515 on Build GRCh38
rs953620522 Chr.1: 39625445-39625445 on Build GRCh38
rs1044222373 Chr.1: 39625652-39625652 on Build GRCh38
rs968828606 Chr.1: 39625467-39625467 on Build GRCh38
rs767575178 Chr.1: 39625696-39625696 on Build GRCh38
rs898264818 Chr.1: 39625613-39625613 on Build GRCh38
rs560722152 Chr.1: 39625519-39625519 on Build GRCh38
rs952151491 Chr.1: 39625780-39625780 on Build GRCh38
rs908161200 Chr.1: 39625779-39625779 on Build GRCh38
rs376278740 Chr.1: 39625582-39625582 on Build GRCh38
rs974930238 Chr.1: 39625484-39625484 on Build GRCh38
rs528765186 Chr.1: 39625661-39625661 on Build GRCh38
rs922079524 Chr.1: 39625507-39625507 on Build GRCh38
rs879044838 Chr.1: 39625341-39625341 on Build GRCh38
rs969977461 Chr.1: 39625378-39625378 on Build GRCh38
rs969235040 Chr.1: 39625695-39625695 on Build GRCh38
rs143934134 Chr.1: 39625535-39625535 on Build GRCh38

View Full Product Details