Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80101397-80101912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAA
Entrez Gene ID
2548
Gene Name
glucosidase alpha, acid
Gene Aliases
LYAG
UniGene
Hs.1437
Gene Chromosome Location
Chr.17: 80101526-80119882 on Build GRCh38
Transcripts
NM_000152.4 NM_001079803.2 NM_001079804.2 NR_134848.1 XM_005257193.2 XM_005257194.4

SNP Detail

SNP ID Chromosome Location
rs544259653 Chr.17: 80101562-80101562 on Build GRCh38
rs547385110 Chr.17: 80101879-80101879 on Build GRCh38
rs900807557 Chr.17: 80101827-80101827 on Build GRCh38
rs969272394 Chr.17: 80101442-80101442 on Build GRCh38
rs1023447167 Chr.17: 80101837-80101837 on Build GRCh38
rs1005312609 Chr.17: 80101631-80101631 on Build GRCh38
rs886053541 Chr.17: 80101687-80101687 on Build GRCh38
rs999193565 Chr.17: 80101788-80101788 on Build GRCh38
rs960427191 Chr.17: 80101819-80101819 on Build GRCh38
rs1055811035 Chr.17: 80101809-80101809 on Build GRCh38
rs939648161 Chr.17: 80101418-80101418 on Build GRCh38
rs1016741188 Chr.17: 80101639-80101639 on Build GRCh38
rs900775636 Chr.17: 80101718-80101718 on Build GRCh38
rs1045840457 Chr.17: 80101572-80101572 on Build GRCh38
rs970649227 Chr.17: 80101473-80101473 on Build GRCh38
rs994001995 Chr.17: 80101883-80101883 on Build GRCh38
rs1046227485 Chr.17: 80101775-80101775 on Build GRCh38
rs527577453 Chr.17: 80101679-80101679 on Build GRCh38
rs1050033501 Chr.17: 80101448-80101449 on Build GRCh38
rs933699991 Chr.17: 80101532-80101532 on Build GRCh38
rs1003854087 Chr.17: 80101476-80101476 on Build GRCh38
rs1010208850 Chr.17: 80101462-80101462 on Build GRCh38
rs955211922 Chr.17: 80101674-80101674 on Build GRCh38
rs978208016 Chr.17: 80101415-80101415 on Build GRCh38
rs972346993 Chr.17: 80101517-80101517 on Build GRCh38
rs190152638 Chr.17: 80101807-80101807 on Build GRCh38
rs772736737 Chr.17: 80101726-80101726 on Build GRCh38
rs1001580052 Chr.17: 80101601-80101601 on Build GRCh38
rs80020206 Chr.17: 80101848-80101848 on Build GRCh38
rs1033973716 Chr.17: 80101606-80101606 on Build GRCh38
rs909599800 Chr.17: 80101513-80101513 on Build GRCh38
rs1800298 Chr.17: 80101841-80101841 on Build GRCh38
rs1050424324 Chr.17: 80101537-80101537 on Build GRCh38
rs959462879 Chr.17: 80101501-80101501 on Build GRCh38
rs936116281 Chr.17: 80101560-80101560 on Build GRCh38
rs1030749054 Chr.17: 80101424-80101424 on Build GRCh38
rs969303535 Chr.17: 80101844-80101844 on Build GRCh38
rs773804868 Chr.17: 80101749-80101749 on Build GRCh38
rs930657076 Chr.17: 80101461-80101461 on Build GRCh38
rs868823899 Chr.17: 80101454-80101454 on Build GRCh38
rs952168973 Chr.17: 80101870-80101870 on Build GRCh38
rs946227791 Chr.17: 80101752-80101752 on Build GRCh38
rs911519006 Chr.17: 80101556-80101556 on Build GRCh38
rs2304849 Chr.17: 80101663-80101663 on Build GRCh38
rs2304850 Chr.17: 80101429-80101429 on Build GRCh38
rs564275180 Chr.17: 80101761-80101761 on Build GRCh38
rs973365355 Chr.17: 80101566-80101577 on Build GRCh38
rs961217909 Chr.17: 80101659-80101659 on Build GRCh38
rs1048311015 Chr.17: 80101463-80101463 on Build GRCh38
rs367731841 Chr.17: 80101861-80101861 on Build GRCh38
rs983920352 Chr.17: 80101507-80101507 on Build GRCh38
rs1043400726 Chr.17: 80101731-80101731 on Build GRCh38
rs557908906 Chr.17: 80101448-80101448 on Build GRCh38
rs1048257025 Chr.17: 80101427-80101427 on Build GRCh38
rs1036663955 Chr.17: 80101425-80101425 on Build GRCh38
rs77514632 Chr.17: 80101745-80101745 on Build GRCh38
rs944897851 Chr.17: 80101698-80101698 on Build GRCh38
rs1013446095 Chr.17: 80101836-80101836 on Build GRCh38
rs1033634476 Chr.17: 80101487-80101487 on Build GRCh38
rs560228296 Chr.17: 80101667-80101667 on Build GRCh38
rs916526687 Chr.17: 80101437-80101437 on Build GRCh38
rs374333139 Chr.17: 80101686-80101686 on Build GRCh38
rs144639114 Chr.17: 80101585-80101585 on Build GRCh38
rs550203457 Chr.17: 80101816-80101816 on Build GRCh38
rs1024325589 Chr.17: 80101465-80101465 on Build GRCh38
rs1040496066 Chr.17: 80101700-80101700 on Build GRCh38
rs980141537 Chr.17: 80101446-80101446 on Build GRCh38
rs918862173 Chr.17: 80101572-80101583 on Build GRCh38
rs972442701 Chr.17: 80101662-80101662 on Build GRCh38
rs576593046 Chr.17: 80101457-80101457 on Build GRCh38
rs574326920 Chr.17: 80101611-80101611 on Build GRCh38
rs934255328 Chr.17: 80101447-80101448 on Build GRCh38
rs907981020 Chr.17: 80101886-80101886 on Build GRCh38
rs1003931341 Chr.17: 80101526-80101526 on Build GRCh38

View Full Product Details