Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80101773-80102273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAA
Entrez Gene ID
2548
Gene Name
glucosidase alpha, acid
Gene Aliases
LYAG
UniGene
Hs.1437
Gene Chromosome Location
Chr.17: 80101526-80119882 on Build GRCh38
Transcripts
NM_000152.4 NM_001079803.2 NM_001079804.2 NR_134848.1 XM_005257193.2 XM_005257194.4

SNP Detail

SNP ID Chromosome Location
rs900807557 Chr.17: 80101827-80101827 on Build GRCh38
rs181336255 Chr.17: 80102037-80102037 on Build GRCh38
rs547385110 Chr.17: 80101879-80101879 on Build GRCh38
rs1023447167 Chr.17: 80101837-80101837 on Build GRCh38
rs897081072 Chr.17: 80102152-80102152 on Build GRCh38
rs1029511272 Chr.17: 80102160-80102160 on Build GRCh38
rs1017959092 Chr.17: 80101920-80101920 on Build GRCh38
rs932171166 Chr.17: 80102161-80102161 on Build GRCh38
rs572564036 Chr.17: 80102177-80102177 on Build GRCh38
rs960427191 Chr.17: 80101819-80101819 on Build GRCh38
rs1055811035 Chr.17: 80101809-80101809 on Build GRCh38
rs186478751 Chr.17: 80102085-80102085 on Build GRCh38
rs570203175 Chr.17: 80102084-80102084 on Build GRCh38
rs758090272 Chr.17: 80101962-80101962 on Build GRCh38
rs572088663 Chr.17: 80102241-80102241 on Build GRCh38
rs994001995 Chr.17: 80101883-80101883 on Build GRCh38
rs4889961 Chr.17: 80102109-80102109 on Build GRCh38
rs751223629 Chr.17: 80102225-80102225 on Build GRCh38
rs981639988 Chr.17: 80101978-80101978 on Build GRCh38
rs928681379 Chr.17: 80102000-80102000 on Build GRCh38
rs190152638 Chr.17: 80101807-80101807 on Build GRCh38
rs976176183 Chr.17: 80102133-80102133 on Build GRCh38
rs1013309408 Chr.17: 80102052-80102052 on Build GRCh38
rs1014843860 Chr.17: 80102121-80102121 on Build GRCh38
rs371769910 Chr.17: 80102206-80102207 on Build GRCh38
rs868102503 Chr.17: 80101893-80101893 on Build GRCh38
rs80020206 Chr.17: 80101848-80101848 on Build GRCh38
rs551673264 Chr.17: 80102059-80102059 on Build GRCh38
rs922063539 Chr.17: 80101921-80101921 on Build GRCh38
rs574436993 Chr.17: 80102163-80102163 on Build GRCh38
rs990389167 Chr.17: 80101925-80101925 on Build GRCh38
rs1800298 Chr.17: 80101841-80101841 on Build GRCh38
rs761168347 Chr.17: 80102057-80102057 on Build GRCh38
rs67074426 Chr.17: 80102206-80102206 on Build GRCh38
rs896117211 Chr.17: 80102240-80102240 on Build GRCh38
rs969303535 Chr.17: 80101844-80101844 on Build GRCh38
rs149485383 Chr.17: 80102183-80102184 on Build GRCh38
rs952168973 Chr.17: 80101870-80101870 on Build GRCh38
rs1052079452 Chr.17: 80101997-80101997 on Build GRCh38
rs999621900 Chr.17: 80101975-80101975 on Build GRCh38
rs1028011474 Chr.17: 80102155-80102155 on Build GRCh38
rs1027337344 Chr.17: 80102099-80102099 on Build GRCh38
rs951303153 Chr.17: 80102104-80102104 on Build GRCh38
rs979606686 Chr.17: 80102162-80102162 on Build GRCh38
rs922013034 Chr.17: 80102159-80102159 on Build GRCh38
rs890848228 Chr.17: 80102184-80102184 on Build GRCh38
rs367731841 Chr.17: 80101861-80101861 on Build GRCh38
rs913523286 Chr.17: 80101926-80101926 on Build GRCh38
rs8077056 Chr.17: 80102207-80102207 on Build GRCh38
rs372648633 Chr.17: 80101940-80101940 on Build GRCh38
rs925544300 Chr.17: 80102185-80102185 on Build GRCh38
rs374851617 Chr.17: 80102189-80102190 on Build GRCh38
rs1023478196 Chr.17: 80102058-80102058 on Build GRCh38
rs565764519 Chr.17: 80101933-80101933 on Build GRCh38
rs1040526680 Chr.17: 80101904-80101904 on Build GRCh38
rs904093120 Chr.17: 80101968-80101968 on Build GRCh38
rs1013446095 Chr.17: 80101836-80101836 on Build GRCh38
rs932168580 Chr.17: 80101922-80101922 on Build GRCh38
rs8077055 Chr.17: 80102206-80102206 on Build GRCh38
rs550203457 Chr.17: 80101816-80101816 on Build GRCh38
rs887765642 Chr.17: 80102068-80102068 on Build GRCh38
rs879766689 Chr.17: 80102127-80102127 on Build GRCh38
rs907981020 Chr.17: 80101886-80101886 on Build GRCh38
rs1044404528 Chr.17: 80101943-80101943 on Build GRCh38

View Full Product Details