Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80107415-80107905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAA
Entrez Gene ID
2548
Gene Name
glucosidase alpha, acid
Gene Aliases
LYAG
UniGene
Hs.1437
Gene Chromosome Location
Chr.17: 80101526-80119882 on Build GRCh38
Transcripts
NM_000152.4 NM_001079803.2 NM_001079804.2 XM_005257193.2 XM_005257194.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
985504 c.783C>T p.A261A Chr.17: 80107647-80107647 on Build GRCh38
6223906 c.862G>A p.G288S Chr.17: 80107803-80107803 on Build GRCh38
3787463 c.711G>A p.A237A Chr.17: 80107575-80107575 on Build GRCh38
2803135 c.900G>A p.A300A Chr.17: 80107841-80107841 on Build GRCh38
2803127 c.761C>T p.S254L Chr.17: 80107625-80107625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754080080 Chr.17: 80107719-80107719 on Build GRCh38
rs121907944 Chr.17: 80107574-80107574 on Build GRCh38
rs121907945 Chr.17: 80107818-80107818 on Build GRCh38
rs74003611 Chr.17: 80107479-80107479 on Build GRCh38
rs779785396 Chr.17: 80107659-80107659 on Build GRCh38
rs763538104 Chr.17: 80107552-80107552 on Build GRCh38
rs886042358 Chr.17: 80107875-80107875 on Build GRCh38
rs752724918 Chr.17: 80107850-80107850 on Build GRCh38
rs386386749 Chr.17: 80107728-80107729 on Build GRCh38
rs375310352 Chr.17: 80107721-80107721 on Build GRCh38
rs372331052 Chr.17: 80107729-80107730 on Build GRCh38
rs769074720 Chr.17: 80107804-80107804 on Build GRCh38
rs150343359 Chr.17: 80107856-80107856 on Build GRCh38
rs977154756 Chr.17: 80107536-80107536 on Build GRCh38
rs766931184 Chr.17: 80107564-80107564 on Build GRCh38
rs748788967 Chr.17: 80107745-80107745 on Build GRCh38
rs764463384 Chr.17: 80107750-80107750 on Build GRCh38
rs533960093 Chr.17: 80107626-80107626 on Build GRCh38
rs775090947 Chr.17: 80107542-80107542 on Build GRCh38
rs142967546 Chr.17: 80107705-80107705 on Build GRCh38
rs768488233 Chr.17: 80107685-80107685 on Build GRCh38
rs975301817 Chr.17: 80107484-80107484 on Build GRCh38
rs752054011 Chr.17: 80107565-80107565 on Build GRCh38
rs534515557 Chr.17: 80107808-80107808 on Build GRCh38
rs778882374 Chr.17: 80107579-80107579 on Build GRCh38
rs768125567 Chr.17: 80107745-80107746 on Build GRCh38
rs543360994 Chr.17: 80107645-80107645 on Build GRCh38
rs201056962 Chr.17: 80107722-80107722 on Build GRCh38
rs761363283 Chr.17: 80107617-80107617 on Build GRCh38
rs577915581 Chr.17: 80107625-80107625 on Build GRCh38
rs941586376 Chr.17: 80107869-80107869 on Build GRCh38
rs763091901 Chr.17: 80107870-80107870 on Build GRCh38
rs148578399 Chr.17: 80107590-80107590 on Build GRCh38
rs772002851 Chr.17: 80107658-80107658 on Build GRCh38
rs759425304 Chr.17: 80107841-80107841 on Build GRCh38
rs758441034 Chr.17: 80107755-80107755 on Build GRCh38
rs121907940 Chr.17: 80107837-80107837 on Build GRCh38
rs762056436 Chr.17: 80107751-80107751 on Build GRCh38
rs746986703 Chr.17: 80107736-80107736 on Build GRCh38
rs758283022 Chr.17: 80107732-80107732 on Build GRCh38
rs539428524 Chr.17: 80107476-80107476 on Build GRCh38
rs1057516600 Chr.17: 80107816-80107816 on Build GRCh38
rs752950890 Chr.17: 80107568-80107568 on Build GRCh38
rs772821371 Chr.17: 80107732-80107733 on Build GRCh38
rs3071247 Chr.17: 80107727-80107728 on Build GRCh38
rs745861849 Chr.17: 80107589-80107589 on Build GRCh38
rs773417785 Chr.17: 80107812-80107812 on Build GRCh38
rs147569830 Chr.17: 80107715-80107715 on Build GRCh38
rs760379754 Chr.17: 80107544-80107544 on Build GRCh38
rs768328783 Chr.17: 80107608-80107608 on Build GRCh38
rs1032949450 Chr.17: 80107840-80107840 on Build GRCh38
rs112025212 Chr.17: 80107863-80107863 on Build GRCh38
rs772607616 Chr.17: 80107706-80107706 on Build GRCh38
rs751639773 Chr.17: 80107824-80107824 on Build GRCh38
rs370502229 Chr.17: 80107509-80107509 on Build GRCh38
rs200856561 Chr.17: 80107616-80107616 on Build GRCh38
rs765295893 Chr.17: 80107728-80107728 on Build GRCh38
rs536976019 Chr.17: 80107530-80107530 on Build GRCh38
rs552929702 Chr.17: 80107809-80107809 on Build GRCh38
rs754919958 Chr.17: 80107778-80107778 on Build GRCh38
rs781731240 Chr.17: 80107567-80107567 on Build GRCh38
rs747820772 Chr.17: 80107704-80107704 on Build GRCh38
rs753249206 Chr.17: 80107746-80107747 on Build GRCh38
rs968966660 Chr.17: 80107729-80107729 on Build GRCh38
rs886042706 Chr.17: 80107726-80107726 on Build GRCh38
rs777262966 Chr.17: 80107793-80107793 on Build GRCh38
rs372614893 Chr.17: 80107644-80107644 on Build GRCh38
rs886042086 Chr.17: 80107717-80107717 on Build GRCh38
rs745762120 Chr.17: 80107652-80107652 on Build GRCh38
rs140301384 Chr.17: 80107851-80107851 on Build GRCh38
rs111888302 Chr.17: 80107748-80107748 on Build GRCh38
rs200509552 Chr.17: 80107631-80107631 on Build GRCh38
rs200154987 Chr.17: 80107854-80107854 on Build GRCh38
rs777964427 Chr.17: 80107570-80107570 on Build GRCh38
rs1015323808 Chr.17: 80107496-80107496 on Build GRCh38
rs770608668 Chr.17: 80107746-80107746 on Build GRCh38
rs200125735 Chr.17: 80107803-80107803 on Build GRCh38
rs771827503 Chr.17: 80107534-80107534 on Build GRCh38
rs750706928 Chr.17: 80107753-80107753 on Build GRCh38
rs369735511 Chr.17: 80107800-80107800 on Build GRCh38
rs773956464 Chr.17: 80107527-80107527 on Build GRCh38
rs776314424 Chr.17: 80107698-80107698 on Build GRCh38
rs143478051 Chr.17: 80107723-80107724 on Build GRCh38
rs200088236 Chr.17: 80107553-80107553 on Build GRCh38
rs373276219 Chr.17: 80107859-80107859 on Build GRCh38
rs1047100216 Chr.17: 80107456-80107456 on Build GRCh38
rs199674786 Chr.17: 80107633-80107633 on Build GRCh38
rs368328598 Chr.17: 80107575-80107575 on Build GRCh38
rs761121318 Chr.17: 80107804-80107805 on Build GRCh38
rs377222482 Chr.17: 80107782-80107782 on Build GRCh38
rs886043920 Chr.17: 80107600-80107600 on Build GRCh38
rs1005297878 Chr.17: 80107493-80107493 on Build GRCh38
rs78855075 Chr.17: 80107508-80107508 on Build GRCh38
rs371980868 Chr.17: 80107739-80107739 on Build GRCh38
rs569345218 Chr.17: 80107765-80107765 on Build GRCh38
rs757426536 Chr.17: 80107725-80107726 on Build GRCh38
rs755337511 Chr.17: 80107566-80107566 on Build GRCh38
rs760016684 Chr.17: 80107714-80107714 on Build GRCh38
rs1005162016 Chr.17: 80107654-80107654 on Build GRCh38
rs368741815 Chr.17: 80107853-80107853 on Build GRCh38
rs543300039 Chr.17: 80107866-80107866 on Build GRCh38
rs753112231 Chr.17: 80107638-80107638 on Build GRCh38
rs2304845 Chr.17: 80107752-80107752 on Build GRCh38
rs747775111 Chr.17: 80107740-80107740 on Build GRCh38
rs771171066 Chr.17: 80107875-80107875 on Build GRCh38
rs367620645 Chr.17: 80107769-80107769 on Build GRCh38
rs79849256 Chr.17: 80107463-80107463 on Build GRCh38
rs551335903 Chr.17: 80107764-80107764 on Build GRCh38
rs370105640 Chr.17: 80107865-80107865 on Build GRCh38
rs769332061 Chr.17: 80107743-80107743 on Build GRCh38
rs185883087 Chr.17: 80107779-80107779 on Build GRCh38
rs5822325 Chr.17: 80107730-80107730 on Build GRCh38
rs200381374 Chr.17: 80107600-80107600 on Build GRCh38
rs1008936778 Chr.17: 80107710-80107710 on Build GRCh38
rs754760986 Chr.17: 80107737-80107737 on Build GRCh38
rs2304846 Chr.17: 80107569-80107569 on Build GRCh38
rs976582329 Chr.17: 80107539-80107539 on Build GRCh38
rs888669731 Chr.17: 80107791-80107791 on Build GRCh38
rs757356178 Chr.17: 80107647-80107647 on Build GRCh38
rs201896815 Chr.17: 80107648-80107648 on Build GRCh38
rs142626724 Chr.17: 80107716-80107716 on Build GRCh38
rs138097673 Chr.17: 80107858-80107858 on Build GRCh38
rs764622267 Chr.17: 80107718-80107718 on Build GRCh38
rs570717926 Chr.17: 80107728-80107734 on Build GRCh38
rs746827767 Chr.17: 80107682-80107682 on Build GRCh38
rs772735040 Chr.17: 80107744-80107744 on Build GRCh38
rs749017942 Chr.17: 80107796-80107796 on Build GRCh38
rs762351035 Chr.17: 80107733-80107734 on Build GRCh38
rs1800303 Chr.17: 80107862-80107862 on Build GRCh38
rs952374058 Chr.17: 80107480-80107480 on Build GRCh38
rs145903608 Chr.17: 80107807-80107807 on Build GRCh38
rs778580823 Chr.17: 80107802-80107802 on Build GRCh38
rs886043779 Chr.17: 80107868-80107868 on Build GRCh38
rs896876542 Chr.17: 80107695-80107695 on Build GRCh38
rs763216519 Chr.17: 80107817-80107817 on Build GRCh38
rs1046519954 Chr.17: 80107597-80107597 on Build GRCh38
rs755990698 Chr.17: 80107786-80107786 on Build GRCh38

View Full Product Details