Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80998278-80998780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP6
Entrez Gene ID
79643
Gene Name
charged multivesicular body protein 6
Gene Aliases
VPS20
UniGene
Hs.514560
Gene Chromosome Location
Chr.17: 80991841-81000133 on Build GRCh38
Transcripts
NM_024591.4 XM_005257668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1710888 c.524_525CC>TT p.S175F Chr.17: 80998394-80998395 on Build GRCh38
5706658 c.527A>G p.E176G Chr.17: 80998397-80998397 on Build GRCh38
4845644 c.524C>T p.S175F Chr.17: 80998394-80998394 on Build GRCh38
3523464 c.521C>T p.P174L Chr.17: 80998391-80998391 on Build GRCh38
6148437 c.496-2A>G p.? Chr.17: 80998364-80998364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78538787 Chr.17: 80998680-80998680 on Build GRCh38
rs745884974 Chr.17: 80998627-80998627 on Build GRCh38
rs773215747 Chr.17: 80998395-80998395 on Build GRCh38
rs757750786 Chr.17: 80998333-80998333 on Build GRCh38
rs372892947 Chr.17: 80998319-80998319 on Build GRCh38
rs929170634 Chr.17: 80998533-80998533 on Build GRCh38
rs141336967 Chr.17: 80998459-80998459 on Build GRCh38
rs767083197 Chr.17: 80998476-80998476 on Build GRCh38
rs780629404 Chr.17: 80998326-80998326 on Build GRCh38
rs751902834 Chr.17: 80998516-80998516 on Build GRCh38
rs758902078 Chr.17: 80998439-80998439 on Build GRCh38
rs771963338 Chr.17: 80998545-80998545 on Build GRCh38
rs755426607 Chr.17: 80998572-80998572 on Build GRCh38
rs776743854 Chr.17: 80998618-80998618 on Build GRCh38
rs374559574 Chr.17: 80998493-80998493 on Build GRCh38
rs762794828 Chr.17: 80998318-80998318 on Build GRCh38
rs753454978 Chr.17: 80998318-80998318 on Build GRCh38
rs759324140 Chr.17: 80998465-80998465 on Build GRCh38
rs867217332 Chr.17: 80998710-80998710 on Build GRCh38
rs151073958 Chr.17: 80998488-80998489 on Build GRCh38
rs751391493 Chr.17: 80998402-80998402 on Build GRCh38
rs986778802 Chr.17: 80998303-80998303 on Build GRCh38
rs767174408 Chr.17: 80998407-80998407 on Build GRCh38
rs770957857 Chr.17: 80998460-80998460 on Build GRCh38
rs772368783 Chr.17: 80998368-80998368 on Build GRCh38
rs556624615 Chr.17: 80998556-80998556 on Build GRCh38
rs960114775 Chr.17: 80998648-80998648 on Build GRCh38
rs778035091 Chr.17: 80998524-80998524 on Build GRCh38
rs943136990 Chr.17: 80998752-80998752 on Build GRCh38
rs768892232 Chr.17: 80998382-80998382 on Build GRCh38
rs763715430 Chr.17: 80998557-80998557 on Build GRCh38
rs745667395 Chr.17: 80998544-80998544 on Build GRCh38
rs985538788 Chr.17: 80998745-80998745 on Build GRCh38
rs915926067 Chr.17: 80998673-80998673 on Build GRCh38
rs746238929 Chr.17: 80998354-80998354 on Build GRCh38
rs58642634 Chr.17: 80998620-80998620 on Build GRCh38
rs748460218 Chr.17: 80998520-80998520 on Build GRCh38
rs745597012 Chr.17: 80998387-80998387 on Build GRCh38
rs1011848674 Chr.17: 80998614-80998614 on Build GRCh38
rs911235550 Chr.17: 80998597-80998597 on Build GRCh38
rs754641674 Chr.17: 80998321-80998321 on Build GRCh38
rs1006153825 Chr.17: 80998522-80998522 on Build GRCh38
rs755301204 Chr.17: 80998517-80998517 on Build GRCh38
rs386799835 Chr.17: 80998620-80998621 on Build GRCh38
rs199819133 Chr.17: 80998467-80998467 on Build GRCh38
rs1014833786 Chr.17: 80998525-80998525 on Build GRCh38
rs533888039 Chr.17: 80998492-80998492 on Build GRCh38
rs371645115 Chr.17: 80998416-80998416 on Build GRCh38
rs749408898 Chr.17: 80998616-80998616 on Build GRCh38
rs538525389 Chr.17: 80998554-80998554 on Build GRCh38
rs550656207 Chr.17: 80998647-80998647 on Build GRCh38
rs776142210 Chr.17: 80998558-80998558 on Build GRCh38
rs1030062281 Chr.17: 80998299-80998299 on Build GRCh38
rs768665616 Chr.17: 80998540-80998540 on Build GRCh38
rs766824077 Chr.17: 80998509-80998509 on Build GRCh38
rs997804347 Chr.17: 80998715-80998715 on Build GRCh38
rs1051550678 Chr.17: 80998700-80998700 on Build GRCh38
rs562597429 Chr.17: 80998691-80998691 on Build GRCh38
rs1021806302 Chr.17: 80998418-80998418 on Build GRCh38
rs766798191 Chr.17: 80998562-80998562 on Build GRCh38
rs911288450 Chr.17: 80998753-80998753 on Build GRCh38
rs747261516 Chr.17: 80998378-80998378 on Build GRCh38
rs767992452 Chr.17: 80998574-80998574 on Build GRCh38
rs963733652 Chr.17: 80998676-80998676 on Build GRCh38
rs756395720 Chr.17: 80998521-80998521 on Build GRCh38
rs61752553 Chr.17: 80998392-80998392 on Build GRCh38
rs763240977 Chr.17: 80998662-80998662 on Build GRCh38
rs759148380 Chr.17: 80998405-80998405 on Build GRCh38
rs548390974 Chr.17: 80998636-80998636 on Build GRCh38
rs35812295 Chr.17: 80998504-80998504 on Build GRCh38
rs192286462 Chr.17: 80998519-80998519 on Build GRCh38
rs752046186 Chr.17: 80998563-80998563 on Build GRCh38
rs779409587 Chr.17: 80998430-80998430 on Build GRCh38
rs567503075 Chr.17: 80998369-80998369 on Build GRCh38
rs562400658 Chr.17: 80998652-80998652 on Build GRCh38
rs201102345 Chr.17: 80998463-80998463 on Build GRCh38
rs769881954 Chr.17: 80998394-80998394 on Build GRCh38
rs575086517 Chr.17: 80998603-80998603 on Build GRCh38
rs11655877 Chr.17: 80998685-80998685 on Build GRCh38
rs760310482 Chr.17: 80998491-80998491 on Build GRCh38
rs111347353 Chr.17: 80998490-80998491 on Build GRCh38
rs1030184605 Chr.17: 80998731-80998731 on Build GRCh38
rs756450650 Chr.17: 80998604-80998604 on Build GRCh38
rs374907086 Chr.17: 80998320-80998320 on Build GRCh38
rs761274974 Chr.17: 80998559-80998559 on Build GRCh38
rs768784343 Chr.17: 80998449-80998449 on Build GRCh38
rs780437898 Chr.17: 80998441-80998441 on Build GRCh38
rs911609457 Chr.17: 80998705-80998705 on Build GRCh38
rs775153003 Chr.17: 80998489-80998489 on Build GRCh38
rs201751751 Chr.17: 80998427-80998427 on Build GRCh38
rs551091387 Chr.17: 80998754-80998754 on Build GRCh38
rs11653865 Chr.17: 80998621-80998621 on Build GRCh38
rs771012946 Chr.17: 80998535-80998535 on Build GRCh38
rs752217922 Chr.17: 80998327-80998327 on Build GRCh38
rs58065351 Chr.17: 80998492-80998493 on Build GRCh38
rs774280529 Chr.17: 80998538-80998538 on Build GRCh38
rs750990282 Chr.17: 80998437-80998437 on Build GRCh38
rs747379209 Chr.17: 80998443-80998443 on Build GRCh38
rs975934354 Chr.17: 80998536-80998536 on Build GRCh38
rs530890415 Chr.17: 80998711-80998711 on Build GRCh38
rs778089912 Chr.17: 80998612-80998612 on Build GRCh38
rs755446388 Chr.17: 80998572-80998583 on Build GRCh38
rs10871491 Chr.17: 80998526-80998526 on Build GRCh38
rs1007143016 Chr.17: 80998573-80998573 on Build GRCh38
rs748179382 Chr.17: 80998458-80998458 on Build GRCh38
rs58412830 Chr.17: 80998341-80998341 on Build GRCh38
rs953305044 Chr.17: 80998739-80998739 on Build GRCh38

View Full Product Details