Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81441875-81442398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHCC1
Entrez Gene ID
57597
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
BAHD2
UniGene
Hs.514580
Gene Chromosome Location
Chr.17: 81395431-81466332 on Build GRCh38
Transcripts
NM_001291324.1 XM_011525063.2 XM_017024898.1 XM_017024899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6312912 c.528G>T p.P176P Chr.17: 81441877-81441877 on Build GRCh38
1184402 c.955C>T p.R319C Chr.17: 81442304-81442304 on Build GRCh38
4130811 c.814G>A p.A272T Chr.17: 81442163-81442163 on Build GRCh38
4791712 c.794G>A p.G265D Chr.17: 81442143-81442143 on Build GRCh38
5733712 c.766G>A p.V256M Chr.17: 81442115-81442115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545586429 Chr.17: 81442339-81442339 on Build GRCh38
rs369843602 Chr.17: 81441995-81441995 on Build GRCh38
rs1006772811 Chr.17: 81442230-81442230 on Build GRCh38
rs782785349 Chr.17: 81442147-81442147 on Build GRCh38
rs562760339 Chr.17: 81442145-81442145 on Build GRCh38
rs782537173 Chr.17: 81441942-81441942 on Build GRCh38
rs1006844904 Chr.17: 81442364-81442364 on Build GRCh38
rs782006187 Chr.17: 81442150-81442150 on Build GRCh38
rs781916395 Chr.17: 81441997-81441997 on Build GRCh38
rs1018326419 Chr.17: 81442252-81442252 on Build GRCh38
rs542765721 Chr.17: 81441954-81441954 on Build GRCh38
rs782589241 Chr.17: 81441955-81441955 on Build GRCh38
rs782682025 Chr.17: 81442038-81442038 on Build GRCh38
rs782784821 Chr.17: 81441999-81441999 on Build GRCh38
rs893020900 Chr.17: 81442370-81442370 on Build GRCh38
rs782358767 Chr.17: 81442302-81442302 on Build GRCh38
rs370107300 Chr.17: 81441923-81441923 on Build GRCh38
rs782395344 Chr.17: 81442043-81442043 on Build GRCh38
rs782637161 Chr.17: 81442354-81442354 on Build GRCh38
rs781825580 Chr.17: 81442273-81442273 on Build GRCh38
rs782582251 Chr.17: 81442035-81442035 on Build GRCh38
rs758323235 Chr.17: 81441992-81441992 on Build GRCh38
rs376051584 Chr.17: 81441905-81441905 on Build GRCh38
rs199920012 Chr.17: 81441957-81441957 on Build GRCh38
rs781973096 Chr.17: 81441908-81441908 on Build GRCh38
rs782531729 Chr.17: 81442277-81442277 on Build GRCh38
rs374880511 Chr.17: 81442225-81442225 on Build GRCh38
rs781895622 Chr.17: 81442000-81442000 on Build GRCh38
rs782052171 Chr.17: 81442263-81442263 on Build GRCh38
rs994030924 Chr.17: 81442180-81442180 on Build GRCh38
rs782254223 Chr.17: 81442042-81442042 on Build GRCh38
rs782424557 Chr.17: 81442190-81442190 on Build GRCh38
rs548283336 Chr.17: 81442161-81442161 on Build GRCh38
rs782331941 Chr.17: 81442137-81442137 on Build GRCh38
rs369124666 Chr.17: 81442264-81442264 on Build GRCh38
rs782335537 Chr.17: 81442360-81442360 on Build GRCh38
rs782665038 Chr.17: 81442096-81442096 on Build GRCh38
rs200849322 Chr.17: 81441994-81441994 on Build GRCh38
rs34122826 Chr.17: 81442154-81442154 on Build GRCh38
rs782652330 Chr.17: 81442294-81442294 on Build GRCh38
rs565437035 Chr.17: 81442082-81442082 on Build GRCh38
rs201597980 Chr.17: 81441899-81441899 on Build GRCh38
rs782765812 Chr.17: 81441931-81441931 on Build GRCh38
rs781800782 Chr.17: 81442276-81442276 on Build GRCh38
rs781802055 Chr.17: 81442153-81442153 on Build GRCh38
rs781976545 Chr.17: 81442248-81442248 on Build GRCh38
rs782159043 Chr.17: 81442285-81442285 on Build GRCh38
rs782378417 Chr.17: 81442066-81442066 on Build GRCh38
rs782230049 Chr.17: 81442285-81442285 on Build GRCh38
rs782116373 Chr.17: 81442006-81442006 on Build GRCh38
rs782316191 Chr.17: 81442061-81442061 on Build GRCh38
rs782063768 Chr.17: 81441998-81441998 on Build GRCh38
rs782178036 Chr.17: 81442046-81442046 on Build GRCh38
rs782110710 Chr.17: 81442270-81442270 on Build GRCh38
rs375318122 Chr.17: 81442115-81442115 on Build GRCh38
rs781916792 Chr.17: 81442303-81442303 on Build GRCh38
rs12952981 Chr.17: 81442163-81442163 on Build GRCh38
rs782539894 Chr.17: 81442345-81442345 on Build GRCh38
rs782503571 Chr.17: 81442095-81442095 on Build GRCh38
rs748213920 Chr.17: 81442160-81442160 on Build GRCh38
rs200635626 Chr.17: 81442340-81442340 on Build GRCh38
rs782546611 Chr.17: 81442036-81442036 on Build GRCh38
rs990731433 Chr.17: 81441897-81441897 on Build GRCh38
rs782773622 Chr.17: 81442271-81442271 on Build GRCh38
rs373285894 Chr.17: 81442275-81442275 on Build GRCh38
rs913420813 Chr.17: 81442094-81442094 on Build GRCh38
rs782018075 Chr.17: 81441917-81441917 on Build GRCh38
rs782180883 Chr.17: 81442136-81442136 on Build GRCh38
rs782421130 Chr.17: 81441988-81441988 on Build GRCh38
rs111391571 Chr.17: 81442363-81442363 on Build GRCh38
rs116138822 Chr.17: 81441898-81441898 on Build GRCh38
rs782674396 Chr.17: 81441948-81441948 on Build GRCh38
rs782495499 Chr.17: 81442274-81442274 on Build GRCh38
rs145916430 Chr.17: 81442151-81442151 on Build GRCh38
rs782742161 Chr.17: 81442073-81442073 on Build GRCh38
rs782496259 Chr.17: 81442338-81442338 on Build GRCh38
rs782368279 Chr.17: 81442297-81442297 on Build GRCh38
rs782285111 Chr.17: 81442185-81442185 on Build GRCh38
rs782336448 Chr.17: 81442126-81442126 on Build GRCh38
rs781969649 Chr.17: 81442127-81442127 on Build GRCh38
rs782743508 Chr.17: 81442152-81442152 on Build GRCh38
rs781903726 Chr.17: 81442090-81442090 on Build GRCh38
rs782617843 Chr.17: 81442368-81442368 on Build GRCh38
rs782281838 Chr.17: 81441986-81441986 on Build GRCh38
rs782222393 Chr.17: 81442164-81442164 on Build GRCh38
rs1039623827 Chr.17: 81442369-81442369 on Build GRCh38
rs782284304 Chr.17: 81442295-81442295 on Build GRCh38
rs782057281 Chr.17: 81441911-81441911 on Build GRCh38
rs782248746 Chr.17: 81442355-81442355 on Build GRCh38
rs782491275 Chr.17: 81442106-81442106 on Build GRCh38
rs782810758 Chr.17: 81442320-81442320 on Build GRCh38
rs928769611 Chr.17: 81442296-81442296 on Build GRCh38
rs782029368 Chr.17: 81442265-81442265 on Build GRCh38
rs552565913 Chr.17: 81442226-81442226 on Build GRCh38
rs781855498 Chr.17: 81442279-81442279 on Build GRCh38
rs782530704 Chr.17: 81442158-81442158 on Build GRCh38
rs12938890 Chr.17: 81442283-81442283 on Build GRCh38
rs370343920 Chr.17: 81442222-81442222 on Build GRCh38
rs782476877 Chr.17: 81441939-81441939 on Build GRCh38
rs782704662 Chr.17: 81441941-81441941 on Build GRCh38
rs782722714 Chr.17: 81441915-81441915 on Build GRCh38
rs782281202 Chr.17: 81441896-81441896 on Build GRCh38
rs377738105 Chr.17: 81441906-81441906 on Build GRCh38
rs1050885053 Chr.17: 81442208-81442208 on Build GRCh38
rs782241889 Chr.17: 81441953-81441953 on Build GRCh38
rs540262888 Chr.17: 81442069-81442069 on Build GRCh38
rs372078407 Chr.17: 81441965-81441965 on Build GRCh38
rs782181995 Chr.17: 81442371-81442371 on Build GRCh38
rs532870321 Chr.17: 81442083-81442083 on Build GRCh38
rs374129514 Chr.17: 81441979-81441979 on Build GRCh38
rs554357521 Chr.17: 81442348-81442348 on Build GRCh38
rs782528927 Chr.17: 81442282-81442282 on Build GRCh38
rs781831555 Chr.17: 81442099-81442099 on Build GRCh38
rs782607836 Chr.17: 81442045-81442045 on Build GRCh38
rs782588265 Chr.17: 81442176-81442176 on Build GRCh38
rs782445850 Chr.17: 81442168-81442168 on Build GRCh38
rs781998556 Chr.17: 81442068-81442068 on Build GRCh38
rs369911036 Chr.17: 81442123-81442123 on Build GRCh38
rs369872052 Chr.17: 81442305-81442305 on Build GRCh38
rs782788520 Chr.17: 81442088-81442088 on Build GRCh38
rs782011046 Chr.17: 81442062-81442062 on Build GRCh38
rs575512055 Chr.17: 81442301-81442301 on Build GRCh38
rs782492484 Chr.17: 81442030-81442030 on Build GRCh38
rs782396498 Chr.17: 81442300-81442301 on Build GRCh38
rs372031993 Chr.17: 81442114-81442114 on Build GRCh38
rs374363244 Chr.17: 81441937-81441937 on Build GRCh38
rs781992152 Chr.17: 81442300-81442300 on Build GRCh38

View Full Product Details