Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81442523-81443012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHCC1
Entrez Gene ID
57597
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
BAHD2
UniGene
Hs.514580
Gene Chromosome Location
Chr.17: 81395431-81466332 on Build GRCh38
Transcripts
NM_001291324.1 XM_011525063.2 XM_017024898.1 XM_017024899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5086925 c.1526A>G p.Q509R Chr.17: 81442875-81442875 on Build GRCh38
6082556 c.1606C>T p.P536S Chr.17: 81442955-81442955 on Build GRCh38
5093833 c.1307C>A p.P436H Chr.17: 81442656-81442656 on Build GRCh38
301738 c.1293C>A p.P431P Chr.17: 81442642-81442642 on Build GRCh38
6768690 c.1483G>A p.E495K Chr.17: 81442832-81442832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367646703 Chr.17: 81442621-81442621 on Build GRCh38
rs113216778 Chr.17: 81442746-81442746 on Build GRCh38
rs782360936 Chr.17: 81442642-81442642 on Build GRCh38
rs138865361 Chr.17: 81442580-81442580 on Build GRCh38
rs782746524 Chr.17: 81442745-81442745 on Build GRCh38
rs374297742 Chr.17: 81442583-81442583 on Build GRCh38
rs375936294 Chr.17: 81442618-81442618 on Build GRCh38
rs376763313 Chr.17: 81442649-81442649 on Build GRCh38
rs782432504 Chr.17: 81442671-81442671 on Build GRCh38
rs569151852 Chr.17: 81442953-81442953 on Build GRCh38
rs781870092 Chr.17: 81442960-81442960 on Build GRCh38
rs782619473 Chr.17: 81442862-81442862 on Build GRCh38
rs572835735 Chr.17: 81442588-81442588 on Build GRCh38
rs782120544 Chr.17: 81442582-81442582 on Build GRCh38
rs782419912 Chr.17: 81442921-81442921 on Build GRCh38
rs370290459 Chr.17: 81442777-81442777 on Build GRCh38
rs781836767 Chr.17: 81442750-81442750 on Build GRCh38
rs977663882 Chr.17: 81442977-81442977 on Build GRCh38
rs550244860 Chr.17: 81442882-81442882 on Build GRCh38
rs368080799 Chr.17: 81442566-81442566 on Build GRCh38
rs782066619 Chr.17: 81442919-81442919 on Build GRCh38
rs782044634 Chr.17: 81442944-81442944 on Build GRCh38
rs74002272 Chr.17: 81442778-81442778 on Build GRCh38
rs530051904 Chr.17: 81442955-81442955 on Build GRCh38
rs782660693 Chr.17: 81442987-81442987 on Build GRCh38
rs782122694 Chr.17: 81442931-81442931 on Build GRCh38
rs782384472 Chr.17: 81442725-81442725 on Build GRCh38
rs781854529 Chr.17: 81442739-81442739 on Build GRCh38
rs376860198 Chr.17: 81442793-81442793 on Build GRCh38
rs377044230 Chr.17: 81442734-81442734 on Build GRCh38
rs782541841 Chr.17: 81442949-81442949 on Build GRCh38
rs782036480 Chr.17: 81442925-81442925 on Build GRCh38
rs782294244 Chr.17: 81442993-81442993 on Build GRCh38
rs938565251 Chr.17: 81442790-81442790 on Build GRCh38
rs782356113 Chr.17: 81442906-81442906 on Build GRCh38
rs782406152 Chr.17: 81442798-81442798 on Build GRCh38
rs924542201 Chr.17: 81442781-81442781 on Build GRCh38
rs914389237 Chr.17: 81442595-81442595 on Build GRCh38
rs782782608 Chr.17: 81442579-81442579 on Build GRCh38
rs782346284 Chr.17: 81442780-81442780 on Build GRCh38
rs782128238 Chr.17: 81442730-81442730 on Build GRCh38
rs781909123 Chr.17: 81442655-81442655 on Build GRCh38
rs782526499 Chr.17: 81442860-81442860 on Build GRCh38
rs782350807 Chr.17: 81442562-81442562 on Build GRCh38
rs1007906262 Chr.17: 81442714-81442714 on Build GRCh38
rs201429848 Chr.17: 81442707-81442707 on Build GRCh38
rs533904909 Chr.17: 81442612-81442612 on Build GRCh38
rs781931093 Chr.17: 81442735-81442735 on Build GRCh38
rs564432457 Chr.17: 81442808-81442808 on Build GRCh38
rs782701798 Chr.17: 81442945-81442945 on Build GRCh38
rs781885572 Chr.17: 81442687-81442687 on Build GRCh38
rs951013161 Chr.17: 81442585-81442585 on Build GRCh38
rs996635001 Chr.17: 81442891-81442891 on Build GRCh38
rs544465445 Chr.17: 81442643-81442643 on Build GRCh38
rs782550650 Chr.17: 81442602-81442602 on Build GRCh38
rs782620463 Chr.17: 81442704-81442704 on Build GRCh38
rs570222358 Chr.17: 81442832-81442832 on Build GRCh38
rs890714498 Chr.17: 81442841-81442841 on Build GRCh38
rs942392950 Chr.17: 81442593-81442593 on Build GRCh38
rs782164754 Chr.17: 81442718-81442718 on Build GRCh38
rs781919013 Chr.17: 81442917-81442917 on Build GRCh38
rs781881364 Chr.17: 81442948-81442948 on Build GRCh38
rs782372513 Chr.17: 81442889-81442889 on Build GRCh38
rs782003982 Chr.17: 81442632-81442632 on Build GRCh38
rs375834153 Chr.17: 81442823-81442823 on Build GRCh38
rs781914861 Chr.17: 81442792-81442792 on Build GRCh38
rs782055507 Chr.17: 81442695-81442695 on Build GRCh38
rs199812758 Chr.17: 81442826-81442826 on Build GRCh38
rs560751046 Chr.17: 81442753-81442753 on Build GRCh38
rs554777397 Chr.17: 81442586-81442586 on Build GRCh38
rs782215129 Chr.17: 81442900-81442900 on Build GRCh38
rs782786342 Chr.17: 81442938-81442938 on Build GRCh38
rs782461900 Chr.17: 81442875-81442875 on Build GRCh38
rs550902726 Chr.17: 81442550-81442550 on Build GRCh38
rs782472181 Chr.17: 81442963-81442963 on Build GRCh38
rs782810003 Chr.17: 81442738-81442738 on Build GRCh38
rs782512018 Chr.17: 81442740-81442740 on Build GRCh38
rs782588831 Chr.17: 81442761-81442761 on Build GRCh38
rs564224428 Chr.17: 81442751-81442751 on Build GRCh38
rs782411538 Chr.17: 81442555-81442555 on Build GRCh38
rs374127888 Chr.17: 81442863-81442863 on Build GRCh38
rs948865584 Chr.17: 81442822-81442822 on Build GRCh38
rs546281324 Chr.17: 81442807-81442807 on Build GRCh38
rs781835723 Chr.17: 81442939-81442939 on Build GRCh38
rs782232814 Chr.17: 81442547-81442547 on Build GRCh38
rs574818751 Chr.17: 81442706-81442706 on Build GRCh38
rs782438830 Chr.17: 81442548-81442548 on Build GRCh38
rs782033885 Chr.17: 81442802-81442802 on Build GRCh38
rs781814739 Chr.17: 81442594-81442594 on Build GRCh38
rs782260970 Chr.17: 81442699-81442699 on Build GRCh38
rs371445776 Chr.17: 81442635-81442635 on Build GRCh38
rs782003359 Chr.17: 81442897-81442897 on Build GRCh38
rs782146993 Chr.17: 81442736-81442736 on Build GRCh38
rs370421012 Chr.17: 81442843-81442843 on Build GRCh38
rs377065491 Chr.17: 81442700-81442700 on Build GRCh38
rs782598915 Chr.17: 81442549-81442549 on Build GRCh38
rs781999779 Chr.17: 81442556-81442556 on Build GRCh38
rs201791070 Chr.17: 81442881-81442881 on Build GRCh38
rs373853835 Chr.17: 81442726-81442726 on Build GRCh38
rs374020665 Chr.17: 81442688-81442688 on Build GRCh38
rs782618803 Chr.17: 81442980-81442980 on Build GRCh38
rs531211102 Chr.17: 81442976-81442976 on Build GRCh38
rs782376318 Chr.17: 81442628-81442628 on Build GRCh38
rs12943867 Chr.17: 81442684-81442684 on Build GRCh38
rs569214613 Chr.17: 81442565-81442565 on Build GRCh38
rs782208958 Chr.17: 81442558-81442558 on Build GRCh38
rs782072753 Chr.17: 81442648-81442648 on Build GRCh38
rs782694274 Chr.17: 81442828-81442828 on Build GRCh38
rs781865961 Chr.17: 81442610-81442610 on Build GRCh38
rs782184927 Chr.17: 81442981-81442981 on Build GRCh38
rs368486632 Chr.17: 81442616-81442616 on Build GRCh38
rs782328775 Chr.17: 81442985-81442985 on Build GRCh38
rs782688480 Chr.17: 81442606-81442606 on Build GRCh38

View Full Product Details