Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81683825-81684318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL16
Entrez Gene ID
339231
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
-
UniGene
Hs.658789
Gene Chromosome Location
Chr.17: 81681194-81683924 on Build GRCh38
Transcripts
NM_001040025.1 NM_004712.4 XM_011525463.2 XM_017025297.1 XM_017025298.1

Gene Symbol
HGS
Entrez Gene ID
9146
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
Gene Aliases
HRS
UniGene
Hs.661056
Gene Chromosome Location
Chr.17: 81683932-81702121 on Build GRCh38
Transcripts
NM_001040025.1 NM_004712.4 XM_011525463.2 XM_017025297.1 XM_017025298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7138050 c.8G>A p.R3Q Chr.17: 81684074-81684074 on Build GRCh38
6401820 c.17G>A p.G6D Chr.17: 81684083-81684083 on Build GRCh38
4421101 c.13A>G p.S5G Chr.17: 81684079-81684079 on Build GRCh38
3523669 c.14G>T p.S5I Chr.17: 81684080-81684080 on Build GRCh38
3691935 c.34C>A p.L12I Chr.17: 81684100-81684100 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201570553 Chr.17: 81684052-81684052 on Build GRCh38
rs946842560 Chr.17: 81684151-81684151 on Build GRCh38
rs747407664 Chr.17: 81684048-81684048 on Build GRCh38
rs895448063 Chr.17: 81684178-81684178 on Build GRCh38
rs1051699219 Chr.17: 81683949-81683949 on Build GRCh38
rs368213492 Chr.17: 81684063-81684063 on Build GRCh38
rs1003825021 Chr.17: 81684009-81684009 on Build GRCh38
rs370999519 Chr.17: 81684228-81684228 on Build GRCh38
rs758581228 Chr.17: 81684138-81684138 on Build GRCh38
rs753903291 Chr.17: 81684038-81684038 on Build GRCh38
rs747461039 Chr.17: 81684020-81684020 on Build GRCh38
rs752782825 Chr.17: 81684132-81684132 on Build GRCh38
rs987312004 Chr.17: 81684254-81684254 on Build GRCh38
rs983431155 Chr.17: 81683891-81683891 on Build GRCh38
rs935484830 Chr.17: 81684060-81684060 on Build GRCh38
rs975167180 Chr.17: 81684207-81684207 on Build GRCh38
rs966135702 Chr.17: 81684205-81684205 on Build GRCh38
rs931978243 Chr.17: 81683939-81683939 on Build GRCh38
rs1039730498 Chr.17: 81683902-81683902 on Build GRCh38
rs374310994 Chr.17: 81683852-81683852 on Build GRCh38
rs1013914996 Chr.17: 81683957-81683957 on Build GRCh38
rs915271876 Chr.17: 81684268-81684268 on Build GRCh38
rs767407000 Chr.17: 81684027-81684027 on Build GRCh38
rs756178766 Chr.17: 81683854-81683854 on Build GRCh38
rs539712600 Chr.17: 81684152-81684152 on Build GRCh38
rs1004564235 Chr.17: 81684204-81684204 on Build GRCh38
rs537595032 Chr.17: 81684039-81684039 on Build GRCh38
rs1004662735 Chr.17: 81684164-81684164 on Build GRCh38
rs557815932 Chr.17: 81684220-81684220 on Build GRCh38
rs771381626 Chr.17: 81684049-81684049 on Build GRCh38
rs992311072 Chr.17: 81684264-81684264 on Build GRCh38
rs376204703 Chr.17: 81683872-81683872 on Build GRCh38
rs911819119 Chr.17: 81684262-81684262 on Build GRCh38
rs755186339 Chr.17: 81683865-81683865 on Build GRCh38
rs543176895 Chr.17: 81684292-81684292 on Build GRCh38
rs577986958 Chr.17: 81684136-81684136 on Build GRCh38
rs1023863962 Chr.17: 81684003-81684004 on Build GRCh38
rs922197449 Chr.17: 81683913-81683913 on Build GRCh38
rs949803388 Chr.17: 81684274-81684274 on Build GRCh38
rs1018981392 Chr.17: 81684025-81684025 on Build GRCh38
rs768046521 Chr.17: 81684059-81684059 on Build GRCh38
rs973206263 Chr.17: 81684216-81684216 on Build GRCh38
rs928306461 Chr.17: 81683881-81683881 on Build GRCh38
rs1052775295 Chr.17: 81683952-81683952 on Build GRCh38
rs765176981 Chr.17: 81684114-81684114 on Build GRCh38
rs946780840 Chr.17: 81684271-81684271 on Build GRCh38
rs189203673 Chr.17: 81683909-81683909 on Build GRCh38
rs200982711 Chr.17: 81684061-81684061 on Build GRCh38
rs561402446 Chr.17: 81683921-81683921 on Build GRCh38
rs114274845 Chr.17: 81684018-81684018 on Build GRCh38
rs757588396 Chr.17: 81684148-81684148 on Build GRCh38
rs577562062 Chr.17: 81684033-81684033 on Build GRCh38
rs926937798 Chr.17: 81684282-81684282 on Build GRCh38
rs146058677 Chr.17: 81684066-81684066 on Build GRCh38
rs766242507 Chr.17: 81684077-81684077 on Build GRCh38
rs995098984 Chr.17: 81683972-81683972 on Build GRCh38
rs866738234 Chr.17: 81684091-81684091 on Build GRCh38
rs907697419 Chr.17: 81683911-81683911 on Build GRCh38
rs776704027 Chr.17: 81684266-81684266 on Build GRCh38
rs1019473553 Chr.17: 81684170-81684170 on Build GRCh38
rs374574980 Chr.17: 81683954-81683954 on Build GRCh38
rs750256484 Chr.17: 81684030-81684030 on Build GRCh38
rs1044464492 Chr.17: 81684278-81684278 on Build GRCh38
rs780223715 Chr.17: 81683856-81683856 on Build GRCh38
rs751829825 Chr.17: 81684143-81684143 on Build GRCh38
rs749331897 Chr.17: 81683858-81683858 on Build GRCh38
rs944978933 Chr.17: 81683900-81683900 on Build GRCh38
rs934366135 Chr.17: 81683950-81683950 on Build GRCh38
rs752874903 Chr.17: 81684051-81684055 on Build GRCh38
rs531444654 Chr.17: 81683850-81683850 on Build GRCh38
rs767282458 Chr.17: 81684064-81684064 on Build GRCh38
rs528968669 Chr.17: 81683935-81683935 on Build GRCh38
rs556666891 Chr.17: 81684196-81684196 on Build GRCh38
rs867309375 Chr.17: 81684155-81684155 on Build GRCh38
rs987383854 Chr.17: 81684072-81684072 on Build GRCh38
rs753682847 Chr.17: 81684086-81684086 on Build GRCh38
rs755203910 Chr.17: 81683871-81683871 on Build GRCh38
rs779022975 Chr.17: 81683866-81683866 on Build GRCh38
rs756047044 Chr.17: 81684037-81684037 on Build GRCh38
rs1028964914 Chr.17: 81684213-81684213 on Build GRCh38
rs746428589 Chr.17: 81684054-81684054 on Build GRCh38
rs148743956 Chr.17: 81684093-81684093 on Build GRCh38
rs868658076 Chr.17: 81684092-81684092 on Build GRCh38
rs1003793886 Chr.17: 81684172-81684172 on Build GRCh38
rs760567020 Chr.17: 81684067-81684067 on Build GRCh38
rs946965084 Chr.17: 81684042-81684042 on Build GRCh38
rs953253678 Chr.17: 81684237-81684237 on Build GRCh38
rs533659840 Chr.17: 81683979-81683979 on Build GRCh38
rs777996111 Chr.17: 81684142-81684142 on Build GRCh38
rs941859694 Chr.17: 81683914-81683914 on Build GRCh38
rs551826459 Chr.17: 81684010-81684010 on Build GRCh38
rs935515785 Chr.17: 81684230-81684230 on Build GRCh38
rs773628468 Chr.17: 81684019-81684019 on Build GRCh38
rs758636311 Chr.17: 81684043-81684043 on Build GRCh38
rs905763770 Chr.17: 81683966-81683966 on Build GRCh38
rs763710819 Chr.17: 81684024-81684024 on Build GRCh38
rs905422628 Chr.17: 81684003-81684003 on Build GRCh38
rs911858153 Chr.17: 81683895-81683895 on Build GRCh38
rs778049575 Chr.17: 81684045-81684045 on Build GRCh38
rs950808659 Chr.17: 81683982-81683982 on Build GRCh38
rs1052498924 Chr.17: 81684130-81684130 on Build GRCh38
rs371698739 Chr.17: 81683980-81683981 on Build GRCh38
rs1043899335 Chr.17: 81683928-81683928 on Build GRCh38
rs890486976 Chr.17: 81683959-81683959 on Build GRCh38
rs983232580 Chr.17: 81683906-81683906 on Build GRCh38
rs1045190157 Chr.17: 81684285-81684285 on Build GRCh38
rs979722139 Chr.17: 81684224-81684224 on Build GRCh38
rs1011501366 Chr.17: 81683981-81683981 on Build GRCh38
rs773844357 Chr.17: 81684026-81684026 on Build GRCh38
rs77511372 Chr.17: 81683962-81683962 on Build GRCh38
rs1035272477 Chr.17: 81684174-81684174 on Build GRCh38
rs9890692 Chr.17: 81683980-81683980 on Build GRCh38
rs781361216 Chr.17: 81684042-81684043 on Build GRCh38

View Full Product Details