Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 616370-616892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUL1
Entrez Gene ID
27098
Gene Name
clusterin like 1
Gene Aliases
RA337M
UniGene
Hs.732341
Gene Chromosome Location
Chr.18: 596998-650298 on Build GRCh38
Transcripts
NM_001289036.1 NM_001318522.1 NM_014410.4 NM_199167.1 XM_005258103.3 XM_006722322.2 XM_011525648.2 XM_011525649.1 XM_011525651.2 XM_011525652.1 XM_011525653.2 XM_017025707.1

SNP Detail

SNP ID Chromosome Location
rs570788864 Chr.18: 616511-616511 on Build GRCh38
rs998303821 Chr.18: 616775-616775 on Build GRCh38
rs479141 Chr.18: 616582-616582 on Build GRCh38
rs186228304 Chr.18: 616703-616703 on Build GRCh38
rs485312 Chr.18: 616783-616783 on Build GRCh38
rs1015080525 Chr.18: 616428-616428 on Build GRCh38
rs1047021906 Chr.18: 616578-616578 on Build GRCh38
rs756840443 Chr.18: 616707-616707 on Build GRCh38
rs386800126 Chr.18: 616576-616582 on Build GRCh38
rs956995362 Chr.18: 616848-616848 on Build GRCh38
rs529678358 Chr.18: 616507-616507 on Build GRCh38
rs899945974 Chr.18: 616760-616760 on Build GRCh38
rs967269521 Chr.18: 616801-616801 on Build GRCh38
rs561663165 Chr.18: 616835-616835 on Build GRCh38
rs888746464 Chr.18: 616745-616745 on Build GRCh38
rs569686026 Chr.18: 616556-616556 on Build GRCh38
rs977737592 Chr.18: 616805-616805 on Build GRCh38
rs483655 Chr.18: 616653-616653 on Build GRCh38
rs114982364 Chr.18: 616601-616601 on Build GRCh38
rs751902579 Chr.18: 616533-616533 on Build GRCh38
rs549518739 Chr.18: 616518-616518 on Build GRCh38
rs957736347 Chr.18: 616522-616522 on Build GRCh38
rs1056077318 Chr.18: 616622-616622 on Build GRCh38
rs545649168 Chr.18: 616756-616756 on Build GRCh38
rs483706 Chr.18: 616576-616576 on Build GRCh38
rs778691191 Chr.18: 616708-616708 on Build GRCh38
rs1006062771 Chr.18: 616412-616412 on Build GRCh38
rs941856474 Chr.18: 616700-616700 on Build GRCh38
rs977003441 Chr.18: 616459-616459 on Build GRCh38
rs1012137768 Chr.18: 616833-616833 on Build GRCh38
rs967532481 Chr.18: 616859-616859 on Build GRCh38
rs1005512141 Chr.18: 616681-616681 on Build GRCh38
rs946156259 Chr.18: 616561-616561 on Build GRCh38
rs1020438659 Chr.18: 616857-616857 on Build GRCh38
rs564300898 Chr.18: 616773-616773 on Build GRCh38
rs190701128 Chr.18: 616751-616751 on Build GRCh38
rs1030033498 Chr.18: 616505-616505 on Build GRCh38
rs1051529424 Chr.18: 616724-616724 on Build GRCh38
rs986093857 Chr.18: 616517-616517 on Build GRCh38
rs997240480 Chr.18: 616714-616714 on Build GRCh38
rs766660050 Chr.18: 616491-616491 on Build GRCh38
rs367814928 Chr.18: 616501-616501 on Build GRCh38
rs990365792 Chr.18: 616550-616550 on Build GRCh38
rs967681018 Chr.18: 616487-616487 on Build GRCh38
rs183462864 Chr.18: 616812-616812 on Build GRCh38
rs897618386 Chr.18: 616459-616459 on Build GRCh38

View Full Product Details