Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 905150-905674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCYAP1
Entrez Gene ID
116
Gene Name
adenylate cyclase activating polypeptide 1
Gene Aliases
PACAP
UniGene
Hs.531719
Gene Chromosome Location
Chr.18: 904342-912172 on Build GRCh38
Transcripts
NM_001099733.1 NM_001117.4 XM_005258081.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
710337 c.15C>A p.S5R Chr.18: 905401-905401 on Build GRCh38
3959441 c.110+4G>T p.? Chr.18: 905500-905500 on Build GRCh38
324648 c.56A>G p.H19R Chr.18: 905442-905442 on Build GRCh38
710336 c.75A>C p.S25S Chr.18: 905461-905461 on Build GRCh38
3422350 c.88G>A p.G30R Chr.18: 905474-905474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533218516 Chr.18: 905572-905573 on Build GRCh38
rs181550079 Chr.18: 905602-905602 on Build GRCh38
rs12605354 Chr.18: 905200-905200 on Build GRCh38
rs547429745 Chr.18: 905182-905182 on Build GRCh38
rs776807884 Chr.18: 905528-905528 on Build GRCh38
rs916823015 Chr.18: 905469-905469 on Build GRCh38
rs140671226 Chr.18: 905622-905623 on Build GRCh38
rs765316211 Chr.18: 905572-905572 on Build GRCh38
rs566002626 Chr.18: 905200-905201 on Build GRCh38
rs754074374 Chr.18: 905239-905239 on Build GRCh38
rs774142783 Chr.18: 905451-905451 on Build GRCh38
rs373510219 Chr.18: 905278-905278 on Build GRCh38
rs958463338 Chr.18: 905464-905464 on Build GRCh38
rs765029234 Chr.18: 905472-905472 on Build GRCh38
rs760792813 Chr.18: 905187-905187 on Build GRCh38
rs375716849 Chr.18: 905642-905642 on Build GRCh38
rs537375314 Chr.18: 905340-905340 on Build GRCh38
rs928341128 Chr.18: 905499-905499 on Build GRCh38
rs186030171 Chr.18: 905653-905653 on Build GRCh38
rs755052187 Chr.18: 905380-905380 on Build GRCh38
rs1893153 Chr.18: 905202-905202 on Build GRCh38
rs924477730 Chr.18: 905241-905241 on Build GRCh38
rs576246004 Chr.18: 905567-905567 on Build GRCh38
rs747342728 Chr.18: 905484-905484 on Build GRCh38
rs906826357 Chr.18: 905399-905399 on Build GRCh38
rs749007403 Chr.18: 905491-905491 on Build GRCh38
rs765402609 Chr.18: 905477-905477 on Build GRCh38
rs757684819 Chr.18: 905538-905538 on Build GRCh38
rs552653569 Chr.18: 905516-905516 on Build GRCh38
rs548688850 Chr.18: 905291-905291 on Build GRCh38
rs938465717 Chr.18: 905257-905257 on Build GRCh38
rs370963358 Chr.18: 905530-905530 on Build GRCh38
rs924669214 Chr.18: 905536-905537 on Build GRCh38
rs766216691 Chr.18: 905527-905527 on Build GRCh38
rs753855693 Chr.18: 905368-905368 on Build GRCh38
rs373051517 Chr.18: 905474-905474 on Build GRCh38
rs377558347 Chr.18: 905427-905427 on Build GRCh38
rs758901557 Chr.18: 905481-905481 on Build GRCh38
rs954774133 Chr.18: 905344-905344 on Build GRCh38
rs948410989 Chr.18: 905236-905236 on Build GRCh38
rs761860008 Chr.18: 905461-905461 on Build GRCh38
rs934075811 Chr.18: 905218-905218 on Build GRCh38
rs562409830 Chr.18: 905588-905588 on Build GRCh38
rs748744145 Chr.18: 905401-905401 on Build GRCh38
rs771985769 Chr.18: 905517-905517 on Build GRCh38
rs779385024 Chr.18: 905391-905391 on Build GRCh38
rs867526464 Chr.18: 905350-905350 on Build GRCh38
rs767403588 Chr.18: 905347-905347 on Build GRCh38
rs886294261 Chr.18: 905279-905279 on Build GRCh38
rs756261455 Chr.18: 905346-905346 on Build GRCh38
rs757703280 Chr.18: 905485-905485 on Build GRCh38
rs569842982 Chr.18: 905304-905304 on Build GRCh38
rs948398868 Chr.18: 905331-905331 on Build GRCh38
rs2231184 Chr.18: 905558-905558 on Build GRCh38
rs752778053 Chr.18: 905533-905533 on Build GRCh38
rs890490675 Chr.18: 905610-905610 on Build GRCh38
rs773871169 Chr.18: 905515-905515 on Build GRCh38
rs2231183 Chr.18: 905536-905536 on Build GRCh38
rs907671106 Chr.18: 905531-905531 on Build GRCh38
rs761954441 Chr.18: 905548-905548 on Build GRCh38
rs938413845 Chr.18: 905508-905508 on Build GRCh38
rs1038942579 Chr.18: 905613-905613 on Build GRCh38
rs559482442 Chr.18: 905612-905612 on Build GRCh38
rs900476891 Chr.18: 905300-905300 on Build GRCh38
rs769265867 Chr.18: 905428-905428 on Build GRCh38
rs761509223 Chr.18: 905345-905345 on Build GRCh38
rs999583467 Chr.18: 905615-905615 on Build GRCh38
rs928393979 Chr.18: 905240-905240 on Build GRCh38
rs760474019 Chr.18: 905470-905470 on Build GRCh38
rs1030371963 Chr.18: 905329-905329 on Build GRCh38
rs750579742 Chr.18: 905359-905359 on Build GRCh38
rs989726208 Chr.18: 905467-905467 on Build GRCh38
rs886174864 Chr.18: 905605-905605 on Build GRCh38
rs780494397 Chr.18: 905190-905191 on Build GRCh38
rs781723482 Chr.18: 905542-905542 on Build GRCh38
rs188757425 Chr.18: 905585-905585 on Build GRCh38
rs778244814 Chr.18: 905420-905420 on Build GRCh38
rs938302041 Chr.18: 905578-905578 on Build GRCh38
rs1031162701 Chr.18: 905448-905448 on Build GRCh38
rs534614195 Chr.18: 905513-905513 on Build GRCh38
rs758664236 Chr.18: 905416-905416 on Build GRCh38
rs760455583 Chr.18: 905519-905519 on Build GRCh38
rs568852695 Chr.18: 905550-905550 on Build GRCh38
rs375496130 Chr.18: 905354-905354 on Build GRCh38
rs544682110 Chr.18: 905556-905556 on Build GRCh38
rs573823992 Chr.18: 905488-905488 on Build GRCh38
rs906878677 Chr.18: 905577-905577 on Build GRCh38
rs202111515 Chr.18: 905189-905190 on Build GRCh38
rs1051750118 Chr.18: 905564-905564 on Build GRCh38
rs533490986 Chr.18: 905650-905650 on Build GRCh38
rs914215413 Chr.18: 905204-905204 on Build GRCh38
rs747367878 Chr.18: 905423-905423 on Build GRCh38
rs773026207 Chr.18: 905518-905518 on Build GRCh38
rs745590193 Chr.18: 905191-905192 on Build GRCh38
rs747374395 Chr.18: 905335-905335 on Build GRCh38
rs1002629931 Chr.18: 905414-905414 on Build GRCh38
rs555648531 Chr.18: 905361-905361 on Build GRCh38
rs773505196 Chr.18: 905654-905654 on Build GRCh38
rs748742011 Chr.18: 905450-905450 on Build GRCh38
rs778168865 Chr.18: 905482-905482 on Build GRCh38
rs754609134 Chr.18: 905543-905543 on Build GRCh38
rs201296866 Chr.18: 905201-905202 on Build GRCh38
rs975425690 Chr.18: 905522-905522 on Build GRCh38
rs111709874 Chr.18: 905283-905283 on Build GRCh38
rs755035529 Chr.18: 905475-905475 on Build GRCh38
rs1037583726 Chr.18: 905547-905547 on Build GRCh38
rs1047550162 Chr.18: 905276-905276 on Build GRCh38
rs529524018 Chr.18: 905592-905592 on Build GRCh38
rs35305428 Chr.18: 905479-905479 on Build GRCh38
rs8192594 Chr.18: 905468-905468 on Build GRCh38
rs1047497942 Chr.18: 905584-905584 on Build GRCh38
rs766609304 Chr.18: 905365-905365 on Build GRCh38

View Full Product Details