Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 908130-908650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCYAP1
Entrez Gene ID
116
Gene Name
adenylate cyclase activating polypeptide 1
Gene Aliases
PACAP
UniGene
Hs.531719
Gene Chromosome Location
Chr.18: 904342-912172 on Build GRCh38
Transcripts
NM_001099733.1 NM_001117.4 XM_005258081.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7274061 c.313C>A p.L105M Chr.18: 908335-908335 on Build GRCh38
7354562 c.296T>A p.L99Q Chr.18: 908318-908318 on Build GRCh38
7354561 c.332G>T p.R111L Chr.18: 908354-908354 on Build GRCh38
990224 c.268G>A p.E90K Chr.18: 908290-908290 on Build GRCh38
4659980 c.311A>G p.H104R Chr.18: 908333-908333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946068660 Chr.18: 908583-908583 on Build GRCh38
rs1042632267 Chr.18: 908473-908473 on Build GRCh38
rs759961514 Chr.18: 908216-908216 on Build GRCh38
rs778889031 Chr.18: 908306-908306 on Build GRCh38
rs770517537 Chr.18: 908414-908414 on Build GRCh38
rs200894663 Chr.18: 908231-908231 on Build GRCh38
rs772983512 Chr.18: 908354-908354 on Build GRCh38
rs146950918 Chr.18: 908294-908294 on Build GRCh38
rs976677527 Chr.18: 908574-908574 on Build GRCh38
rs148570945 Chr.18: 908346-908346 on Build GRCh38
rs2231186 Chr.18: 908470-908470 on Build GRCh38
rs1025395028 Chr.18: 908502-908502 on Build GRCh38
rs568000959 Chr.18: 908333-908333 on Build GRCh38
rs144732014 Chr.18: 908299-908299 on Build GRCh38
rs189295968 Chr.18: 908474-908474 on Build GRCh38
rs936128641 Chr.18: 908610-908610 on Build GRCh38
rs746714533 Chr.18: 908412-908412 on Build GRCh38
rs766571865 Chr.18: 908235-908235 on Build GRCh38
rs760235350 Chr.18: 908300-908300 on Build GRCh38
rs574690187 Chr.18: 908303-908303 on Build GRCh38
rs772243726 Chr.18: 908253-908253 on Build GRCh38
rs752573601 Chr.18: 908411-908412 on Build GRCh38
rs770776837 Chr.18: 908172-908172 on Build GRCh38
rs757266670 Chr.18: 908398-908398 on Build GRCh38
rs965278360 Chr.18: 908432-908432 on Build GRCh38
rs751869012 Chr.18: 908391-908391 on Build GRCh38
rs774513955 Chr.18: 908238-908241 on Build GRCh38
rs1010343271 Chr.18: 908207-908207 on Build GRCh38
rs1041099913 Chr.18: 908186-908186 on Build GRCh38
rs747132092 Chr.18: 908275-908275 on Build GRCh38
rs753686646 Chr.18: 908359-908359 on Build GRCh38
rs901249310 Chr.18: 908201-908201 on Build GRCh38
rs58719451 Chr.18: 908168-908168 on Build GRCh38
rs536947825 Chr.18: 908169-908169 on Build GRCh38
rs530358705 Chr.18: 908563-908563 on Build GRCh38
rs952604069 Chr.18: 908519-908519 on Build GRCh38
rs769120911 Chr.18: 908407-908407 on Build GRCh38
rs750169852 Chr.18: 908361-908361 on Build GRCh38
rs752566792 Chr.18: 908314-908314 on Build GRCh38
rs928978 Chr.18: 908534-908534 on Build GRCh38
rs542086481 Chr.18: 908312-908312 on Build GRCh38
rs963942904 Chr.18: 908535-908535 on Build GRCh38
rs367884952 Chr.18: 908296-908296 on Build GRCh38
rs765284213 Chr.18: 908378-908378 on Build GRCh38
rs991161788 Chr.18: 908209-908209 on Build GRCh38
rs191294273 Chr.18: 908593-908593 on Build GRCh38
rs746426997 Chr.18: 908344-908344 on Build GRCh38
rs369947878 Chr.18: 908343-908343 on Build GRCh38
rs759573329 Chr.18: 908241-908251 on Build GRCh38
rs902479672 Chr.18: 908316-908316 on Build GRCh38
rs755964964 Chr.18: 908242-908242 on Build GRCh38
rs751090918 Chr.18: 908462-908462 on Build GRCh38
rs958999224 Chr.18: 908271-908271 on Build GRCh38
rs974933035 Chr.18: 908455-908455 on Build GRCh38
rs545768800 Chr.18: 908383-908383 on Build GRCh38
rs894729381 Chr.18: 908442-908442 on Build GRCh38
rs777856894 Chr.18: 908386-908386 on Build GRCh38
rs773121956 Chr.18: 908269-908269 on Build GRCh38
rs564179201 Chr.18: 908415-908415 on Build GRCh38
rs555945368 Chr.18: 908622-908622 on Build GRCh38
rs936636338 Chr.18: 908500-908500 on Build GRCh38
rs531423257 Chr.18: 908423-908423 on Build GRCh38
rs772717753 Chr.18: 908409-908409 on Build GRCh38
rs771281885 Chr.18: 908277-908277 on Build GRCh38
rs1055924822 Chr.18: 908434-908434 on Build GRCh38
rs535531202 Chr.18: 908413-908414 on Build GRCh38
rs970433185 Chr.18: 908488-908488 on Build GRCh38
rs113464270 Chr.18: 908396-908396 on Build GRCh38
rs771568197 Chr.18: 908353-908353 on Build GRCh38
rs371625594 Chr.18: 908212-908212 on Build GRCh38
rs922569717 Chr.18: 908577-908577 on Build GRCh38
rs758642838 Chr.18: 908318-908318 on Build GRCh38
rs889768084 Chr.18: 908180-908180 on Build GRCh38
rs1025273504 Chr.18: 908400-908400 on Build GRCh38
rs551501911 Chr.18: 908241-908241 on Build GRCh38
rs767349047 Chr.18: 908310-908310 on Build GRCh38
rs778067047 Chr.18: 908319-908319 on Build GRCh38
rs779263517 Chr.18: 908249-908249 on Build GRCh38
rs748429062 Chr.18: 908252-908252 on Build GRCh38
rs1020856725 Chr.18: 908221-908221 on Build GRCh38
rs1046231542 Chr.18: 908548-908548 on Build GRCh38
rs1012532896 Chr.18: 908547-908547 on Build GRCh38
rs149799520 Chr.18: 908289-908289 on Build GRCh38
rs895003276 Chr.18: 908356-908356 on Build GRCh38
rs566840802 Chr.18: 908183-908183 on Build GRCh38
rs1032184592 Chr.18: 908189-908189 on Build GRCh38
rs1034569765 Chr.18: 908260-908260 on Build GRCh38
rs139671216 Chr.18: 908288-908288 on Build GRCh38
rs1025344813 Chr.18: 908358-908358 on Build GRCh38
rs752480565 Chr.18: 908380-908380 on Build GRCh38
rs753870869 Chr.18: 908302-908302 on Build GRCh38
rs780064378 Chr.18: 908244-908244 on Build GRCh38
rs926052718 Chr.18: 908602-908602 on Build GRCh38
rs994299284 Chr.18: 908490-908490 on Build GRCh38
rs773973973 Chr.18: 908292-908292 on Build GRCh38
rs923955440 Chr.18: 908496-908496 on Build GRCh38
rs911133758 Chr.18: 908323-908323 on Build GRCh38
rs994452283 Chr.18: 908387-908387 on Build GRCh38
rs780554545 Chr.18: 908402-908402 on Build GRCh38
rs372460905 Chr.18: 908324-908324 on Build GRCh38
rs745978982 Chr.18: 908401-908401 on Build GRCh38
rs75827920 Chr.18: 908273-908273 on Build GRCh38
rs372357255 Chr.18: 908342-908342 on Build GRCh38
rs754884744 Chr.18: 908246-908246 on Build GRCh38
rs748968090 Chr.18: 908245-908245 on Build GRCh38
rs200379850 Chr.18: 908357-908357 on Build GRCh38
rs2231185 Chr.18: 908228-908228 on Build GRCh38
rs141381060 Chr.18: 908176-908176 on Build GRCh38
rs375346025 Chr.18: 908220-908220 on Build GRCh38
rs904647258 Chr.18: 908404-908404 on Build GRCh38
rs777202463 Chr.18: 908281-908281 on Build GRCh38
rs983952502 Chr.18: 908532-908532 on Build GRCh38
rs772017243 Chr.18: 908255-908255 on Build GRCh38

View Full Product Details