Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 11689326-11689868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAL
Entrez Gene ID
2774
Gene Name
G protein subunit alpha L
Gene Aliases
DYT25
UniGene
Hs.136295
Gene Chromosome Location
Chr.18: 11688956-11885685 on Build GRCh38
Transcripts
NM_182978.3 XM_006722324.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5343088 c.268A>G p.K90E Chr.18: 11689831-11689831 on Build GRCh38
6556849 c.102G>T p.P34P Chr.18: 11689665-11689665 on Build GRCh38
5959648 c.107_108insGGCCCT p.A39_P40insLA Chr.18: 11689670-11689671 on Build GRCh38
4130911 c.107C>T p.P36L Chr.18: 11689670-11689670 on Build GRCh38
4887633 c.224G>A p.R75Q Chr.18: 11689787-11689787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901017195 Chr.18: 11689525-11689525 on Build GRCh38
rs758969552 Chr.18: 11689769-11689769 on Build GRCh38
rs564907176 Chr.18: 11689713-11689713 on Build GRCh38
rs893681985 Chr.18: 11689771-11689771 on Build GRCh38
rs964964441 Chr.18: 11689656-11689656 on Build GRCh38
rs954433506 Chr.18: 11689623-11689623 on Build GRCh38
rs1044393018 Chr.18: 11689568-11689568 on Build GRCh38
rs1032625784 Chr.18: 11689600-11689600 on Build GRCh38
rs901005939 Chr.18: 11689740-11689740 on Build GRCh38
rs531745431 Chr.18: 11689676-11689677 on Build GRCh38
rs987117042 Chr.18: 11689632-11689632 on Build GRCh38
rs893639863 Chr.18: 11689601-11689601 on Build GRCh38
rs755250899 Chr.18: 11689745-11689745 on Build GRCh38
rs750015298 Chr.18: 11689695-11689695 on Build GRCh38
rs950042838 Chr.18: 11689529-11689529 on Build GRCh38
rs1047810300 Chr.18: 11689512-11689512 on Build GRCh38
rs7236061 Chr.18: 11689670-11689670 on Build GRCh38
rs776673691 Chr.18: 11689814-11689814 on Build GRCh38
rs760225719 Chr.18: 11689834-11689834 on Build GRCh38
rs753562019 Chr.18: 11689724-11689724 on Build GRCh38
rs868491990 Chr.18: 11689805-11689805 on Build GRCh38
rs61495657 Chr.18: 11689453-11689453 on Build GRCh38
rs1006776567 Chr.18: 11689518-11689518 on Build GRCh38
rs532055871 Chr.18: 11689701-11689701 on Build GRCh38
rs905132944 Chr.18: 11689572-11689572 on Build GRCh38
rs768781193 Chr.18: 11689684-11689684 on Build GRCh38
rs761832554 Chr.18: 11689829-11689829 on Build GRCh38
rs7236433 Chr.18: 11689700-11689700 on Build GRCh38
rs750891634 Chr.18: 11689813-11689814 on Build GRCh38
rs1002109765 Chr.18: 11689575-11689575 on Build GRCh38
rs1005314490 Chr.18: 11689757-11689757 on Build GRCh38
rs577163323 Chr.18: 11689671-11689676 on Build GRCh38
rs181443061 Chr.18: 11689824-11689824 on Build GRCh38
rs993990807 Chr.18: 11689749-11689749 on Build GRCh38
rs1032157087 Chr.18: 11689402-11689402 on Build GRCh38
rs370118361 Chr.18: 11689815-11689815 on Build GRCh38
rs747139554 Chr.18: 11689787-11689787 on Build GRCh38
rs890228437 Chr.18: 11689812-11689812 on Build GRCh38
rs917356429 Chr.18: 11689706-11689706 on Build GRCh38
rs950092521 Chr.18: 11689711-11689711 on Build GRCh38
rs753374463 Chr.18: 11689847-11689847 on Build GRCh38
rs765936963 Chr.18: 11689840-11689840 on Build GRCh38
rs1046793623 Chr.18: 11689756-11689756 on Build GRCh38
rs920804434 Chr.18: 11689688-11689688 on Build GRCh38
rs987902641 Chr.18: 11689422-11689422 on Build GRCh38
rs958057295 Chr.18: 11689406-11689406 on Build GRCh38
rs910818338 Chr.18: 11689480-11689480 on Build GRCh38
rs1042963344 Chr.18: 11689491-11689491 on Build GRCh38
rs767243364 Chr.18: 11689659-11689660 on Build GRCh38
rs1017206708 Chr.18: 11689652-11689652 on Build GRCh38
rs773260769 Chr.18: 11689481-11689481 on Build GRCh38
rs1023317715 Chr.18: 11689604-11689604 on Build GRCh38
rs928129701 Chr.18: 11689383-11689383 on Build GRCh38
rs889680075 Chr.18: 11689514-11689514 on Build GRCh38
rs779349517 Chr.18: 11689761-11689761 on Build GRCh38
rs867696410 Chr.18: 11689792-11689792 on Build GRCh38
rs1017262747 Chr.18: 11689822-11689822 on Build GRCh38
rs201898548 Chr.18: 11689670-11689671 on Build GRCh38
rs969044588 Chr.18: 11689775-11689775 on Build GRCh38
rs774289119 Chr.18: 11689825-11689825 on Build GRCh38
rs9303742 Chr.18: 11689523-11689523 on Build GRCh38
rs543723400 Chr.18: 11689705-11689705 on Build GRCh38
rs767065649 Chr.18: 11689683-11689683 on Build GRCh38
rs772820493 Chr.18: 11689833-11689833 on Build GRCh38
rs956895925 Chr.18: 11689626-11689626 on Build GRCh38
rs905080526 Chr.18: 11689387-11689387 on Build GRCh38
rs554303443 Chr.18: 11689493-11689493 on Build GRCh38
rs982367401 Chr.18: 11689617-11689617 on Build GRCh38
rs758880466 Chr.18: 11689814-11689831 on Build GRCh38
rs779605505 Chr.18: 11689809-11689810 on Build GRCh38
rs1005653996 Chr.18: 11689578-11689578 on Build GRCh38
rs1002054179 Chr.18: 11689399-11689399 on Build GRCh38
rs767615710 Chr.18: 11689831-11689831 on Build GRCh38
rs1040980055 Chr.18: 11689726-11689726 on Build GRCh38
rs543230597 Chr.18: 11689611-11689611 on Build GRCh38
rs915337719 Chr.18: 11689664-11689664 on Build GRCh38
rs1009415810 Chr.18: 11689776-11689776 on Build GRCh38
rs771151753 Chr.18: 11689813-11689813 on Build GRCh38
rs937952329 Chr.18: 11689737-11689737 on Build GRCh38
rs952400171 Chr.18: 11689576-11689576 on Build GRCh38
rs993876212 Chr.18: 11689527-11689527 on Build GRCh38
rs1021080904 Chr.18: 11689430-11689430 on Build GRCh38
rs1024903557 Chr.18: 11689353-11689353 on Build GRCh38
rs1009777479 Chr.18: 11689609-11689609 on Build GRCh38
rs866744675 Chr.18: 11689798-11689798 on Build GRCh38
rs973567089 Chr.18: 11689488-11689488 on Build GRCh38
rs1044443716 Chr.18: 11689719-11689719 on Build GRCh38
rs1042590918 Chr.18: 11689791-11689791 on Build GRCh38
rs920764543 Chr.18: 11689490-11689490 on Build GRCh38
rs748589252 Chr.18: 11689768-11689768 on Build GRCh38
rs761851127 Chr.18: 11689680-11689680 on Build GRCh38
rs910562901 Chr.18: 11689715-11689715 on Build GRCh38
rs577915504 Chr.18: 11689431-11689431 on Build GRCh38

View Full Product Details