Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 11751394-11751893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAL
Entrez Gene ID
2774
Gene Name
G protein subunit alpha L
Gene Aliases
DYT25
UniGene
Hs.136295
Gene Chromosome Location
Chr.18: 11688956-11885685 on Build GRCh38
Transcripts
NM_001142339.2 NM_001261443.1 NM_182978.3 XM_006722324.2

SNP Detail

SNP ID Chromosome Location
rs928899192 Chr.18: 11751644-11751644 on Build GRCh38
rs1011635381 Chr.18: 11751550-11751550 on Build GRCh38
rs978867142 Chr.18: 11751432-11751432 on Build GRCh38
rs950554973 Chr.18: 11751620-11751620 on Build GRCh38
rs934398460 Chr.18: 11751469-11751469 on Build GRCh38
rs997357231 Chr.18: 11751591-11751591 on Build GRCh38
rs1006038978 Chr.18: 11751776-11751776 on Build GRCh38
rs922441554 Chr.18: 11751510-11751510 on Build GRCh38
rs959737842 Chr.18: 11751788-11751788 on Build GRCh38
rs553992851 Chr.18: 11751536-11751536 on Build GRCh38
rs899160726 Chr.18: 11751682-11751682 on Build GRCh38
rs1014877533 Chr.18: 11751578-11751578 on Build GRCh38
rs938843138 Chr.18: 11751488-11751488 on Build GRCh38
rs933832711 Chr.18: 11751637-11751637 on Build GRCh38
rs574672025 Chr.18: 11751735-11751735 on Build GRCh38
rs895971534 Chr.18: 11751570-11751570 on Build GRCh38
rs955672389 Chr.18: 11751801-11751801 on Build GRCh38
rs911307378 Chr.18: 11751823-11751823 on Build GRCh38
rs565958288 Chr.18: 11751486-11751486 on Build GRCh38
rs560953948 Chr.18: 11751753-11751753 on Build GRCh38
rs970249011 Chr.18: 11751783-11751783 on Build GRCh38
rs973945175 Chr.18: 11751862-11751862 on Build GRCh38
rs922369286 Chr.18: 11751629-11751629 on Build GRCh38
rs889735921 Chr.18: 11751524-11751524 on Build GRCh38
rs1015945551 Chr.18: 11751777-11751777 on Build GRCh38
rs1026532352 Chr.18: 11751601-11751601 on Build GRCh38
rs1043167348 Chr.18: 11751551-11751551 on Build GRCh38
rs969679898 Chr.18: 11751635-11751635 on Build GRCh38
rs911059484 Chr.18: 11751844-11751844 on Build GRCh38
rs1007315678 Chr.18: 11751558-11751558 on Build GRCh38
rs993558684 Chr.18: 11751562-11751562 on Build GRCh38
rs549882319 Chr.18: 11751778-11751778 on Build GRCh38
rs970721225 Chr.18: 11751589-11751589 on Build GRCh38
rs955303831 Chr.18: 11751835-11751835 on Build GRCh38
rs997802540 Chr.18: 11751748-11751748 on Build GRCh38
rs1033291002 Chr.18: 11751794-11751794 on Build GRCh38
rs533439536 Chr.18: 11751596-11751596 on Build GRCh38
rs576979904 Chr.18: 11751574-11751574 on Build GRCh38
rs973834482 Chr.18: 11751695-11751718 on Build GRCh38
rs979783153 Chr.18: 11751785-11751785 on Build GRCh38
rs955745605 Chr.18: 11751603-11751603 on Build GRCh38
rs938964527 Chr.18: 11751646-11751646 on Build GRCh38
rs868384412 Chr.18: 11751763-11751763 on Build GRCh38
rs989447126 Chr.18: 11751799-11751799 on Build GRCh38
rs777100069 Chr.18: 11751836-11751836 on Build GRCh38
rs185268200 Chr.18: 11751643-11751643 on Build GRCh38
rs925656317 Chr.18: 11751447-11751447 on Build GRCh38
rs1029386222 Chr.18: 11751834-11751834 on Build GRCh38
rs1009943857 Chr.18: 11751805-11751805 on Build GRCh38
rs542651738 Chr.18: 11751740-11751740 on Build GRCh38
rs901144510 Chr.18: 11751870-11751870 on Build GRCh38
rs559415228 Chr.18: 11751657-11751657 on Build GRCh38
rs901752598 Chr.18: 11751742-11751742 on Build GRCh38
rs1049646587 Chr.18: 11751523-11751523 on Build GRCh38
rs978409045 Chr.18: 11751597-11751597 on Build GRCh38
rs941293877 Chr.18: 11751658-11751658 on Build GRCh38
rs112400833 Chr.18: 11751600-11751600 on Build GRCh38
rs966381820 Chr.18: 11751587-11751587 on Build GRCh38
rs1044958337 Chr.18: 11751770-11751770 on Build GRCh38
rs761058872 Chr.18: 11751710-11751710 on Build GRCh38
rs988546101 Chr.18: 11751628-11751628 on Build GRCh38
rs1038110587 Chr.18: 11751537-11751537 on Build GRCh38
rs988619602 Chr.18: 11751481-11751481 on Build GRCh38
rs1032696570 Chr.18: 11751599-11751599 on Build GRCh38
rs899232711 Chr.18: 11751545-11751545 on Build GRCh38
rs755419983 Chr.18: 11751491-11751491 on Build GRCh38
rs867440799 Chr.18: 11751672-11751672 on Build GRCh38
rs978949969 Chr.18: 11751810-11751810 on Build GRCh38
rs1023207935 Chr.18: 11751581-11751581 on Build GRCh38
rs1026388644 Chr.18: 11751564-11751564 on Build GRCh38
rs565342420 Chr.18: 11751839-11751839 on Build GRCh38
rs764760074 Chr.18: 11751624-11751624 on Build GRCh38
rs754329094 Chr.18: 11751452-11751452 on Build GRCh38
rs962605631 Chr.18: 11751851-11751851 on Build GRCh38
rs1000868770 Chr.18: 11751790-11751790 on Build GRCh38
rs943350133 Chr.18: 11751529-11751529 on Build GRCh38
rs1035728399 Chr.18: 11751787-11751787 on Build GRCh38
rs933839676 Chr.18: 11751520-11751520 on Build GRCh38
rs1053733517 Chr.18: 11751534-11751534 on Build GRCh38
rs947523928 Chr.18: 11751803-11751803 on Build GRCh38
rs531550231 Chr.18: 11751765-11751765 on Build GRCh38
rs889769791 Chr.18: 11751554-11751554 on Build GRCh38

View Full Product Details