Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 11753532-11754073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAL
Entrez Gene ID
2774
Gene Name
G protein subunit alpha L
Gene Aliases
DYT25
UniGene
Hs.136295
Gene Chromosome Location
Chr.18: 11688956-11885685 on Build GRCh38
Transcripts
NM_001142339.2 NM_001261443.1 NM_182978.3 XM_006722324.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4918871 c.608A>G p.D203G Chr.18: 11753929-11753929 on Build GRCh38
1680238 c.356A>T p.K119M Chr.18: 11753908-11753908 on Build GRCh38
338908 c.575C>G p.S192* Chr.18: 11753896-11753896 on Build GRCh38
4885350 c.479G>A p.R160Q Chr.18: 11753657-11753657 on Build GRCh38
3388360 c.533T>C p.I178T Chr.18: 11753854-11753854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759495975 Chr.18: 11753945-11753945 on Build GRCh38
rs746681260 Chr.18: 11753867-11753867 on Build GRCh38
rs573304987 Chr.18: 11753595-11753595 on Build GRCh38
rs202011060 Chr.18: 11753711-11753711 on Build GRCh38
rs376063491 Chr.18: 11753886-11753886 on Build GRCh38
rs770838797 Chr.18: 11753802-11753802 on Build GRCh38
rs777529067 Chr.18: 11753864-11753864 on Build GRCh38
rs760133344 Chr.18: 11753724-11753724 on Build GRCh38
rs752199100 Chr.18: 11753853-11753853 on Build GRCh38
rs775495200 Chr.18: 11753777-11753777 on Build GRCh38
rs769856826 Chr.18: 11753691-11753692 on Build GRCh38
rs770258090 Chr.18: 11753900-11753900 on Build GRCh38
rs746368758 Chr.18: 11753891-11753891 on Build GRCh38
rs763709763 Chr.18: 11753981-11753981 on Build GRCh38
rs112485987 Chr.18: 11753749-11753749 on Build GRCh38
rs372718658 Chr.18: 11753696-11753696 on Build GRCh38
rs373310932 Chr.18: 11753718-11753718 on Build GRCh38
rs746198959 Chr.18: 11753974-11753974 on Build GRCh38
rs375535450 Chr.18: 11753715-11753715 on Build GRCh38
rs398122926 Chr.18: 11753835-11753836 on Build GRCh38
rs535954358 Chr.18: 11754025-11754025 on Build GRCh38
rs187797500 Chr.18: 11753799-11753799 on Build GRCh38
rs182262644 Chr.18: 11753861-11753861 on Build GRCh38
rs754431169 Chr.18: 11753659-11753659 on Build GRCh38
rs763219592 Chr.18: 11753779-11753779 on Build GRCh38
rs201531513 Chr.18: 11753969-11753969 on Build GRCh38
rs777228288 Chr.18: 11753721-11753721 on Build GRCh38
rs1042482502 Chr.18: 11753760-11753760 on Build GRCh38
rs774048722 Chr.18: 11753803-11753803 on Build GRCh38
rs771680652 Chr.18: 11753712-11753712 on Build GRCh38
rs747832369 Chr.18: 11753705-11753705 on Build GRCh38
rs374773009 Chr.18: 11753788-11753788 on Build GRCh38
rs201760953 Chr.18: 11753987-11753987 on Build GRCh38
rs989285758 Chr.18: 11753568-11753568 on Build GRCh38
rs9963594 Chr.18: 11753993-11753993 on Build GRCh38
rs778473594 Chr.18: 11753841-11753841 on Build GRCh38
rs1057519098 Chr.18: 11753639-11753640 on Build GRCh38
rs557461258 Chr.18: 11754031-11754031 on Build GRCh38
rs753692101 Chr.18: 11753942-11753942 on Build GRCh38
rs370473544 Chr.18: 11753948-11753948 on Build GRCh38
rs762981193 Chr.18: 11753909-11753909 on Build GRCh38
rs762815069 Chr.18: 11753798-11753798 on Build GRCh38
rs775615136 Chr.18: 11753901-11753901 on Build GRCh38
rs199675814 Chr.18: 11753609-11753609 on Build GRCh38
rs76888098 Chr.18: 11753912-11753912 on Build GRCh38
rs752714566 Chr.18: 11753954-11753954 on Build GRCh38
rs72865259 Chr.18: 11754035-11754035 on Build GRCh38
rs8095592 Chr.18: 11753717-11753717 on Build GRCh38
rs374549917 Chr.18: 11753625-11753625 on Build GRCh38
rs747231872 Chr.18: 11754012-11754012 on Build GRCh38
rs762031221 Chr.18: 11753936-11753936 on Build GRCh38
rs902622873 Chr.18: 11753778-11753778 on Build GRCh38
rs756788642 Chr.18: 11753728-11753728 on Build GRCh38
rs371094261 Chr.18: 11753826-11753826 on Build GRCh38
rs774502271 Chr.18: 11753918-11753918 on Build GRCh38
rs753798643 Chr.18: 11753628-11753628 on Build GRCh38
rs781693236 Chr.18: 11753888-11753888 on Build GRCh38
rs757795779 Chr.18: 11753956-11753956 on Build GRCh38
rs754942072 Chr.18: 11753828-11753828 on Build GRCh38
rs766177601 Chr.18: 11753821-11753821 on Build GRCh38
rs372560141 Chr.18: 11753700-11753700 on Build GRCh38
rs1015814681 Chr.18: 11753927-11753927 on Build GRCh38
rs199647946 Chr.18: 11753991-11753991 on Build GRCh38
rs959087602 Chr.18: 11754003-11754003 on Build GRCh38
rs192684515 Chr.18: 11753800-11753800 on Build GRCh38
rs1027050665 Chr.18: 11753845-11753845 on Build GRCh38
rs766162763 Chr.18: 11753941-11753941 on Build GRCh38

View Full Product Details