Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12328856-12329361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFG3L2
Entrez Gene ID
10939
Gene Name
AFG3 like matrix AAA peptidase subunit 2
Gene Aliases
SCA28, SPAX5
UniGene
Hs.726355
Gene Chromosome Location
Chr.18: 12328944-12377309 on Build GRCh38
Transcripts
NM_006796.2 XM_011525601.2 NM_001303525.1

Gene Symbol
TUBB6
Entrez Gene ID
84617
Gene Name
tubulin beta 6 class V
Gene Aliases
HsT1601, TUBB-5
UniGene
Hs.193491
Gene Chromosome Location
Chr.18: 12307669-12329826 on Build GRCh38
Transcripts
NM_006796.2 XM_011525601.2 NM_001303525.1

SNP Detail

SNP ID Chromosome Location
rs936844940 Chr.18: 12329145-12329145 on Build GRCh38
rs1045301825 Chr.18: 12329114-12329114 on Build GRCh38
rs550591123 Chr.18: 12329240-12329240 on Build GRCh38
rs757183547 Chr.18: 12329222-12329222 on Build GRCh38
rs1016135801 Chr.18: 12328935-12328935 on Build GRCh38
rs79884272 Chr.18: 12329254-12329254 on Build GRCh38
rs886053612 Chr.18: 12329192-12329192 on Build GRCh38
rs12327107 Chr.18: 12328929-12328929 on Build GRCh38
rs992211515 Chr.18: 12329072-12329072 on Build GRCh38
rs551371844 Chr.18: 12329275-12329275 on Build GRCh38
rs932981283 Chr.18: 12329192-12329192 on Build GRCh38
rs963224091 Chr.18: 12328937-12328937 on Build GRCh38
rs533285669 Chr.18: 12329261-12329261 on Build GRCh38
rs150224434 Chr.18: 12329186-12329186 on Build GRCh38
rs918684914 Chr.18: 12329040-12329040 on Build GRCh38
rs773307826 Chr.18: 12329121-12329121 on Build GRCh38
rs939147580 Chr.18: 12329282-12329282 on Build GRCh38
rs964683115 Chr.18: 12329066-12329066 on Build GRCh38
rs1018433556 Chr.18: 12329007-12329007 on Build GRCh38
rs180917336 Chr.18: 12329239-12329239 on Build GRCh38
rs533711447 Chr.18: 12329321-12329321 on Build GRCh38
rs969378273 Chr.18: 12329334-12329334 on Build GRCh38
rs754369427 Chr.18: 12329328-12329328 on Build GRCh38
rs565352114 Chr.18: 12329081-12329081 on Build GRCh38
rs546862017 Chr.18: 12329076-12329076 on Build GRCh38
rs371759084 Chr.18: 12329232-12329232 on Build GRCh38
rs1051466041 Chr.18: 12329193-12329193 on Build GRCh38
rs951448975 Chr.18: 12329068-12329068 on Build GRCh38
rs566345570 Chr.18: 12329310-12329310 on Build GRCh38
rs886053611 Chr.18: 12328980-12328980 on Build GRCh38
rs759810038 Chr.18: 12329217-12329217 on Build GRCh38
rs760718826 Chr.18: 12329168-12329168 on Build GRCh38
rs951519155 Chr.18: 12329279-12329279 on Build GRCh38
rs112783401 Chr.18: 12329303-12329303 on Build GRCh38
rs577014991 Chr.18: 12329335-12329335 on Build GRCh38
rs974112845 Chr.18: 12328953-12328953 on Build GRCh38
rs948335187 Chr.18: 12329086-12329086 on Build GRCh38
rs760930166 Chr.18: 12329281-12329281 on Build GRCh38
rs951081800 Chr.18: 12329128-12329128 on Build GRCh38
rs773895356 Chr.18: 12329220-12329220 on Build GRCh38
rs561717400 Chr.18: 12329221-12329221 on Build GRCh38
rs768114348 Chr.18: 12329160-12329160 on Build GRCh38
rs199853626 Chr.18: 12328985-12328985 on Build GRCh38
rs1052582214 Chr.18: 12329178-12329178 on Build GRCh38

View Full Product Details