Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12351007-12351511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFG3L2
Entrez Gene ID
10939
Gene Name
AFG3 like matrix AAA peptidase subunit 2
Gene Aliases
SCA28, SPAX5
UniGene
Hs.726355
Gene Chromosome Location
Chr.18: 12328944-12377309 on Build GRCh38
Transcripts
NM_006796.2 XM_011525601.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165579 c.1408G>T p.D470Y Chr.18: 12351324-12351324 on Build GRCh38
2883709 c.1384G>A p.A462T Chr.18: 12351348-12351348 on Build GRCh38
6595948 c.1358C>T p.T453I Chr.18: 12351374-12351374 on Build GRCh38
148399 c.1389G>A p.L463L Chr.18: 12351343-12351343 on Build GRCh38
986610 c.1408G>A p.D470N Chr.18: 12351324-12351324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754078029 Chr.18: 12351060-12351060 on Build GRCh38
rs767868939 Chr.18: 12351140-12351140 on Build GRCh38
rs771360106 Chr.18: 12351427-12351427 on Build GRCh38
rs748208993 Chr.18: 12351182-12351184 on Build GRCh38
rs756148691 Chr.18: 12351039-12351039 on Build GRCh38
rs745808937 Chr.18: 12351238-12351238 on Build GRCh38
rs747793869 Chr.18: 12351419-12351419 on Build GRCh38
rs762665023 Chr.18: 12351109-12351109 on Build GRCh38
rs574904377 Chr.18: 12351275-12351276 on Build GRCh38
rs750575479 Chr.18: 12351291-12351291 on Build GRCh38
rs778448352 Chr.18: 12351418-12351418 on Build GRCh38
rs781280640 Chr.18: 12351064-12351066 on Build GRCh38
rs760102437 Chr.18: 12351439-12351439 on Build GRCh38
rs758755204 Chr.18: 12351341-12351341 on Build GRCh38
rs774012113 Chr.18: 12351115-12351115 on Build GRCh38
rs368011609 Chr.18: 12351390-12351390 on Build GRCh38
rs550200835 Chr.18: 12351112-12351112 on Build GRCh38
rs755220273 Chr.18: 12351061-12351061 on Build GRCh38
rs370562426 Chr.18: 12351349-12351349 on Build GRCh38
rs773632739 Chr.18: 12351281-12351281 on Build GRCh38
rs761143170 Chr.18: 12351158-12351158 on Build GRCh38
rs774859640 Chr.18: 12351384-12351384 on Build GRCh38
rs547346676 Chr.18: 12351334-12351334 on Build GRCh38
rs562127649 Chr.18: 12351284-12351284 on Build GRCh38
rs574474685 Chr.18: 12351472-12351472 on Build GRCh38
rs752910099 Chr.18: 12351337-12351337 on Build GRCh38
rs769294488 Chr.18: 12351085-12351085 on Build GRCh38
rs868044931 Chr.18: 12351380-12351380 on Build GRCh38
rs757045676 Chr.18: 12351234-12351234 on Build GRCh38
rs762123700 Chr.18: 12351126-12351126 on Build GRCh38
rs908738247 Chr.18: 12351468-12351468 on Build GRCh38
rs373106745 Chr.18: 12351078-12351078 on Build GRCh38
rs200110947 Chr.18: 12351178-12351178 on Build GRCh38
rs376789505 Chr.18: 12351064-12351064 on Build GRCh38
rs770874216 Chr.18: 12351068-12351068 on Build GRCh38
rs118126456 Chr.18: 12351464-12351464 on Build GRCh38
rs753902034 Chr.18: 12351163-12351163 on Build GRCh38
rs888647132 Chr.18: 12351075-12351075 on Build GRCh38
rs746851204 Chr.18: 12351067-12351067 on Build GRCh38
rs377761529 Chr.18: 12351255-12351255 on Build GRCh38
rs912546325 Chr.18: 12351347-12351347 on Build GRCh38
rs1005670966 Chr.18: 12351097-12351097 on Build GRCh38
rs538784507 Chr.18: 12351471-12351471 on Build GRCh38
rs371117466 Chr.18: 12351258-12351258 on Build GRCh38
rs773542472 Chr.18: 12351421-12351421 on Build GRCh38
rs747286053 Chr.18: 12351425-12351425 on Build GRCh38
rs531301543 Chr.18: 12351159-12351159 on Build GRCh38
rs755204659 Chr.18: 12351174-12351174 on Build GRCh38
rs750868061 Chr.18: 12351142-12351142 on Build GRCh38
rs373885372 Chr.18: 12351457-12351457 on Build GRCh38
rs1020764190 Chr.18: 12351330-12351330 on Build GRCh38
rs931434708 Chr.18: 12351034-12351034 on Build GRCh38
rs762474283 Chr.18: 12351069-12351069 on Build GRCh38
rs777218796 Chr.18: 12351324-12351324 on Build GRCh38
rs920131526 Chr.18: 12351422-12351422 on Build GRCh38
rs375098002 Chr.18: 12351335-12351335 on Build GRCh38
rs768121027 Chr.18: 12351101-12351101 on Build GRCh38
rs749105981 Chr.18: 12351396-12351398 on Build GRCh38
rs182327153 Chr.18: 12351420-12351420 on Build GRCh38
rs775783680 Chr.18: 12351455-12351455 on Build GRCh38
rs776937935 Chr.18: 12351433-12351433 on Build GRCh38
rs753000167 Chr.18: 12351187-12351187 on Build GRCh38
rs571114609 Chr.18: 12351266-12351266 on Build GRCh38
rs754423066 Chr.18: 12351399-12351399 on Build GRCh38
rs549787555 Chr.18: 12351181-12351181 on Build GRCh38
rs778918433 Chr.18: 12351059-12351059 on Build GRCh38
rs377339236 Chr.18: 12351297-12351297 on Build GRCh38
rs779587743 Chr.18: 12351252-12351252 on Build GRCh38
rs761212062 Chr.18: 12351296-12351296 on Build GRCh38
rs11080572 Chr.18: 12351343-12351343 on Build GRCh38
rs914992281 Chr.18: 12351332-12351332 on Build GRCh38
rs538927944 Chr.18: 12351325-12351325 on Build GRCh38
rs529264477 Chr.18: 12351289-12351289 on Build GRCh38
rs1004231616 Chr.18: 12351474-12351474 on Build GRCh38
rs745742578 Chr.18: 12351451-12351452 on Build GRCh38
rs772411404 Chr.18: 12351270-12351270 on Build GRCh38
rs186981770 Chr.18: 12351453-12351453 on Build GRCh38
rs769692015 Chr.18: 12351246-12351246 on Build GRCh38
rs1032710173 Chr.18: 12351295-12351295 on Build GRCh38
rs867453730 Chr.18: 12351377-12351377 on Build GRCh38
rs749249858 Chr.18: 12351391-12351391 on Build GRCh38
rs969627467 Chr.18: 12351233-12351233 on Build GRCh38
rs779230346 Chr.18: 12351176-12351176 on Build GRCh38
rs751832459 Chr.18: 12351458-12351458 on Build GRCh38
rs763241242 Chr.18: 12351456-12351456 on Build GRCh38
rs967408146 Chr.18: 12351331-12351331 on Build GRCh38
rs756058528 Chr.18: 12351356-12351356 on Build GRCh38
rs373273761 Chr.18: 12351264-12351264 on Build GRCh38
rs374874087 Chr.18: 12351379-12351379 on Build GRCh38
rs773169376 Chr.18: 12351388-12351390 on Build GRCh38

View Full Product Details