Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 21535813-21536310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESCO1
Entrez Gene ID
114799
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 1
Gene Aliases
A930014I12Rik, CTF, ECO1, EFO1, ESO1
UniGene
Hs.464733
Gene Chromosome Location
Chr.18: 21529281-21601015 on Build GRCh38
Transcripts
NM_052911.2 XM_011525798.1 XM_011525799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6656300 c.2171C>T p.A724V Chr.18: 21536058-21536058 on Build GRCh38
708345 c.2172G>A p.A724A Chr.18: 21536057-21536057 on Build GRCh38
1680257 c.2050G>A p.E684K Chr.18: 21536179-21536179 on Build GRCh38
368088 c.2092G>T p.A698S Chr.18: 21536137-21536137 on Build GRCh38
385441 c.2052G>C p.E684D Chr.18: 21536177-21536177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200076587 Chr.18: 21536041-21536041 on Build GRCh38
rs550861984 Chr.18: 21535942-21535942 on Build GRCh38
rs377543839 Chr.18: 21536167-21536167 on Build GRCh38
rs762284408 Chr.18: 21536204-21536204 on Build GRCh38
rs570784002 Chr.18: 21535951-21535951 on Build GRCh38
rs938983386 Chr.18: 21535910-21535910 on Build GRCh38
rs373928942 Chr.18: 21536129-21536129 on Build GRCh38
rs1045801299 Chr.18: 21536206-21536206 on Build GRCh38
rs139003214 Chr.18: 21535988-21535988 on Build GRCh38
rs1043504917 Chr.18: 21535953-21535953 on Build GRCh38
rs1026306382 Chr.18: 21536010-21536010 on Build GRCh38
rs4800780 Chr.18: 21535935-21535935 on Build GRCh38
rs574734522 Chr.18: 21535862-21535862 on Build GRCh38
rs762231208 Chr.18: 21536085-21536085 on Build GRCh38
rs748993198 Chr.18: 21536195-21536195 on Build GRCh38
rs184900300 Chr.18: 21535990-21535990 on Build GRCh38
rs751385732 Chr.18: 21536141-21536141 on Build GRCh38
rs774100819 Chr.18: 21536201-21536201 on Build GRCh38
rs960623375 Chr.18: 21536229-21536229 on Build GRCh38
rs775073407 Chr.18: 21536035-21536035 on Build GRCh38
rs769282537 Chr.18: 21536032-21536032 on Build GRCh38
rs755170614 Chr.18: 21536138-21536138 on Build GRCh38
rs750317804 Chr.18: 21535978-21535978 on Build GRCh38
rs992064356 Chr.18: 21536279-21536279 on Build GRCh38
rs781142646 Chr.18: 21536027-21536027 on Build GRCh38
rs774828921 Chr.18: 21536075-21536075 on Build GRCh38
rs769195779 Chr.18: 21536069-21536069 on Build GRCh38
rs1053166878 Chr.18: 21536274-21536274 on Build GRCh38
rs368120711 Chr.18: 21536218-21536218 on Build GRCh38
rs532912309 Chr.18: 21535979-21535979 on Build GRCh38
rs35830116 Chr.18: 21536175-21536175 on Build GRCh38
rs538070159 Chr.18: 21536286-21536286 on Build GRCh38
rs770823437 Chr.18: 21536011-21536011 on Build GRCh38
rs200593486 Chr.18: 21535993-21535993 on Build GRCh38
rs777584527 Chr.18: 21536140-21536140 on Build GRCh38
rs561945279 Chr.18: 21535911-21535911 on Build GRCh38
rs768163703 Chr.18: 21536090-21536090 on Build GRCh38
rs527759945 Chr.18: 21535989-21535989 on Build GRCh38
rs769246270 Chr.18: 21536189-21536189 on Build GRCh38
rs368988268 Chr.18: 21536124-21536124 on Build GRCh38
rs369669502 Chr.18: 21536180-21536180 on Build GRCh38
rs569397951 Chr.18: 21536121-21536121 on Build GRCh38
rs199553670 Chr.18: 21536123-21536123 on Build GRCh38
rs770222976 Chr.18: 21536196-21536196 on Build GRCh38
rs745385928 Chr.18: 21536028-21536028 on Build GRCh38
rs761228362 Chr.18: 21536225-21536225 on Build GRCh38
rs926569233 Chr.18: 21535889-21535889 on Build GRCh38
rs201053826 Chr.18: 21536034-21536034 on Build GRCh38
rs998950635 Chr.18: 21536248-21536248 on Build GRCh38
rs373973338 Chr.18: 21536126-21536126 on Build GRCh38
rs745815213 Chr.18: 21536188-21536188 on Build GRCh38
rs768458264 Chr.18: 21536200-21536200 on Build GRCh38
rs762654521 Chr.18: 21536057-21536057 on Build GRCh38
rs772412648 Chr.18: 21536217-21536217 on Build GRCh38
rs779594072 Chr.18: 21536194-21536194 on Build GRCh38
rs773981355 Chr.18: 21536029-21536029 on Build GRCh38

View Full Product Details