Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22171602-22172089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA6
Entrez Gene ID
2627
Gene Name
GATA binding protein 6
Gene Aliases
-
UniGene
Hs.514746
Gene Chromosome Location
Chr.18: 22169437-22202528 on Build GRCh38
Transcripts
NM_005257.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6395230 c.916C>T p.R306C Chr.18: 22172060-22172060 on Build GRCh38
6035296 c.895_930del36 p.L300_S311del12 Chr.18: 22172039-22172074 on Build GRCh38
7339860 c.655G>A p.A219T Chr.18: 22171799-22171799 on Build GRCh38
6386896 c.923C>T p.P308L Chr.18: 22172067-22172067 on Build GRCh38
4413073 c.885C>T p.G295G Chr.18: 22172029-22172029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs387906814 Chr.18: 22171736-22171736 on Build GRCh38
rs759587190 Chr.18: 22171669-22171669 on Build GRCh38
rs1014349671 Chr.18: 22171999-22171999 on Build GRCh38
rs769277383 Chr.18: 22171660-22171661 on Build GRCh38
rs1031755167 Chr.18: 22171879-22171879 on Build GRCh38
rs762101807 Chr.18: 22171681-22171682 on Build GRCh38
rs1039427988 Chr.18: 22171847-22171847 on Build GRCh38
rs771297576 Chr.18: 22171621-22171621 on Build GRCh38
rs941759643 Chr.18: 22171666-22171667 on Build GRCh38
rs387906815 Chr.18: 22171677-22171677 on Build GRCh38
rs904551659 Chr.18: 22171850-22171850 on Build GRCh38
rs929089960 Chr.18: 22171914-22171914 on Build GRCh38
rs926270539 Chr.18: 22171909-22171909 on Build GRCh38
rs753682981 Chr.18: 22171662-22171662 on Build GRCh38
rs919012580 Chr.18: 22171912-22171912 on Build GRCh38
rs387906816 Chr.18: 22171695-22171695 on Build GRCh38
rs770251106 Chr.18: 22171643-22171643 on Build GRCh38
rs1024528816 Chr.18: 22171894-22171894 on Build GRCh38
rs904519303 Chr.18: 22171983-22171983 on Build GRCh38
rs1038983311 Chr.18: 22171968-22171968 on Build GRCh38
rs1000153828 Chr.18: 22171993-22171993 on Build GRCh38
rs568069587 Chr.18: 22171935-22171935 on Build GRCh38
rs775485094 Chr.18: 22171644-22171644 on Build GRCh38
rs897655941 Chr.18: 22171682-22171682 on Build GRCh38
rs911752926 Chr.18: 22171919-22171919 on Build GRCh38
rs774639835 Chr.18: 22171646-22171646 on Build GRCh38
rs1000227951 Chr.18: 22171858-22171858 on Build GRCh38
rs777216159 Chr.18: 22171664-22171678 on Build GRCh38
rs777087132 Chr.18: 22171719-22171719 on Build GRCh38
rs1024878588 Chr.18: 22172001-22172001 on Build GRCh38
rs185325359 Chr.18: 22171995-22171995 on Build GRCh38
rs890377104 Chr.18: 22171810-22171810 on Build GRCh38
rs751281703 Chr.18: 22171725-22171725 on Build GRCh38
rs917494899 Chr.18: 22171647-22171647 on Build GRCh38
rs559922666 Chr.18: 22171625-22171625 on Build GRCh38
rs980861173 Chr.18: 22171651-22171651 on Build GRCh38
rs943267073 Chr.18: 22171963-22171963 on Build GRCh38
rs949048690 Chr.18: 22171648-22171648 on Build GRCh38
rs1007535542 Chr.18: 22171814-22171814 on Build GRCh38
rs1051642564 Chr.18: 22171771-22171771 on Build GRCh38
rs1014382084 Chr.18: 22171887-22171887 on Build GRCh38
rs35248194 Chr.18: 22171626-22171626 on Build GRCh38
rs1052045980 Chr.18: 22171918-22171918 on Build GRCh38
rs746437124 Chr.18: 22171642-22171642 on Build GRCh38
rs956166871 Chr.18: 22171881-22171881 on Build GRCh38
rs587777710 Chr.18: 22171856-22171856 on Build GRCh38
rs761885207 Chr.18: 22171657-22171657 on Build GRCh38
rs755907806 Chr.18: 22171631-22171631 on Build GRCh38

View Full Product Details