Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23136520-23137008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABLES1
Entrez Gene ID
91768
Gene Name
Cdk5 and Abl enzyme substrate 1
Gene Aliases
CABL1, CABLES, HsT2563, IK3-1
UniGene
Hs.11108
Gene Chromosome Location
Chr.18: 23134564-23260470 on Build GRCh38
Transcripts
NM_001100619.2 NM_001256438.1 NR_023359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6148149 c.800G>T p.G267V Chr.18: 23136562-23136562 on Build GRCh38
1247056 c.784_794del11 p.S262fs*34 Chr.18: 23136546-23136556 on Build GRCh38
6082277 c.845+1G>T p.? Chr.18: 23136608-23136608 on Build GRCh38
5464270 c.806G>A p.G269D Chr.18: 23136568-23136568 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777942979 Chr.18: 23136648-23136648 on Build GRCh38
rs745630739 Chr.18: 23136546-23136546 on Build GRCh38
rs559323349 Chr.18: 23136794-23136794 on Build GRCh38
rs540638285 Chr.18: 23136773-23136773 on Build GRCh38
rs188642060 Chr.18: 23136890-23136890 on Build GRCh38
rs902199718 Chr.18: 23136672-23136672 on Build GRCh38
rs745745791 Chr.18: 23136652-23136652 on Build GRCh38
rs373320155 Chr.18: 23136650-23136650 on Build GRCh38
rs574838497 Chr.18: 23136912-23136912 on Build GRCh38
rs755442114 Chr.18: 23136633-23136633 on Build GRCh38
rs761579105 Chr.18: 23136587-23136587 on Build GRCh38
rs768294719 Chr.18: 23136578-23136578 on Build GRCh38
rs184899180 Chr.18: 23136884-23136884 on Build GRCh38
rs769812959 Chr.18: 23136549-23136549 on Build GRCh38
rs548906024 Chr.18: 23136666-23136666 on Build GRCh38
rs944669161 Chr.18: 23136810-23136810 on Build GRCh38
rs1031744081 Chr.18: 23136887-23136887 on Build GRCh38
rs750837884 Chr.18: 23136601-23136601 on Build GRCh38
rs775286667 Chr.18: 23136559-23136559 on Build GRCh38
rs543859343 Chr.18: 23136985-23136985 on Build GRCh38
rs915375138 Chr.18: 23136734-23136734 on Build GRCh38
rs986264530 Chr.18: 23136955-23136955 on Build GRCh38
rs766820007 Chr.18: 23136680-23136680 on Build GRCh38
rs989946937 Chr.18: 23136703-23136703 on Build GRCh38
rs1040773362 Chr.18: 23136833-23136833 on Build GRCh38
rs984139952 Chr.18: 23136749-23136749 on Build GRCh38
rs773931728 Chr.18: 23136579-23136579 on Build GRCh38
rs754311831 Chr.18: 23136631-23136631 on Build GRCh38
rs778991998 Chr.18: 23136638-23136638 on Build GRCh38
rs753357044 Chr.18: 23136855-23136855 on Build GRCh38
rs762583399 Chr.18: 23136567-23136567 on Build GRCh38
rs567680463 Chr.18: 23136685-23136685 on Build GRCh38
rs932029564 Chr.18: 23136809-23136809 on Build GRCh38
rs150139393 Chr.18: 23136699-23136699 on Build GRCh38
rs952186206 Chr.18: 23136742-23136742 on Build GRCh38
rs369906945 Chr.18: 23136590-23136590 on Build GRCh38
rs955754370 Chr.18: 23136902-23136902 on Build GRCh38
rs142120848 Chr.18: 23136664-23136664 on Build GRCh38
rs1012765288 Chr.18: 23136871-23136871 on Build GRCh38
rs944713809 Chr.18: 23136641-23136641 on Build GRCh38
rs968151483 Chr.18: 23136936-23136936 on Build GRCh38
rs1042714926 Chr.18: 23136899-23136899 on Build GRCh38
rs769464742 Chr.18: 23136653-23136653 on Build GRCh38
rs552509113 Chr.18: 23136539-23136539 on Build GRCh38
rs772023920 Chr.18: 23136646-23136646 on Build GRCh38
rs552872063 Chr.18: 23136882-23136882 on Build GRCh38
rs990280402 Chr.18: 23136906-23136906 on Build GRCh38
rs752729389 Chr.18: 23136639-23136639 on Build GRCh38
rs111830385 Chr.18: 23136842-23136842 on Build GRCh38
rs760942253 Chr.18: 23136606-23136606 on Build GRCh38
rs4800451 Chr.18: 23136841-23136841 on Build GRCh38
rs368272682 Chr.18: 23136784-23136784 on Build GRCh38
rs559928727 Chr.18: 23136644-23136644 on Build GRCh38
rs966460891 Chr.18: 23136781-23136781 on Build GRCh38
rs1022850490 Chr.18: 23136874-23136874 on Build GRCh38
rs760772501 Chr.18: 23136673-23136673 on Build GRCh38
rs765653143 Chr.18: 23136878-23136878 on Build GRCh38
rs115481168 Chr.18: 23136907-23136907 on Build GRCh38
rs946902467 Chr.18: 23136764-23136764 on Build GRCh38
rs192886399 Chr.18: 23136863-23136863 on Build GRCh38
rs113759905 Chr.18: 23136667-23136667 on Build GRCh38
rs971019092 Chr.18: 23136668-23136668 on Build GRCh38
rs929368063 Chr.18: 23136853-23136853 on Build GRCh38
rs73964439 Chr.18: 23136796-23136796 on Build GRCh38
rs990191501 Chr.18: 23136812-23136812 on Build GRCh38

View Full Product Details