Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 30992298-30992808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC3
Entrez Gene ID
1825
Gene Name
desmocollin 3
Gene Aliases
CDHF3, DSC, DSC1, DSC2, DSC4, HT-CP
UniGene
Hs.41690
Gene Chromosome Location
Chr.18: 30989365-31042815 on Build GRCh38
Transcripts
NM_001941.4 NM_024423.3

SNP Detail

SNP ID Chromosome Location
rs920083064 Chr.18: 30992484-30992484 on Build GRCh38
rs767102278 Chr.18: 30992492-30992492 on Build GRCh38
rs888710180 Chr.18: 30992445-30992445 on Build GRCh38
rs753505168 Chr.18: 30992667-30992667 on Build GRCh38
rs762742510 Chr.18: 30992460-30992460 on Build GRCh38
rs561003473 Chr.18: 30992468-30992468 on Build GRCh38
rs959892065 Chr.18: 30992636-30992636 on Build GRCh38
rs938513049 Chr.18: 30992343-30992343 on Build GRCh38
rs1032173152 Chr.18: 30992359-30992359 on Build GRCh38
rs1012156924 Chr.18: 30992404-30992404 on Build GRCh38
rs148696475 Chr.18: 30992580-30992580 on Build GRCh38
rs906403455 Chr.18: 30992657-30992657 on Build GRCh38
rs746345147 Chr.18: 30992743-30992743 on Build GRCh38
rs1053899969 Chr.18: 30992591-30992591 on Build GRCh38
rs766340162 Chr.18: 30992483-30992483 on Build GRCh38
rs894549586 Chr.18: 30992362-30992362 on Build GRCh38
rs71361309 Chr.18: 30992417-30992417 on Build GRCh38
rs144015964 Chr.18: 30992485-30992485 on Build GRCh38
rs76343607 Chr.18: 30992560-30992560 on Build GRCh38
rs555662376 Chr.18: 30992705-30992705 on Build GRCh38
rs528552938 Chr.18: 30992595-30992595 on Build GRCh38
rs977347494 Chr.18: 30992603-30992603 on Build GRCh38
rs760699188 Chr.18: 30992349-30992349 on Build GRCh38
rs375987296 Chr.18: 30992462-30992462 on Build GRCh38
rs973111051 Chr.18: 30992673-30992673 on Build GRCh38
rs529346780 Chr.18: 30992461-30992461 on Build GRCh38
rs760744901 Chr.18: 30992456-30992456 on Build GRCh38
rs998777760 Chr.18: 30992773-30992773 on Build GRCh38
rs539402005 Chr.18: 30992510-30992510 on Build GRCh38
rs1028678263 Chr.18: 30992716-30992716 on Build GRCh38
rs940001319 Chr.18: 30992699-30992699 on Build GRCh38
rs998347070 Chr.18: 30992478-30992478 on Build GRCh38
rs78985117 Chr.18: 30992665-30992665 on Build GRCh38
rs902550614 Chr.18: 30992552-30992552 on Build GRCh38
rs944309111 Chr.18: 30992498-30992498 on Build GRCh38
rs909845881 Chr.18: 30992678-30992678 on Build GRCh38
rs895013015 Chr.18: 30992363-30992363 on Build GRCh38
rs948756525 Chr.18: 30992411-30992411 on Build GRCh38
rs948805692 Chr.18: 30992669-30992669 on Build GRCh38
rs1042739538 Chr.18: 30992516-30992516 on Build GRCh38
rs1042406525 Chr.18: 30992334-30992334 on Build GRCh38
rs973161830 Chr.18: 30992482-30992482 on Build GRCh38
rs142599311 Chr.18: 30992355-30992355 on Build GRCh38
rs901413363 Chr.18: 30992723-30992723 on Build GRCh38
rs779299098 Chr.18: 30992676-30992676 on Build GRCh38
rs963095970 Chr.18: 30992481-30992481 on Build GRCh38
rs983489518 Chr.18: 30992443-30992443 on Build GRCh38
rs1037321817 Chr.18: 30992720-30992720 on Build GRCh38
rs755594923 Chr.18: 30992622-30992622 on Build GRCh38
rs776357256 Chr.18: 30992610-30992610 on Build GRCh38
rs1007650287 Chr.18: 30992509-30992509 on Build GRCh38
rs764690013 Chr.18: 30992330-30992332 on Build GRCh38
rs768871833 Chr.18: 30992365-30992365 on Build GRCh38
rs1021256273 Chr.18: 30992558-30992558 on Build GRCh38
rs113769668 Chr.18: 30992672-30992672 on Build GRCh38
rs565847275 Chr.18: 30992495-30992495 on Build GRCh38

View Full Product Details