Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 30993016-30993560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC3
Entrez Gene ID
1825
Gene Name
desmocollin 3
Gene Aliases
CDHF3, DSC, DSC1, DSC2, DSC4, HT-CP
UniGene
Hs.41690
Gene Chromosome Location
Chr.18: 30989365-31042815 on Build GRCh38
Transcripts
NM_001941.4 NM_024423.3

SNP Detail

SNP ID Chromosome Location
rs770825092 Chr.18: 30993257-30993257 on Build GRCh38
rs140760517 Chr.18: 30993291-30993291 on Build GRCh38
rs76488122 Chr.18: 30993121-30993121 on Build GRCh38
rs911494650 Chr.18: 30993470-30993470 on Build GRCh38
rs961425146 Chr.18: 30993235-30993235 on Build GRCh38
rs141625117 Chr.18: 30993450-30993450 on Build GRCh38
rs1016742023 Chr.18: 30993428-30993428 on Build GRCh38
rs533233119 Chr.18: 30993478-30993478 on Build GRCh38
rs969951936 Chr.18: 30993209-30993209 on Build GRCh38
rs191216022 Chr.18: 30993098-30993098 on Build GRCh38
rs551346477 Chr.18: 30993059-30993059 on Build GRCh38
rs916624083 Chr.18: 30993167-30993167 on Build GRCh38
rs946927454 Chr.18: 30993279-30993279 on Build GRCh38
rs972736995 Chr.18: 30993244-30993244 on Build GRCh38
rs561190888 Chr.18: 30993422-30993422 on Build GRCh38
rs1020192111 Chr.18: 30993042-30993042 on Build GRCh38
rs144607647 Chr.18: 30993313-30993313 on Build GRCh38
rs74990424 Chr.18: 30993424-30993424 on Build GRCh38
rs79252469 Chr.18: 30993374-30993374 on Build GRCh38
rs905757719 Chr.18: 30993417-30993417 on Build GRCh38
rs1050045450 Chr.18: 30993250-30993250 on Build GRCh38
rs946864108 Chr.18: 30993101-30993101 on Build GRCh38
rs984349308 Chr.18: 30993211-30993211 on Build GRCh38
rs752177728 Chr.18: 30993375-30993379 on Build GRCh38
rs911449824 Chr.18: 30993275-30993275 on Build GRCh38
rs567874997 Chr.18: 30993161-30993161 on Build GRCh38
rs192404929 Chr.18: 30993479-30993479 on Build GRCh38
rs985762091 Chr.18: 30993270-30993270 on Build GRCh38
rs946931349 Chr.18: 30993477-30993477 on Build GRCh38
rs187978005 Chr.18: 30993321-30993321 on Build GRCh38
rs904261264 Chr.18: 30993104-30993104 on Build GRCh38
rs935501925 Chr.18: 30993488-30993488 on Build GRCh38
rs775298845 Chr.18: 30993492-30993492 on Build GRCh38
rs1017054680 Chr.18: 30993221-30993221 on Build GRCh38
rs969439723 Chr.18: 30993468-30993468 on Build GRCh38
rs899055720 Chr.18: 30993224-30993224 on Build GRCh38
rs1031270942 Chr.18: 30993128-30993128 on Build GRCh38
rs533266477 Chr.18: 30993040-30993040 on Build GRCh38
rs895747408 Chr.18: 30993187-30993187 on Build GRCh38
rs183831014 Chr.18: 30993179-30993179 on Build GRCh38
rs145575883 Chr.18: 30993452-30993452 on Build GRCh38
rs1022910455 Chr.18: 30993403-30993403 on Build GRCh38
rs151037205 Chr.18: 30993077-30993077 on Build GRCh38
rs568032829 Chr.18: 30993429-30993429 on Build GRCh38
rs1023302337 Chr.18: 30993205-30993205 on Build GRCh38
rs528897289 Chr.18: 30993442-30993442 on Build GRCh38
rs138894174 Chr.18: 30993202-30993202 on Build GRCh38
rs972851166 Chr.18: 30993445-30993445 on Build GRCh38
rs369896657 Chr.18: 30993139-30993139 on Build GRCh38
rs1014328801 Chr.18: 30993389-30993389 on Build GRCh38
rs985846080 Chr.18: 30993469-30993469 on Build GRCh38
rs1050349706 Chr.18: 30993063-30993063 on Build GRCh38
rs774324199 Chr.18: 30993443-30993443 on Build GRCh38
rs538771968 Chr.18: 30993280-30993280 on Build GRCh38
rs958274707 Chr.18: 30993411-30993411 on Build GRCh38
rs549927006 Chr.18: 30993200-30993200 on Build GRCh38
rs933980621 Chr.18: 30993259-30993259 on Build GRCh38
rs904295770 Chr.18: 30993292-30993292 on Build GRCh38
rs17656284 Chr.18: 30993201-30993201 on Build GRCh38
rs537343676 Chr.18: 30993103-30993103 on Build GRCh38
rs889997625 Chr.18: 30993095-30993095 on Build GRCh38

View Full Product Details