Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31388103-31388636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1-AS1
Entrez Gene ID
101927718
Gene Name
DSG1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.625473
Gene Chromosome Location
Chr.18: 31343368-31426988 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
UniGene
Hs.407618
Gene Chromosome Location
Chr.18: 31376777-31415791 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6556906 c.315C>T p.R105R Chr.18: 31388465-31388465 on Build GRCh38
5692626 c.257G>T p.R86L Chr.18: 31388407-31388407 on Build GRCh38
393341 c.314G>T p.R105L Chr.18: 31388464-31388464 on Build GRCh38
352031 c.340G>A p.V114M Chr.18: 31388490-31388490 on Build GRCh38
3890899 c.271G>A p.G91R Chr.18: 31388421-31388421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781483894 Chr.18: 31388376-31388376 on Build GRCh38
rs757836441 Chr.18: 31388346-31388346 on Build GRCh38
rs756647119 Chr.18: 31388445-31388445 on Build GRCh38
rs770937760 Chr.18: 31388471-31388471 on Build GRCh38
rs753294135 Chr.18: 31388425-31388425 on Build GRCh38
rs1022420815 Chr.18: 31388137-31388137 on Build GRCh38
rs16959856 Chr.18: 31388408-31388408 on Build GRCh38
rs1016569161 Chr.18: 31388135-31388135 on Build GRCh38
rs111309674 Chr.18: 31388353-31388353 on Build GRCh38
rs145872108 Chr.18: 31388127-31388127 on Build GRCh38
rs915736147 Chr.18: 31388304-31388304 on Build GRCh38
rs543482376 Chr.18: 31388328-31388328 on Build GRCh38
rs765398241 Chr.18: 31388520-31388521 on Build GRCh38
rs1045588877 Chr.18: 31388363-31388363 on Build GRCh38
rs767429901 Chr.18: 31388316-31388316 on Build GRCh38
rs374554073 Chr.18: 31388463-31388463 on Build GRCh38
rs536662472 Chr.18: 31388157-31388157 on Build GRCh38
rs762562609 Chr.18: 31388417-31388417 on Build GRCh38
rs548077429 Chr.18: 31388394-31388394 on Build GRCh38
rs555326265 Chr.18: 31388170-31388170 on Build GRCh38
rs746951483 Chr.18: 31388464-31388464 on Build GRCh38
rs150084716 Chr.18: 31388269-31388269 on Build GRCh38
rs368482528 Chr.18: 31388481-31388481 on Build GRCh38
rs759510780 Chr.18: 31388477-31388479 on Build GRCh38
rs751004337 Chr.18: 31388539-31388539 on Build GRCh38
rs537127889 Chr.18: 31388585-31388585 on Build GRCh38
rs769990795 Chr.18: 31388382-31388382 on Build GRCh38
rs999684617 Chr.18: 31388517-31388517 on Build GRCh38
rs201692493 Chr.18: 31388338-31388338 on Build GRCh38
rs759127869 Chr.18: 31388406-31388406 on Build GRCh38
rs36040686 Chr.18: 31388386-31388386 on Build GRCh38
rs768880705 Chr.18: 31388454-31388454 on Build GRCh38
rs764662007 Chr.18: 31388339-31388339 on Build GRCh38
rs1003812753 Chr.18: 31388130-31388130 on Build GRCh38
rs773332757 Chr.18: 31388393-31388393 on Build GRCh38
rs756419111 Chr.18: 31388435-31388435 on Build GRCh38
rs769442408 Chr.18: 31388496-31388496 on Build GRCh38
rs977383449 Chr.18: 31388242-31388242 on Build GRCh38
rs753567685 Chr.18: 31388337-31388337 on Build GRCh38
rs74615251 Chr.18: 31388143-31388143 on Build GRCh38
rs768827569 Chr.18: 31388390-31388390 on Build GRCh38
rs146500402 Chr.18: 31388398-31388398 on Build GRCh38
rs972647567 Chr.18: 31388330-31388330 on Build GRCh38
rs762540649 Chr.18: 31388534-31388534 on Build GRCh38
rs765993971 Chr.18: 31388384-31388384 on Build GRCh38
rs762320136 Chr.18: 31388569-31388569 on Build GRCh38
rs201790595 Chr.18: 31388334-31388334 on Build GRCh38
rs763569094 Chr.18: 31388430-31388430 on Build GRCh38
rs569446463 Chr.18: 31388518-31388518 on Build GRCh38
rs939846540 Chr.18: 31388308-31388308 on Build GRCh38
rs868142301 Chr.18: 31388472-31388472 on Build GRCh38
rs368463545 Chr.18: 31388407-31388407 on Build GRCh38
rs960728069 Chr.18: 31388224-31388224 on Build GRCh38
rs868569457 Chr.18: 31388437-31388437 on Build GRCh38
rs145324949 Chr.18: 31388396-31388396 on Build GRCh38
rs140661448 Chr.18: 31388203-31388203 on Build GRCh38
rs765906278 Chr.18: 31388418-31388418 on Build GRCh38
rs750739975 Chr.18: 31388433-31388433 on Build GRCh38
rs573878756 Chr.18: 31388195-31388195 on Build GRCh38
rs898206436 Chr.18: 31388400-31388400 on Build GRCh38
rs569380892 Chr.18: 31388156-31388156 on Build GRCh38
rs909752067 Chr.18: 31388286-31388286 on Build GRCh38
rs749440898 Chr.18: 31388387-31388387 on Build GRCh38
rs991846430 Chr.18: 31388288-31388288 on Build GRCh38
rs780365981 Chr.18: 31388440-31388440 on Build GRCh38
rs1049360047 Chr.18: 31388349-31388349 on Build GRCh38
rs778317241 Chr.18: 31388368-31388368 on Build GRCh38
rs577916884 Chr.18: 31388236-31388236 on Build GRCh38
rs182027794 Chr.18: 31388277-31388277 on Build GRCh38
rs773279734 Chr.18: 31388321-31388321 on Build GRCh38
rs1009449805 Chr.18: 31388443-31388443 on Build GRCh38
rs73410275 Chr.18: 31388233-31388233 on Build GRCh38
rs145443659 Chr.18: 31388371-31388371 on Build GRCh38
rs886053704 Chr.18: 31388409-31388409 on Build GRCh38
rs368838232 Chr.18: 31388344-31388344 on Build GRCh38
rs1019117710 Chr.18: 31388470-31388470 on Build GRCh38
rs200935383 Chr.18: 31388381-31388381 on Build GRCh38
rs768069326 Chr.18: 31388570-31388570 on Build GRCh38
rs551522777 Chr.18: 31388431-31388431 on Build GRCh38
rs371390432 Chr.18: 31388536-31388536 on Build GRCh38
rs545196049 Chr.18: 31388249-31388249 on Build GRCh38
rs1035286847 Chr.18: 31388522-31388522 on Build GRCh38
rs1024159333 Chr.18: 31388246-31388246 on Build GRCh38

View Full Product Details