Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31388753-31389295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1-AS1
Entrez Gene ID
101927718
Gene Name
DSG1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.625473
Gene Chromosome Location
Chr.18: 31343368-31426988 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
UniGene
Hs.407618
Gene Chromosome Location
Chr.18: 31376777-31415791 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5464282 c.434T>G p.V145G Chr.18: 31388935-31388935 on Build GRCh38
6083124 c.383G>T p.R128L Chr.18: 31388884-31388884 on Build GRCh38
3717878 c.373-2A>G p.? Chr.18: 31388872-31388872 on Build GRCh38
172483 c.473C>T p.S158L Chr.18: 31388974-31388974 on Build GRCh38
5686983 c.446A>G p.D149G Chr.18: 31388947-31388947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887997189 Chr.18: 31389223-31389223 on Build GRCh38
rs760021293 Chr.18: 31388997-31388997 on Build GRCh38
rs201177179 Chr.18: 31388975-31388975 on Build GRCh38
rs755185752 Chr.18: 31388848-31388848 on Build GRCh38
rs1056218875 Chr.18: 31389142-31389142 on Build GRCh38
rs897920456 Chr.18: 31389237-31389237 on Build GRCh38
rs748300234 Chr.18: 31388860-31388860 on Build GRCh38
rs961223012 Chr.18: 31389197-31389201 on Build GRCh38
rs775104246 Chr.18: 31388877-31388877 on Build GRCh38
rs546603536 Chr.18: 31389092-31389092 on Build GRCh38
rs750889268 Chr.18: 31388835-31388835 on Build GRCh38
rs534276605 Chr.18: 31388785-31388785 on Build GRCh38
rs967755657 Chr.18: 31389085-31389085 on Build GRCh38
rs763042725 Chr.18: 31389010-31389010 on Build GRCh38
rs368593554 Chr.18: 31388876-31388876 on Build GRCh38
rs371642087 Chr.18: 31388900-31388900 on Build GRCh38
rs773978247 Chr.18: 31388883-31388883 on Build GRCh38
rs752938690 Chr.18: 31388919-31388919 on Build GRCh38
rs374759265 Chr.18: 31389083-31389083 on Build GRCh38
rs752822527 Chr.18: 31388843-31388843 on Build GRCh38
rs16959863 Chr.18: 31389045-31389045 on Build GRCh38
rs766917303 Chr.18: 31388886-31388886 on Build GRCh38
rs768239218 Chr.18: 31388879-31388879 on Build GRCh38
rs149265994 Chr.18: 31388840-31388840 on Build GRCh38
rs138080453 Chr.18: 31388935-31388935 on Build GRCh38
rs76399598 Chr.18: 31388918-31388918 on Build GRCh38
rs745767888 Chr.18: 31388869-31388869 on Build GRCh38
rs755934013 Chr.18: 31388946-31388946 on Build GRCh38
rs764234368 Chr.18: 31389026-31389026 on Build GRCh38
rs781233097 Chr.18: 31388866-31388866 on Build GRCh38
rs749138368 Chr.18: 31388960-31388960 on Build GRCh38
rs979487376 Chr.18: 31389104-31389104 on Build GRCh38
rs567223520 Chr.18: 31389072-31389072 on Build GRCh38
rs538933300 Chr.18: 31389143-31389143 on Build GRCh38
rs557173600 Chr.18: 31388884-31388884 on Build GRCh38
rs146993377 Chr.18: 31388841-31388841 on Build GRCh38
rs769650327 Chr.18: 31388872-31388872 on Build GRCh38
rs369812803 Chr.18: 31389069-31389069 on Build GRCh38
rs200217327 Chr.18: 31388985-31388985 on Build GRCh38
rs751531908 Chr.18: 31389030-31389030 on Build GRCh38
rs961222783 Chr.18: 31388821-31388821 on Build GRCh38
rs189844610 Chr.18: 31389207-31389207 on Build GRCh38
rs541994591 Chr.18: 31389136-31389136 on Build GRCh38
rs957389630 Chr.18: 31388827-31388827 on Build GRCh38
rs9956865 Chr.18: 31388996-31388996 on Build GRCh38
rs9946162 Chr.18: 31389055-31389055 on Build GRCh38
rs758597836 Chr.18: 31388927-31388927 on Build GRCh38
rs369555342 Chr.18: 31388974-31388974 on Build GRCh38
rs971682308 Chr.18: 31389082-31389082 on Build GRCh38
rs779911399 Chr.18: 31388953-31388953 on Build GRCh38
rs1056608617 Chr.18: 31388936-31388936 on Build GRCh38
rs761259085 Chr.18: 31388988-31388988 on Build GRCh38
rs779152664 Chr.18: 31388849-31388849 on Build GRCh38
rs764376432 Chr.18: 31389054-31389054 on Build GRCh38
rs567243631 Chr.18: 31388783-31388783 on Build GRCh38
rs765676569 Chr.18: 31388998-31388998 on Build GRCh38
rs778642207 Chr.18: 31389061-31389061 on Build GRCh38
rs9946169 Chr.18: 31389081-31389081 on Build GRCh38
rs955252103 Chr.18: 31388870-31388870 on Build GRCh38
rs908375316 Chr.18: 31389179-31389179 on Build GRCh38
rs923690394 Chr.18: 31389135-31389135 on Build GRCh38
rs758659944 Chr.18: 31388883-31388897 on Build GRCh38
rs773793326 Chr.18: 31388898-31388898 on Build GRCh38
rs757345992 Chr.18: 31389032-31389032 on Build GRCh38
rs779253679 Chr.18: 31388999-31388999 on Build GRCh38
rs948184607 Chr.18: 31389164-31389164 on Build GRCh38
rs569045832 Chr.18: 31389235-31389235 on Build GRCh38
rs13381457 Chr.18: 31388961-31388961 on Build GRCh38
rs143110911 Chr.18: 31388899-31388899 on Build GRCh38
rs758550163 Chr.18: 31388863-31388863 on Build GRCh38
rs530559568 Chr.18: 31389054-31389054 on Build GRCh38
rs186165345 Chr.18: 31388902-31388902 on Build GRCh38
rs919574084 Chr.18: 31388909-31388909 on Build GRCh38
rs1033001644 Chr.18: 31388780-31388780 on Build GRCh38
rs918130161 Chr.18: 31389157-31389157 on Build GRCh38
rs1045349001 Chr.18: 31389222-31389222 on Build GRCh38
rs147207368 Chr.18: 31389053-31389053 on Build GRCh38
rs748939245 Chr.18: 31388878-31388878 on Build GRCh38
rs981422555 Chr.18: 31389122-31389122 on Build GRCh38
rs553326731 Chr.18: 31388809-31388809 on Build GRCh38
rs745845853 Chr.18: 31388942-31388942 on Build GRCh38
rs1006239509 Chr.18: 31389231-31389231 on Build GRCh38

View Full Product Details