Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31390888-31391423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1-AS1
Entrez Gene ID
101927718
Gene Name
DSG1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.625473
Gene Chromosome Location
Chr.18: 31343368-31426988 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
UniGene
Hs.407618
Gene Chromosome Location
Chr.18: 31376777-31415791 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1680313 c.769A>T p.I257F Chr.18: 31391162-31391162 on Build GRCh38
4637369 c.709G>A p.V237M Chr.18: 31391102-31391102 on Build GRCh38
5018209 c.784G>A p.V262I Chr.18: 31391177-31391177 on Build GRCh38
4394663 c.774G>A p.K258K Chr.18: 31391167-31391167 on Build GRCh38
3525108 c.724C>T p.R242W Chr.18: 31391117-31391117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779516049 Chr.18: 31391241-31391241 on Build GRCh38
rs778504245 Chr.18: 31391243-31391244 on Build GRCh38
rs367712480 Chr.18: 31391096-31391096 on Build GRCh38
rs890212247 Chr.18: 31391340-31391340 on Build GRCh38
rs867571290 Chr.18: 31391117-31391117 on Build GRCh38
rs375090363 Chr.18: 31391347-31391347 on Build GRCh38
rs376466681 Chr.18: 31391055-31391055 on Build GRCh38
rs755680547 Chr.18: 31391051-31391051 on Build GRCh38
rs956051550 Chr.18: 31390999-31390999 on Build GRCh38
rs148802079 Chr.18: 31391182-31391182 on Build GRCh38
rs1047347948 Chr.18: 31391271-31391271 on Build GRCh38
rs530042422 Chr.18: 31391377-31391377 on Build GRCh38
rs778074197 Chr.18: 31391257-31391257 on Build GRCh38
rs760788420 Chr.18: 31391093-31391093 on Build GRCh38
rs780859407 Chr.18: 31391029-31391029 on Build GRCh38
rs745425699 Chr.18: 31391038-31391038 on Build GRCh38
rs773419927 Chr.18: 31391088-31391088 on Build GRCh38
rs755733906 Chr.18: 31391120-31391120 on Build GRCh38
rs1017737867 Chr.18: 31390919-31390919 on Build GRCh38
rs540804083 Chr.18: 31391166-31391166 on Build GRCh38
rs747186527 Chr.18: 31391167-31391167 on Build GRCh38
rs75969922 Chr.18: 31391056-31391056 on Build GRCh38
rs753448411 Chr.18: 31391226-31391226 on Build GRCh38
rs893519627 Chr.18: 31391085-31391085 on Build GRCh38
rs768691954 Chr.18: 31391183-31391183 on Build GRCh38
rs754442470 Chr.18: 31391236-31391236 on Build GRCh38
rs760362395 Chr.18: 31391221-31391221 on Build GRCh38
rs267606776 Chr.18: 31391193-31391193 on Build GRCh38
rs16961784 Chr.18: 31391019-31391019 on Build GRCh38
rs769120117 Chr.18: 31391369-31391369 on Build GRCh38
rs1035877652 Chr.18: 31391152-31391152 on Build GRCh38
rs746430548 Chr.18: 31391191-31391191 on Build GRCh38
rs768923193 Chr.18: 31391058-31391058 on Build GRCh38
rs373224014 Chr.18: 31390931-31390931 on Build GRCh38
rs767397078 Chr.18: 31391204-31391204 on Build GRCh38
rs754409805 Chr.18: 31391241-31391242 on Build GRCh38
rs748579354 Chr.18: 31391151-31391151 on Build GRCh38
rs146853509 Chr.18: 31391108-31391108 on Build GRCh38
rs544221978 Chr.18: 31391291-31391291 on Build GRCh38
rs539833212 Chr.18: 31390934-31390934 on Build GRCh38
rs750068426 Chr.18: 31391118-31391118 on Build GRCh38
rs779565461 Chr.18: 31391121-31391121 on Build GRCh38
rs775315191 Chr.18: 31391098-31391098 on Build GRCh38
rs866353795 Chr.18: 31391211-31391211 on Build GRCh38
rs776703259 Chr.18: 31391177-31391177 on Build GRCh38
rs192421503 Chr.18: 31391266-31391266 on Build GRCh38
rs371706500 Chr.18: 31391298-31391298 on Build GRCh38
rs765822300 Chr.18: 31391222-31391222 on Build GRCh38
rs763798387 Chr.18: 31391100-31391100 on Build GRCh38
rs761740344 Chr.18: 31391201-31391201 on Build GRCh38
rs143657134 Chr.18: 31390994-31390994 on Build GRCh38
rs757100375 Chr.18: 31391106-31391106 on Build GRCh38
rs7228968 Chr.18: 31390970-31390970 on Build GRCh38
rs112037677 Chr.18: 31391320-31391321 on Build GRCh38
rs758789448 Chr.18: 31391244-31391244 on Build GRCh38
rs944506078 Chr.18: 31391361-31391361 on Build GRCh38
rs772398849 Chr.18: 31391155-31391155 on Build GRCh38
rs779275029 Chr.18: 31391150-31391150 on Build GRCh38
rs751308464 Chr.18: 31391104-31391104 on Build GRCh38
rs984648968 Chr.18: 31391350-31391350 on Build GRCh38
rs771033904 Chr.18: 31391176-31391176 on Build GRCh38
rs942627323 Chr.18: 31391034-31391034 on Build GRCh38
rs201222279 Chr.18: 31390968-31390970 on Build GRCh38
rs28380082 Chr.18: 31391195-31391195 on Build GRCh38
rs971430605 Chr.18: 31391078-31391078 on Build GRCh38
rs774711511 Chr.18: 31391071-31391071 on Build GRCh38
rs377332513 Chr.18: 31390996-31390996 on Build GRCh38
rs749789051 Chr.18: 31391130-31391130 on Build GRCh38
rs576994924 Chr.18: 31391269-31391269 on Build GRCh38
rs144324362 Chr.18: 31391083-31391083 on Build GRCh38
rs753183587 Chr.18: 31391243-31391243 on Build GRCh38
rs750121324 Chr.18: 31391209-31391209 on Build GRCh38
rs952827307 Chr.18: 31391312-31391312 on Build GRCh38
rs747240010 Chr.18: 31391260-31391260 on Build GRCh38
rs972892475 Chr.18: 31390922-31390922 on Build GRCh38
rs755456442 Chr.18: 31391146-31391146 on Build GRCh38
rs551516939 Chr.18: 31390943-31390943 on Build GRCh38
rs1041389942 Chr.18: 31391040-31391040 on Build GRCh38
rs756900453 Chr.18: 31391028-31391028 on Build GRCh38
rs562338814 Chr.18: 31391341-31391341 on Build GRCh38

View Full Product Details