Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32265978-32266486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM1
Entrez Gene ID
64762
Gene Name
GRB2 associated regulator of MAPK1 subtype 1
Gene Aliases
C18orf11, FAM59A, GAREM, Gm944
UniGene
Hs.444314
Gene Chromosome Location
Chr.18: 32263521-32470518 on Build GRCh38
Transcripts
NM_001242409.1 NM_022751.2

SNP Detail

SNP ID Chromosome Location
rs192768727 Chr.18: 32266114-32266114 on Build GRCh38
rs993304899 Chr.18: 32266342-32266342 on Build GRCh38
rs772989462 Chr.18: 32266422-32266422 on Build GRCh38
rs557400907 Chr.18: 32266165-32266165 on Build GRCh38
rs937758546 Chr.18: 32266320-32266320 on Build GRCh38
rs538804634 Chr.18: 32266419-32266419 on Build GRCh38
rs951380614 Chr.18: 32266100-32266100 on Build GRCh38
rs569408399 Chr.18: 32266195-32266195 on Build GRCh38
rs936808796 Chr.18: 32266163-32266163 on Build GRCh38
rs1024374563 Chr.18: 32266016-32266018 on Build GRCh38
rs898467222 Chr.18: 32266375-32266375 on Build GRCh38
rs899009596 Chr.18: 32266329-32266329 on Build GRCh38
rs995491967 Chr.18: 32266387-32266387 on Build GRCh38
rs567277370 Chr.18: 32266391-32266391 on Build GRCh38
rs368127491 Chr.18: 32266292-32266293 on Build GRCh38
rs746046073 Chr.18: 32266184-32266184 on Build GRCh38
rs188213344 Chr.18: 32266042-32266042 on Build GRCh38
rs58057202 Chr.18: 32266187-32266187 on Build GRCh38
rs1046520362 Chr.18: 32266196-32266196 on Build GRCh38
rs890407963 Chr.18: 32266372-32266372 on Build GRCh38
rs771256900 Chr.18: 32266106-32266106 on Build GRCh38
rs147584158 Chr.18: 32266386-32266387 on Build GRCh38
rs966913255 Chr.18: 32266408-32266408 on Build GRCh38
rs1004209835 Chr.18: 32266054-32266054 on Build GRCh38
rs1015685409 Chr.18: 32266352-32266352 on Build GRCh38
rs987230535 Chr.18: 32266111-32266111 on Build GRCh38
rs605522 Chr.18: 32266164-32266164 on Build GRCh38
rs915820707 Chr.18: 32266459-32266459 on Build GRCh38
rs772282812 Chr.18: 32266193-32266193 on Build GRCh38
rs977202858 Chr.18: 32266154-32266154 on Build GRCh38
rs944496641 Chr.18: 32266152-32266152 on Build GRCh38
rs988296248 Chr.18: 32266449-32266449 on Build GRCh38
rs547035800 Chr.18: 32266404-32266404 on Build GRCh38
rs986959845 Chr.18: 32266132-32266132 on Build GRCh38
rs182733960 Chr.18: 32266142-32266142 on Build GRCh38
rs148443548 Chr.18: 32266081-32266081 on Build GRCh38
rs398120288 Chr.18: 32266390-32266391 on Build GRCh38
rs142337714 Chr.18: 32266127-32266128 on Build GRCh38
rs571373519 Chr.18: 32266418-32266418 on Build GRCh38
rs760655193 Chr.18: 32266298-32266298 on Build GRCh38
rs545408251 Chr.18: 32266117-32266117 on Build GRCh38
rs962744673 Chr.18: 32266109-32266109 on Build GRCh38
rs964512395 Chr.18: 32266126-32266126 on Build GRCh38
rs774733237 Chr.18: 32266147-32266147 on Build GRCh38
rs4121653 Chr.18: 32266448-32266448 on Build GRCh38
rs142741293 Chr.18: 32266323-32266323 on Build GRCh38
rs975521331 Chr.18: 32266128-32266128 on Build GRCh38
rs73419886 Chr.18: 32266397-32266397 on Build GRCh38
rs1026134953 Chr.18: 32266361-32266361 on Build GRCh38
rs764281228 Chr.18: 32266357-32266357 on Build GRCh38
rs904854215 Chr.18: 32266250-32266250 on Build GRCh38
rs898957482 Chr.18: 32266017-32266017 on Build GRCh38
rs559715415 Chr.18: 32266108-32266108 on Build GRCh38
rs964039501 Chr.18: 32266403-32266403 on Build GRCh38
rs879324477 Chr.18: 32266117-32266117 on Build GRCh38

View Full Product Details