Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 35972613-35973055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf21
Entrez Gene ID
83608
Gene Name
chromosome 18 open reading frame 21
Gene Aliases
HsT3108, PNAS-124, PNAS-131, XTP13
UniGene
Hs.37883
Gene Chromosome Location
Chr.18: 35972625-35979287 on Build GRCh38
Transcripts
NM_001201474.1 NM_001201475.1 NM_001201476.1 NM_031446.4 XM_005258364.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5854157 c.73C>T p.L25F Chr.18: 35972801-35972801 on Build GRCh38
383925 c.96G>T p.S32S Chr.18: 35972903-35972903 on Build GRCh38
5649304 c.92C>T p.S31L Chr.18: 35972899-35972899 on Build GRCh38
6614794 c.78-3C>T p.? Chr.18: 35972882-35972882 on Build GRCh38
4427428 c.33G>T p.A11A Chr.18: 35972761-35972761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761061704 Chr.18: 35972694-35972694 on Build GRCh38
rs556345526 Chr.18: 35972725-35972725 on Build GRCh38
rs750356233 Chr.18: 35972806-35972806 on Build GRCh38
rs746845494 Chr.18: 35972721-35972721 on Build GRCh38
rs916245327 Chr.18: 35972767-35972767 on Build GRCh38
rs1010950796 Chr.18: 35972962-35972962 on Build GRCh38
rs534737677 Chr.18: 35972651-35972651 on Build GRCh38
rs374869352 Chr.18: 35972696-35972696 on Build GRCh38
rs1008605058 Chr.18: 35972886-35972886 on Build GRCh38
rs369714222 Chr.18: 35972922-35972922 on Build GRCh38
rs541952483 Chr.18: 35972774-35972774 on Build GRCh38
rs764199607 Chr.18: 35972959-35972959 on Build GRCh38
rs565747261 Chr.18: 35972641-35972641 on Build GRCh38
rs764832634 Chr.18: 35972763-35972763 on Build GRCh38
rs1032822961 Chr.18: 35972938-35972938 on Build GRCh38
rs766170311 Chr.18: 35972953-35972953 on Build GRCh38
rs61731892 Chr.18: 35972899-35972899 on Build GRCh38
rs770135416 Chr.18: 35972925-35972925 on Build GRCh38
rs773398630 Chr.18: 35972740-35972740 on Build GRCh38
rs772383831 Chr.18: 35972736-35972736 on Build GRCh38
rs761151882 Chr.18: 35972746-35972746 on Build GRCh38
rs746268515 Chr.18: 35972924-35972924 on Build GRCh38
rs748035888 Chr.18: 35972716-35972717 on Build GRCh38
rs17648305 Chr.18: 35972634-35972634 on Build GRCh38
rs928276396 Chr.18: 35972797-35972797 on Build GRCh38
rs781671504 Chr.18: 35972921-35972921 on Build GRCh38
rs758133444 Chr.18: 35972717-35972717 on Build GRCh38
rs780070613 Chr.18: 35972890-35972890 on Build GRCh38
rs773892133 Chr.18: 35972935-35972935 on Build GRCh38
rs916582790 Chr.18: 35972708-35972708 on Build GRCh38
rs771405619 Chr.18: 35972845-35972845 on Build GRCh38
rs201221269 Chr.18: 35972876-35972876 on Build GRCh38
rs748280454 Chr.18: 35972816-35972816 on Build GRCh38
rs371862193 Chr.18: 35972850-35972850 on Build GRCh38
rs781022770 Chr.18: 35972803-35972803 on Build GRCh38
rs751212214 Chr.18: 35972874-35972874 on Build GRCh38
rs371871535 Chr.18: 35972681-35972681 on Build GRCh38
rs903130871 Chr.18: 35972918-35972918 on Build GRCh38
rs199532304 Chr.18: 35972928-35972928 on Build GRCh38
rs377277789 Chr.18: 35972702-35972702 on Build GRCh38
rs754934105 Chr.18: 35972958-35972958 on Build GRCh38
rs757150727 Chr.18: 35972722-35972722 on Build GRCh38
rs760860919 Chr.18: 35972869-35972872 on Build GRCh38
rs550079045 Chr.18: 35972989-35972989 on Build GRCh38
rs545645196 Chr.18: 35972699-35972699 on Build GRCh38
rs773404225 Chr.18: 35972867-35972868 on Build GRCh38
rs373659583 Chr.18: 35972941-35972941 on Build GRCh38
rs748289475 Chr.18: 35972735-35972735 on Build GRCh38
rs753797006 Chr.18: 35972954-35972954 on Build GRCh38
rs368700045 Chr.18: 35973023-35973024 on Build GRCh38
rs755994591 Chr.18: 35972889-35972889 on Build GRCh38
rs1023706363 Chr.18: 35972963-35972963 on Build GRCh38
rs752276630 Chr.18: 35972715-35972715 on Build GRCh38
rs776081362 Chr.18: 35972927-35972927 on Build GRCh38
rs766670307 Chr.18: 35972871-35972872 on Build GRCh38
rs750270086 Chr.18: 35972884-35972884 on Build GRCh38
rs577101546 Chr.18: 35972671-35972671 on Build GRCh38
rs771315707 Chr.18: 35972907-35972907 on Build GRCh38
rs371255234 Chr.18: 35972966-35972966 on Build GRCh38
rs747315225 Chr.18: 35972842-35972842 on Build GRCh38
rs373403147 Chr.18: 35972812-35972812 on Build GRCh38
rs2298716 Chr.18: 35972635-35972635 on Build GRCh38
rs761522229 Chr.18: 35972875-35972875 on Build GRCh38
rs766926524 Chr.18: 35972747-35972747 on Build GRCh38
rs774559836 Chr.18: 35972729-35972729 on Build GRCh38
rs772949453 Chr.18: 35972950-35972950 on Build GRCh38
rs373993468 Chr.18: 35972868-35972868 on Build GRCh38
rs767771687 Chr.18: 35972688-35972688 on Build GRCh38
rs899817680 Chr.18: 35972902-35972902 on Build GRCh38
rs75299916 Chr.18: 35973023-35973023 on Build GRCh38
rs763600150 Chr.18: 35972871-35972871 on Build GRCh38
rs762153914 Chr.18: 35972682-35972682 on Build GRCh38
rs374307763 Chr.18: 35972833-35972833 on Build GRCh38
rs369956559 Chr.18: 35972866-35972866 on Build GRCh38
rs764920603 Chr.18: 35972709-35972709 on Build GRCh38
rs1015214393 Chr.18: 35972741-35972742 on Build GRCh38
rs935615106 Chr.18: 35972639-35972639 on Build GRCh38
rs759885739 Chr.18: 35972923-35972923 on Build GRCh38
rs773664741 Chr.18: 35972691-35972691 on Build GRCh38
rs989498240 Chr.18: 35972701-35972701 on Build GRCh38
rs1013484210 Chr.18: 35973013-35973015 on Build GRCh38
rs765504978 Chr.18: 35972925-35972926 on Build GRCh38
rs969324375 Chr.18: 35972724-35972724 on Build GRCh38
rs777769100 Chr.18: 35972903-35972903 on Build GRCh38
rs778060885 Chr.18: 35972834-35972834 on Build GRCh38
rs752289559 Chr.18: 35972765-35972765 on Build GRCh38
rs756087163 Chr.18: 35972808-35972808 on Build GRCh38
rs761434063 Chr.18: 35972939-35972939 on Build GRCh38
rs749309802 Chr.18: 35972726-35972726 on Build GRCh38
rs757061008 Chr.18: 35972785-35972785 on Build GRCh38
rs780863722 Chr.18: 35972741-35972741 on Build GRCh38
rs960410701 Chr.18: 35972743-35972743 on Build GRCh38
rs866122136 Chr.18: 35973031-35973031 on Build GRCh38
rs1008607733 Chr.18: 35972660-35972660 on Build GRCh38
rs553759096 Chr.18: 35972655-35972655 on Build GRCh38
rs762465890 Chr.18: 35972768-35972768 on Build GRCh38
rs768709594 Chr.18: 35972728-35972728 on Build GRCh38
rs763840961 Chr.18: 35972773-35972773 on Build GRCh38
rs999150836 Chr.18: 35972926-35972926 on Build GRCh38
rs17562811 Chr.18: 35972858-35972858 on Build GRCh38
rs776999445 Chr.18: 35972918-35972919 on Build GRCh38
rs371003240 Chr.18: 35973012-35973013 on Build GRCh38
rs760174508 Chr.18: 35972751-35972751 on Build GRCh38
rs770358598 Chr.18: 35972854-35972854 on Build GRCh38
rs184896173 Chr.18: 35972934-35972934 on Build GRCh38
rs774826193 Chr.18: 35972855-35972855 on Build GRCh38
rs373045082 Chr.18: 35972974-35972974 on Build GRCh38
rs763431664 Chr.18: 35972933-35972933 on Build GRCh38
rs555296789 Chr.18: 35972640-35972640 on Build GRCh38
rs369854960 Chr.18: 35972716-35972716 on Build GRCh38
rs570235857 Chr.18: 35973018-35973018 on Build GRCh38
rs750844462 Chr.18: 35973017-35973017 on Build GRCh38
rs199497571 Chr.18: 35972766-35972766 on Build GRCh38
rs774262241 Chr.18: 35972713-35972713 on Build GRCh38
rs747226938 Chr.18: 35972904-35972904 on Build GRCh38

View Full Product Details