Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 35978645-35979145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf21
Entrez Gene ID
83608
Gene Name
chromosome 18 open reading frame 21
Gene Aliases
HsT3108, PNAS-124, PNAS-131, XTP13
UniGene
Hs.37883
Gene Chromosome Location
Chr.18: 35972625-35979287 on Build GRCh38
Transcripts
NM_001201474.1 NM_001201475.1 NM_001201476.1 NM_031446.4 XM_005258364.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
988182 c.603G>T p.K201N Chr.18: 35978946-35978946 on Build GRCh38
4752433 c.524G>A p.C175Y Chr.18: 35978867-35978867 on Build GRCh38
988183 c.629T>A p.F210Y Chr.18: 35978972-35978972 on Build GRCh38
3422154 c.626A>C p.N209T Chr.18: 35978969-35978969 on Build GRCh38
5464295 c.628T>G p.F210V Chr.18: 35978971-35978971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759058188 Chr.18: 35979012-35979012 on Build GRCh38
rs368478405 Chr.18: 35978758-35978758 on Build GRCh38
rs767364644 Chr.18: 35978799-35978799 on Build GRCh38
rs80004835 Chr.18: 35978772-35978772 on Build GRCh38
rs749449588 Chr.18: 35978885-35978885 on Build GRCh38
rs903976688 Chr.18: 35978721-35978721 on Build GRCh38
rs535443844 Chr.18: 35978792-35978792 on Build GRCh38
rs377292504 Chr.18: 35978993-35978993 on Build GRCh38
rs544591480 Chr.18: 35978738-35978738 on Build GRCh38
rs753532990 Chr.18: 35978798-35978798 on Build GRCh38
rs1036049054 Chr.18: 35978682-35978682 on Build GRCh38
rs184260582 Chr.18: 35978793-35978793 on Build GRCh38
rs762246199 Chr.18: 35978933-35978933 on Build GRCh38
rs753478453 Chr.18: 35978973-35978973 on Build GRCh38
rs547627462 Chr.18: 35978689-35978689 on Build GRCh38
rs774544109 Chr.18: 35978920-35978920 on Build GRCh38
rs1031006763 Chr.18: 35978781-35978781 on Build GRCh38
rs545276810 Chr.18: 35978812-35978812 on Build GRCh38
rs765953729 Chr.18: 35978989-35978989 on Build GRCh38
rs757553965 Chr.18: 35979065-35979065 on Build GRCh38
rs772403832 Chr.18: 35978949-35978949 on Build GRCh38
rs982291041 Chr.18: 35978938-35978938 on Build GRCh38
rs751469319 Chr.18: 35979041-35979041 on Build GRCh38
rs780149019 Chr.18: 35978871-35978871 on Build GRCh38
rs747996473 Chr.18: 35978807-35978807 on Build GRCh38
rs367835365 Chr.18: 35978808-35978808 on Build GRCh38
rs951919346 Chr.18: 35979011-35979011 on Build GRCh38
rs569844013 Chr.18: 35978764-35978764 on Build GRCh38
rs773845572 Chr.18: 35978953-35978953 on Build GRCh38
rs553820165 Chr.18: 35979023-35979023 on Build GRCh38
rs777613342 Chr.18: 35979029-35979029 on Build GRCh38
rs193092568 Chr.18: 35979007-35979007 on Build GRCh38
rs533402182 Chr.18: 35978745-35978745 on Build GRCh38
rs767718928 Chr.18: 35979078-35979078 on Build GRCh38
rs762264289 Chr.18: 35978879-35978879 on Build GRCh38
rs1050811803 Chr.18: 35978716-35978716 on Build GRCh38
rs186869265 Chr.18: 35978692-35978692 on Build GRCh38
rs769862746 Chr.18: 35978859-35978859 on Build GRCh38
rs761329617 Chr.18: 35978956-35978956 on Build GRCh38
rs867103202 Chr.18: 35979032-35979032 on Build GRCh38
rs983185361 Chr.18: 35979022-35979022 on Build GRCh38
rs372712333 Chr.18: 35978886-35978886 on Build GRCh38
rs757234722 Chr.18: 35979049-35979049 on Build GRCh38
rs920350333 Chr.18: 35979091-35979091 on Build GRCh38
rs1018844108 Chr.18: 35978867-35978867 on Build GRCh38
rs758223610 Chr.18: 35979028-35979028 on Build GRCh38
rs1040153996 Chr.18: 35978733-35978733 on Build GRCh38
rs191535806 Chr.18: 35978762-35978762 on Build GRCh38
rs1008713301 Chr.18: 35978724-35978724 on Build GRCh38
rs928095344 Chr.18: 35978996-35978996 on Build GRCh38
rs774166745 Chr.18: 35978723-35978723 on Build GRCh38
rs930327950 Chr.18: 35979094-35979094 on Build GRCh38
rs764973461 Chr.18: 35979018-35979018 on Build GRCh38
rs930586536 Chr.18: 35978693-35978693 on Build GRCh38
rs189214391 Chr.18: 35978842-35978842 on Build GRCh38

View Full Product Details