Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 59433112-59433609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCBE1
Entrez Gene ID
147372
Gene Name
collagen and calcium binding EGF domains 1
Gene Aliases
HKLLS1
UniGene
Hs.34333
Gene Chromosome Location
Chr.18: 59430939-59698181 on Build GRCh38
Transcripts
NM_133459.3

SNP Detail

SNP ID Chromosome Location
rs115205666 Chr.18: 59433235-59433235 on Build GRCh38
rs958678471 Chr.18: 59433414-59433414 on Build GRCh38
rs1000862997 Chr.18: 59433215-59433215 on Build GRCh38
rs886054051 Chr.18: 59433269-59433269 on Build GRCh38
rs1006579898 Chr.18: 59433319-59433319 on Build GRCh38
rs1040966582 Chr.18: 59433331-59433331 on Build GRCh38
rs910837879 Chr.18: 59433154-59433154 on Build GRCh38
rs943497467 Chr.18: 59433166-59433166 on Build GRCh38
rs931039483 Chr.18: 59433269-59433269 on Build GRCh38
rs961088104 Chr.18: 59433311-59433311 on Build GRCh38
rs1048627511 Chr.18: 59433291-59433291 on Build GRCh38
rs9959752 Chr.18: 59433547-59433547 on Build GRCh38
rs1010117755 Chr.18: 59433544-59433544 on Build GRCh38
rs1048122096 Chr.18: 59433504-59433504 on Build GRCh38
rs911000731 Chr.18: 59433519-59433519 on Build GRCh38
rs1034383126 Chr.18: 59433581-59433581 on Build GRCh38
rs528130337 Chr.18: 59433568-59433568 on Build GRCh38
rs968963430 Chr.18: 59433369-59433369 on Build GRCh38
rs1000458412 Chr.18: 59433570-59433570 on Build GRCh38
rs758679541 Chr.18: 59433162-59433162 on Build GRCh38
rs188777368 Chr.18: 59433400-59433400 on Build GRCh38
rs992816977 Chr.18: 59433420-59433420 on Build GRCh38
rs566233610 Chr.18: 59433148-59433148 on Build GRCh38
rs1036565587 Chr.18: 59433217-59433217 on Build GRCh38
rs150355335 Chr.18: 59433180-59433180 on Build GRCh38
rs559638765 Chr.18: 59433316-59433316 on Build GRCh38
rs530647353 Chr.18: 59433535-59433535 on Build GRCh38
rs76216795 Chr.18: 59433448-59433448 on Build GRCh38
rs917491093 Chr.18: 59433444-59433444 on Build GRCh38
rs903848161 Chr.18: 59433207-59433207 on Build GRCh38
rs996099606 Chr.18: 59433401-59433401 on Build GRCh38
rs886054050 Chr.18: 59433137-59433137 on Build GRCh38
rs750720598 Chr.18: 59433160-59433160 on Build GRCh38
rs901120885 Chr.18: 59433347-59433347 on Build GRCh38
rs1000254736 Chr.18: 59433373-59433373 on Build GRCh38
rs1020158328 Chr.18: 59433366-59433366 on Build GRCh38
rs113143061 Chr.18: 59433167-59433167 on Build GRCh38
rs764839664 Chr.18: 59433460-59433460 on Build GRCh38
rs866382710 Chr.18: 59433459-59433459 on Build GRCh38
rs576137278 Chr.18: 59433170-59433170 on Build GRCh38
rs895071629 Chr.18: 59433236-59433236 on Build GRCh38
rs942332331 Chr.18: 59433531-59433531 on Build GRCh38
rs5825337 Chr.18: 59433270-59433270 on Build GRCh38
rs1010478494 Chr.18: 59433350-59433350 on Build GRCh38
rs866586391 Chr.18: 59433363-59433363 on Build GRCh38
rs1024827600 Chr.18: 59433289-59433289 on Build GRCh38
rs760587700 Chr.18: 59433530-59433530 on Build GRCh38
rs370758874 Chr.18: 59433506-59433507 on Build GRCh38
rs1042451845 Chr.18: 59433507-59433507 on Build GRCh38
rs930821759 Chr.18: 59433497-59433497 on Build GRCh38
rs534077467 Chr.18: 59433504-59433505 on Build GRCh38
rs9959080 Chr.18: 59433536-59433536 on Build GRCh38
rs563442191 Chr.18: 59433534-59433534 on Build GRCh38
rs1042481682 Chr.18: 59433200-59433200 on Build GRCh38
rs972457789 Chr.18: 59433330-59433330 on Build GRCh38
rs1975499 Chr.18: 59433178-59433178 on Build GRCh38
rs937353232 Chr.18: 59433176-59433176 on Build GRCh38
rs574005370 Chr.18: 59433223-59433223 on Build GRCh38
rs917522099 Chr.18: 59433179-59433179 on Build GRCh38
rs1013798937 Chr.18: 59433282-59433282 on Build GRCh38
rs114107592 Chr.18: 59433150-59433150 on Build GRCh38

View Full Product Details