Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63329966-63330507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
UniGene
Hs.74050
Gene Chromosome Location
Chr.18: 63327736-63367273 on Build GRCh38
Transcripts
NM_002035.2 XM_005266677.2 XM_017025690.1

SNP Detail

SNP ID Chromosome Location
rs112172911 Chr.18: 63330316-63330316 on Build GRCh38
rs753426065 Chr.18: 63330294-63330294 on Build GRCh38
rs998421102 Chr.18: 63330217-63330218 on Build GRCh38
rs186879790 Chr.18: 63330282-63330282 on Build GRCh38
rs965253616 Chr.18: 63330202-63330202 on Build GRCh38
rs937535345 Chr.18: 63330298-63330298 on Build GRCh38
rs904278628 Chr.18: 63330406-63330406 on Build GRCh38
rs1037211031 Chr.18: 63330096-63330096 on Build GRCh38
rs2849383 Chr.18: 63330148-63330148 on Build GRCh38
rs940035196 Chr.18: 63330082-63330082 on Build GRCh38
rs561479821 Chr.18: 63330120-63330121 on Build GRCh38
rs1006765193 Chr.18: 63330185-63330185 on Build GRCh38
rs984191024 Chr.18: 63330264-63330264 on Build GRCh38
rs868821722 Chr.18: 63330026-63330026 on Build GRCh38
rs780666682 Chr.18: 63330412-63330412 on Build GRCh38
rs1031556369 Chr.18: 63330205-63330205 on Build GRCh38
rs1036469063 Chr.18: 63330073-63330073 on Build GRCh38
rs1056011790 Chr.18: 63329991-63329991 on Build GRCh38
rs904160799 Chr.18: 63330191-63330191 on Build GRCh38
rs928221839 Chr.18: 63330113-63330113 on Build GRCh38
rs972747301 Chr.18: 63330293-63330293 on Build GRCh38
rs978482668 Chr.18: 63330265-63330265 on Build GRCh38
rs901070071 Chr.18: 63330439-63330439 on Build GRCh38
rs897804121 Chr.18: 63330121-63330121 on Build GRCh38
rs552549977 Chr.18: 63330126-63330126 on Build GRCh38
rs888404112 Chr.18: 63330179-63330179 on Build GRCh38
rs767237100 Chr.18: 63330000-63330000 on Build GRCh38
rs917399889 Chr.18: 63330317-63330317 on Build GRCh38
rs991379175 Chr.18: 63330315-63330315 on Build GRCh38
rs1042796198 Chr.18: 63330402-63330402 on Build GRCh38
rs909861287 Chr.18: 63330069-63330069 on Build GRCh38
rs1036844443 Chr.18: 63330392-63330392 on Build GRCh38
rs181772990 Chr.18: 63330257-63330257 on Build GRCh38
rs760267541 Chr.18: 63330195-63330195 on Build GRCh38
rs1052806228 Chr.18: 63330427-63330427 on Build GRCh38
rs984508169 Chr.18: 63330051-63330052 on Build GRCh38
rs2850765 Chr.18: 63330216-63330216 on Build GRCh38
rs756714765 Chr.18: 63330324-63330324 on Build GRCh38
rs1016979900 Chr.18: 63330269-63330269 on Build GRCh38
rs1020055589 Chr.18: 63330228-63330228 on Build GRCh38
rs1012692636 Chr.18: 63330173-63330173 on Build GRCh38
rs887691320 Chr.18: 63330299-63330299 on Build GRCh38
rs946888598 Chr.18: 63330352-63330352 on Build GRCh38
rs950150877 Chr.18: 63330023-63330023 on Build GRCh38
rs991755385 Chr.18: 63330309-63330309 on Build GRCh38

View Full Product Details