Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63330704-63331247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
UniGene
Hs.74050
Gene Chromosome Location
Chr.18: 63327736-63367273 on Build GRCh38
Transcripts
NM_002035.2 XM_005266677.2 XM_017025690.1

SNP Detail

SNP ID Chromosome Location
rs1042046124 Chr.18: 63331191-63331191 on Build GRCh38
rs189836495 Chr.18: 63331119-63331119 on Build GRCh38
rs79558815 Chr.18: 63331077-63331077 on Build GRCh38
rs530235237 Chr.18: 63330728-63330728 on Build GRCh38
rs150690456 Chr.18: 63331123-63331123 on Build GRCh38
rs919405205 Chr.18: 63331093-63331093 on Build GRCh38
rs900807227 Chr.18: 63330798-63330801 on Build GRCh38
rs1032787877 Chr.18: 63331002-63331002 on Build GRCh38
rs553164019 Chr.18: 63330931-63330931 on Build GRCh38
rs968741450 Chr.18: 63330962-63330962 on Build GRCh38
rs142034651 Chr.18: 63330773-63330773 on Build GRCh38
rs540694008 Chr.18: 63330758-63330758 on Build GRCh38
rs910528436 Chr.18: 63330822-63330822 on Build GRCh38
rs201719061 Chr.18: 63330799-63330800 on Build GRCh38
rs911126797 Chr.18: 63331103-63331103 on Build GRCh38
rs759355464 Chr.18: 63331064-63331064 on Build GRCh38
rs999924808 Chr.18: 63330965-63330965 on Build GRCh38
rs1035081125 Chr.18: 63330926-63330926 on Build GRCh38
rs778805326 Chr.18: 63330839-63330839 on Build GRCh38
rs923266299 Chr.18: 63331182-63331182 on Build GRCh38
rs533642615 Chr.18: 63331157-63331157 on Build GRCh38
rs925686104 Chr.18: 63330782-63330782 on Build GRCh38
rs746977417 Chr.18: 63331020-63331020 on Build GRCh38
rs1049625170 Chr.18: 63330802-63330802 on Build GRCh38
rs924081399 Chr.18: 63330967-63330967 on Build GRCh38
rs377432947 Chr.18: 63330843-63330843 on Build GRCh38
rs547798387 Chr.18: 63331112-63331112 on Build GRCh38
rs1005333804 Chr.18: 63330824-63330825 on Build GRCh38
rs1024087291 Chr.18: 63331194-63331194 on Build GRCh38
rs941304559 Chr.18: 63331110-63331110 on Build GRCh38
rs943396870 Chr.18: 63330899-63330899 on Build GRCh38
rs777761633 Chr.18: 63330940-63330940 on Build GRCh38
rs1038194357 Chr.18: 63330888-63330888 on Build GRCh38
rs933656647 Chr.18: 63330800-63330800 on Build GRCh38
rs370756076 Chr.18: 63331117-63331117 on Build GRCh38
rs919423465 Chr.18: 63330730-63330730 on Build GRCh38
rs902999945 Chr.18: 63331220-63331220 on Build GRCh38
rs774806589 Chr.18: 63330810-63330810 on Build GRCh38
rs532645370 Chr.18: 63330840-63330840 on Build GRCh38
rs879227252 Chr.18: 63330809-63330810 on Build GRCh38
rs181309339 Chr.18: 63330839-63330839 on Build GRCh38
rs901121139 Chr.18: 63330929-63330929 on Build GRCh38
rs775308361 Chr.18: 63331096-63331096 on Build GRCh38
rs774352195 Chr.18: 63331029-63331029 on Build GRCh38
rs1040390383 Chr.18: 63331170-63331170 on Build GRCh38
rs185131595 Chr.18: 63330935-63330935 on Build GRCh38
rs1039582479 Chr.18: 63330916-63330916 on Build GRCh38
rs552720173 Chr.18: 63330912-63330912 on Build GRCh38
rs574752083 Chr.18: 63331010-63331010 on Build GRCh38
rs6810 Chr.18: 63331168-63331168 on Build GRCh38
rs903022189 Chr.18: 63330964-63330964 on Build GRCh38
rs536417444 Chr.18: 63330869-63330869 on Build GRCh38
rs1041511173 Chr.18: 63330959-63330959 on Build GRCh38
rs910664821 Chr.18: 63331167-63331167 on Build GRCh38
rs200054441 Chr.18: 63330809-63330809 on Build GRCh38
rs984821659 Chr.18: 63331118-63331118 on Build GRCh38
rs74881875 Chr.18: 63330825-63330825 on Build GRCh38
rs2849367 Chr.18: 63330784-63330784 on Build GRCh38
rs1056513339 Chr.18: 63331130-63331130 on Build GRCh38
rs958349570 Chr.18: 63331006-63331006 on Build GRCh38
rs968272935 Chr.18: 63331195-63331195 on Build GRCh38
rs570519022 Chr.18: 63331189-63331189 on Build GRCh38
rs754710969 Chr.18: 63330783-63330783 on Build GRCh38
rs1049146184 Chr.18: 63330792-63330792 on Build GRCh38

View Full Product Details