Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74256752-74257264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5A
Entrez Gene ID
1528
Gene Name
cytochrome b5 type A
Gene Aliases
CYB5, MCB5
UniGene
Hs.465413
Gene Chromosome Location
Chr.18: 74253292-74292016 on Build GRCh38
Transcripts
NM_001190807.2 NM_001914.3 NM_148923.3 XM_011525835.2

SNP Detail

SNP ID Chromosome Location
rs901931228 Chr.18: 74257105-74257105 on Build GRCh38
rs1050258194 Chr.18: 74257138-74257138 on Build GRCh38
rs1010827548 Chr.18: 74256945-74256945 on Build GRCh38
rs940754558 Chr.18: 74257096-74257096 on Build GRCh38
rs986135180 Chr.18: 74256990-74256990 on Build GRCh38
rs376205710 Chr.18: 74256868-74256868 on Build GRCh38
rs892316795 Chr.18: 74256901-74256901 on Build GRCh38
rs181246796 Chr.18: 74257071-74257071 on Build GRCh38
rs933113275 Chr.18: 74257137-74257137 on Build GRCh38
rs370904381 Chr.18: 74256936-74256936 on Build GRCh38
rs748511232 Chr.18: 74256823-74256823 on Build GRCh38
rs1028830583 Chr.18: 74256841-74256841 on Build GRCh38
rs774139236 Chr.18: 74256879-74256879 on Build GRCh38
rs916794964 Chr.18: 74257018-74257018 on Build GRCh38
rs948326054 Chr.18: 74257027-74257027 on Build GRCh38
rs779600610 Chr.18: 74256822-74256822 on Build GRCh38
rs573634481 Chr.18: 74257172-74257172 on Build GRCh38
rs746935745 Chr.18: 74257173-74257173 on Build GRCh38
rs548078743 Chr.18: 74256974-74256974 on Build GRCh38
rs540059631 Chr.18: 74257167-74257167 on Build GRCh38
rs916166293 Chr.18: 74257143-74257143 on Build GRCh38
rs1044721796 Chr.18: 74257155-74257155 on Build GRCh38
rs1011477034 Chr.18: 74257160-74257160 on Build GRCh38
rs559996169 Chr.18: 74257170-74257170 on Build GRCh38
rs1043627625 Chr.18: 74257201-74257201 on Build GRCh38
rs1018933697 Chr.18: 74257227-74257227 on Build GRCh38
rs947697806 Chr.18: 74257145-74257145 on Build GRCh38
rs1010755420 Chr.18: 74257188-74257188 on Build GRCh38
rs992335599 Chr.18: 74257012-74257012 on Build GRCh38
rs972277272 Chr.18: 74256785-74256785 on Build GRCh38
rs112662392 Chr.18: 74256978-74256978 on Build GRCh38
rs371569832 Chr.18: 74256861-74256861 on Build GRCh38
rs1036418361 Chr.18: 74257100-74257100 on Build GRCh38
rs376476347 Chr.18: 74256865-74256865 on Build GRCh38
rs984972857 Chr.18: 74257130-74257130 on Build GRCh38
rs534142634 Chr.18: 74256961-74256961 on Build GRCh38
rs765726746 Chr.18: 74256857-74256858 on Build GRCh38
rs202053517 Chr.18: 74256791-74256791 on Build GRCh38
rs3819168 Chr.18: 74257202-74257202 on Build GRCh38
rs2848931 Chr.18: 74256827-74256827 on Build GRCh38
rs761684030 Chr.18: 74256893-74256893 on Build GRCh38
rs576455808 Chr.18: 74257052-74257052 on Build GRCh38
rs1011194361 Chr.18: 74256859-74256859 on Build GRCh38
rs927646535 Chr.18: 74257169-74257169 on Build GRCh38
rs770027459 Chr.18: 74256860-74256860 on Build GRCh38
rs767469713 Chr.18: 74256904-74256904 on Build GRCh38
rs200375466 Chr.18: 74256840-74256840 on Build GRCh38
rs997315210 Chr.18: 74256831-74256831 on Build GRCh38
rs556634420 Chr.18: 74257036-74257036 on Build GRCh38
rs371803785 Chr.18: 74256954-74256954 on Build GRCh38
rs536631362 Chr.18: 74257007-74257007 on Build GRCh38
rs748178717 Chr.18: 74257073-74257073 on Build GRCh38
rs755124385 Chr.18: 74256933-74256933 on Build GRCh38
rs560247436 Chr.18: 74257220-74257220 on Build GRCh38
rs150657475 Chr.18: 74256845-74256845 on Build GRCh38
rs189941855 Chr.18: 74256839-74256839 on Build GRCh38
rs8091957 Chr.18: 74256817-74256817 on Build GRCh38
rs367955790 Chr.18: 74256858-74256858 on Build GRCh38
rs1003756103 Chr.18: 74256853-74256853 on Build GRCh38
rs972587914 Chr.18: 74257044-74257044 on Build GRCh38
rs966168138 Chr.18: 74257034-74257034 on Build GRCh38
rs766644443 Chr.18: 74256804-74256804 on Build GRCh38
rs577733857 Chr.18: 74257144-74257144 on Build GRCh38
rs138565253 Chr.18: 74257088-74257088 on Build GRCh38
rs562430514 Chr.18: 74256832-74256832 on Build GRCh38
rs377153930 Chr.18: 74256825-74256825 on Build GRCh38

View Full Product Details