Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42830686-42831192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMAP
Entrez Gene ID
114625
Gene Name
erythroblast membrane associated protein (Scianna blood group)
Gene Aliases
BTN5, PRO2801, RD, SC
UniGene
Hs.439437
Gene Chromosome Location
Chr.1: 42817105-42844991 on Build GRCh38
Transcripts
NM_001017922.1 NM_018538.3 XM_006710313.3 XM_011540570.2 XM_011540571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3805272 c.381G>C p.L127L Chr.1: 42831063-42831063 on Build GRCh38
6440198 c.160C>T p.L54F Chr.1: 42830842-42830842 on Build GRCh38
7323532 c.415G>A p.V139M Chr.1: 42831097-42831097 on Build GRCh38
7366577 c.103G>A p.G35S Chr.1: 42830785-42830785 on Build GRCh38
6451160 c.345C>T p.D115D Chr.1: 42831027-42831027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758090366 Chr.1: 42830918-42830918 on Build GRCh38
rs759288632 Chr.1: 42831150-42831150 on Build GRCh38
rs370783701 Chr.1: 42830894-42830894 on Build GRCh38
rs776692427 Chr.1: 42830932-42830932 on Build GRCh38
rs556539598 Chr.1: 42830855-42830855 on Build GRCh38
rs139264039 Chr.1: 42831035-42831035 on Build GRCh38
rs1024300229 Chr.1: 42830809-42830809 on Build GRCh38
rs771020983 Chr.1: 42830881-42830881 on Build GRCh38
rs146247630 Chr.1: 42830900-42830900 on Build GRCh38
rs1010517765 Chr.1: 42830764-42830764 on Build GRCh38
rs746860461 Chr.1: 42830970-42830970 on Build GRCh38
rs33954154 Chr.1: 42830901-42830901 on Build GRCh38
rs761839957 Chr.1: 42831002-42831002 on Build GRCh38
rs758771948 Chr.1: 42830850-42830850 on Build GRCh38
rs778474168 Chr.1: 42830927-42830927 on Build GRCh38
rs981372230 Chr.1: 42830775-42830775 on Build GRCh38
rs753298061 Chr.1: 42830766-42830766 on Build GRCh38
rs375116495 Chr.1: 42830773-42830773 on Build GRCh38
rs892956744 Chr.1: 42830752-42830752 on Build GRCh38
rs943692367 Chr.1: 42830921-42830921 on Build GRCh38
rs770534773 Chr.1: 42831133-42831133 on Build GRCh38
rs781594970 Chr.1: 42830784-42830784 on Build GRCh38
rs199680522 Chr.1: 42830906-42830906 on Build GRCh38
rs774480445 Chr.1: 42830882-42830882 on Build GRCh38
rs910840430 Chr.1: 42831037-42831037 on Build GRCh38
rs779076532 Chr.1: 42830957-42830957 on Build GRCh38
rs773441122 Chr.1: 42830797-42830797 on Build GRCh38
rs565545614 Chr.1: 42830730-42830730 on Build GRCh38
rs149787850 Chr.1: 42830899-42830899 on Build GRCh38
rs756534595 Chr.1: 42830926-42830926 on Build GRCh38
rs769047388 Chr.1: 42830778-42830778 on Build GRCh38
rs201419224 Chr.1: 42830866-42830866 on Build GRCh38
rs763062647 Chr.1: 42830802-42830802 on Build GRCh38
rs200923604 Chr.1: 42831094-42831094 on Build GRCh38
rs768412880 Chr.1: 42830975-42830975 on Build GRCh38
rs773347684 Chr.1: 42831097-42831097 on Build GRCh38
rs909319532 Chr.1: 42830813-42830813 on Build GRCh38
rs757800023 Chr.1: 42831048-42831048 on Build GRCh38
rs200071273 Chr.1: 42831109-42831109 on Build GRCh38
rs778168880 Chr.1: 42830912-42830912 on Build GRCh38
rs756587594 Chr.1: 42831036-42831036 on Build GRCh38
rs369767907 Chr.1: 42831161-42831161 on Build GRCh38
rs945797784 Chr.1: 42831123-42831123 on Build GRCh38
rs942340358 Chr.1: 42831039-42831039 on Build GRCh38
rs56047316 Chr.1: 42830821-42830821 on Build GRCh38
rs972224952 Chr.1: 42831027-42831027 on Build GRCh38
rs755662659 Chr.1: 42830735-42830735 on Build GRCh38
rs865897030 Chr.1: 42830818-42830818 on Build GRCh38
rs899696208 Chr.1: 42830749-42830749 on Build GRCh38
rs747837466 Chr.1: 42831105-42831105 on Build GRCh38
rs760631140 Chr.1: 42830838-42830838 on Build GRCh38
rs764081661 Chr.1: 42830902-42830902 on Build GRCh38
rs201143776 Chr.1: 42830933-42830933 on Build GRCh38
rs1014002008 Chr.1: 42831059-42831059 on Build GRCh38
rs775682230 Chr.1: 42830969-42830969 on Build GRCh38
rs1023914732 Chr.1: 42831081-42831081 on Build GRCh38
rs150664549 Chr.1: 42830923-42830923 on Build GRCh38
rs183863963 Chr.1: 42831032-42831032 on Build GRCh38
rs375125882 Chr.1: 42830864-42830864 on Build GRCh38
rs772341251 Chr.1: 42830958-42830958 on Build GRCh38
rs749786187 Chr.1: 42830929-42830929 on Build GRCh38
rs764589228 Chr.1: 42831043-42831043 on Build GRCh38
rs563298024 Chr.1: 42831028-42831028 on Build GRCh38
rs1034252057 Chr.1: 42830954-42830954 on Build GRCh38
rs750066994 Chr.1: 42830856-42830856 on Build GRCh38
rs763959022 Chr.1: 42830844-42830844 on Build GRCh38
rs746767041 Chr.1: 42830855-42830856 on Build GRCh38
rs773962354 Chr.1: 42831145-42831145 on Build GRCh38
rs186792314 Chr.1: 42831165-42831165 on Build GRCh38
rs749437755 Chr.1: 42830878-42830878 on Build GRCh38
rs200355434 Chr.1: 42831031-42831031 on Build GRCh38
rs769626904 Chr.1: 42831043-42831044 on Build GRCh38
rs140203431 Chr.1: 42830976-42830976 on Build GRCh38
rs773011605 Chr.1: 42831120-42831120 on Build GRCh38
rs756915487 Chr.1: 42830772-42830772 on Build GRCh38
rs1031029067 Chr.1: 42830786-42830786 on Build GRCh38
rs200998492 Chr.1: 42830895-42830895 on Build GRCh38
rs528557088 Chr.1: 42830731-42830731 on Build GRCh38
rs754107057 Chr.1: 42831046-42831046 on Build GRCh38
rs376310520 Chr.1: 42830796-42830796 on Build GRCh38
rs764772807 Chr.1: 42830914-42830914 on Build GRCh38
rs767315853 Chr.1: 42830827-42830827 on Build GRCh38
rs368064875 Chr.1: 42830924-42830924 on Build GRCh38
rs889506281 Chr.1: 42830711-42830711 on Build GRCh38
rs773307424 Chr.1: 42831062-42831063 on Build GRCh38
rs150122881 Chr.1: 42830820-42830820 on Build GRCh38
rs772790363 Chr.1: 42831022-42831022 on Build GRCh38
rs772256111 Chr.1: 42831055-42831055 on Build GRCh38
rs373770988 Chr.1: 42830963-42830963 on Build GRCh38
rs201520271 Chr.1: 42831014-42831014 on Build GRCh38
rs752451851 Chr.1: 42830831-42830831 on Build GRCh38
rs780299336 Chr.1: 42831056-42831056 on Build GRCh38
rs762574815 Chr.1: 42831126-42831126 on Build GRCh38
rs200211952 Chr.1: 42830867-42830867 on Build GRCh38
rs748036779 Chr.1: 42831037-42831039 on Build GRCh38
rs56136737 Chr.1: 42830860-42830860 on Build GRCh38
rs528295732 Chr.1: 42831096-42831096 on Build GRCh38
rs146429994 Chr.1: 42830785-42830785 on Build GRCh38
rs749859392 Chr.1: 42830917-42830917 on Build GRCh38
rs138466210 Chr.1: 42830708-42830708 on Build GRCh38
rs1054125822 Chr.1: 42831060-42831060 on Build GRCh38
rs777297202 Chr.1: 42830758-42830758 on Build GRCh38
rs56025238 Chr.1: 42830851-42830851 on Build GRCh38
rs767223296 Chr.1: 42831155-42831155 on Build GRCh38
rs387906265 Chr.1: 42830989-42830990 on Build GRCh38
rs768637323 Chr.1: 42830923-42830924 on Build GRCh38
rs778421271 Chr.1: 42830869-42830869 on Build GRCh38
rs375207937 Chr.1: 42830980-42830980 on Build GRCh38
rs771128741 Chr.1: 42830816-42830816 on Build GRCh38
rs779223927 Chr.1: 42831053-42831053 on Build GRCh38

View Full Product Details