Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42838766-42839234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMAP
Entrez Gene ID
114625
Gene Name
erythroblast membrane associated protein (Scianna blood group)
Gene Aliases
BTN5, PRO2801, RD, SC
UniGene
Hs.439437
Gene Chromosome Location
Chr.1: 42817105-42844991 on Build GRCh38
Transcripts
NM_001017922.1 NM_018538.3 XM_006710313.3 XM_011540570.2 XM_011540571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4958139 c.632C>T p.A211V Chr.1: 42838916-42838916 on Build GRCh38
1736404 c.630A>C p.K210N Chr.1: 42838914-42838914 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770209755 Chr.1: 42838942-42838942 on Build GRCh38
rs890619004 Chr.1: 42838793-42838793 on Build GRCh38
rs762803517 Chr.1: 42838932-42838932 on Build GRCh38
rs1052754319 Chr.1: 42838832-42838832 on Build GRCh38
rs935726666 Chr.1: 42839127-42839127 on Build GRCh38
rs746278370 Chr.1: 42839021-42839021 on Build GRCh38
rs561736272 Chr.1: 42838842-42838842 on Build GRCh38
rs752166038 Chr.1: 42839139-42839139 on Build GRCh38
rs552304100 Chr.1: 42839039-42839039 on Build GRCh38
rs34228099 Chr.1: 42838954-42838954 on Build GRCh38
rs1039246680 Chr.1: 42838837-42838837 on Build GRCh38
rs754832984 Chr.1: 42838948-42838948 on Build GRCh38
rs777401931 Chr.1: 42839019-42839019 on Build GRCh38
rs754688072 Chr.1: 42838918-42838921 on Build GRCh38
rs750247958 Chr.1: 42838909-42838909 on Build GRCh38
rs530683768 Chr.1: 42838890-42838890 on Build GRCh38
rs778273956 Chr.1: 42839084-42839084 on Build GRCh38
rs912872397 Chr.1: 42839170-42839170 on Build GRCh38
rs911965420 Chr.1: 42839201-42839201 on Build GRCh38
rs746181079 Chr.1: 42838854-42838854 on Build GRCh38
rs188661428 Chr.1: 42839152-42839152 on Build GRCh38
rs779264242 Chr.1: 42838851-42838851 on Build GRCh38
rs902880341 Chr.1: 42838918-42838918 on Build GRCh38
rs375700723 Chr.1: 42839114-42839114 on Build GRCh38
rs950001005 Chr.1: 42839178-42839178 on Build GRCh38
rs34303058 Chr.1: 42838970-42838970 on Build GRCh38
rs943571308 Chr.1: 42838797-42838797 on Build GRCh38
rs751309805 Chr.1: 42838885-42838888 on Build GRCh38
rs954173356 Chr.1: 42839063-42839063 on Build GRCh38
rs929323767 Chr.1: 42839109-42839109 on Build GRCh38
rs752945726 Chr.1: 42838792-42838792 on Build GRCh38
rs374469176 Chr.1: 42838979-42838979 on Build GRCh38
rs367645800 Chr.1: 42838856-42838856 on Build GRCh38
rs998549193 Chr.1: 42838931-42838931 on Build GRCh38
rs765493600 Chr.1: 42838906-42838906 on Build GRCh38
rs779749834 Chr.1: 42838879-42838880 on Build GRCh38
rs755771222 Chr.1: 42839014-42839014 on Build GRCh38
rs760719907 Chr.1: 42838875-42838875 on Build GRCh38
rs985264588 Chr.1: 42839081-42839081 on Build GRCh38
rs766284214 Chr.1: 42838935-42838935 on Build GRCh38
rs905744718 Chr.1: 42839208-42839208 on Build GRCh38
rs1005973190 Chr.1: 42838989-42838989 on Build GRCh38
rs776810787 Chr.1: 42838885-42838885 on Build GRCh38
rs961640008 Chr.1: 42839023-42839024 on Build GRCh38
rs369458235 Chr.1: 42838882-42838882 on Build GRCh38
rs772463784 Chr.1: 42839022-42839022 on Build GRCh38
rs976971514 Chr.1: 42839028-42839028 on Build GRCh38
rs757850169 Chr.1: 42839156-42839156 on Build GRCh38
rs1022849693 Chr.1: 42838998-42838998 on Build GRCh38
rs776054655 Chr.1: 42838863-42838863 on Build GRCh38

View Full Product Details