Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43150670-43151120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM183A
Entrez Gene ID
440585
Gene Name
family with sequence similarity 183 member A
Gene Aliases
-
UniGene
Hs.657614
Gene Chromosome Location
Chr.1: 43142959-43156396 on Build GRCh38
Transcripts
NM_001101376.2 XM_005270875.4 XM_005270876.4 XM_017001322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260781 c.161C>A p.S54Y Chr.1: 43150788-43150788 on Build GRCh38
464641 c.189A>C p.A63A Chr.1: 43150816-43150816 on Build GRCh38
6050837 c.192T>C p.D64D Chr.1: 43150819-43150819 on Build GRCh38
260782 c.161C>A p.S54Y Chr.1: 43150788-43150788 on Build GRCh38
86386 c.156C>G p.P52P Chr.1: 43150783-43150783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377327826 Chr.1: 43150815-43150815 on Build GRCh38
rs1019556896 Chr.1: 43150702-43150702 on Build GRCh38
rs769148078 Chr.1: 43150802-43150802 on Build GRCh38
rs535423049 Chr.1: 43151036-43151036 on Build GRCh38
rs560083705 Chr.1: 43150978-43150978 on Build GRCh38
rs184714550 Chr.1: 43151083-43151083 on Build GRCh38
rs763828494 Chr.1: 43150721-43150721 on Build GRCh38
rs636969 Chr.1: 43150755-43150755 on Build GRCh38
rs780730298 Chr.1: 43150797-43150797 on Build GRCh38
rs610200 Chr.1: 43151082-43151082 on Build GRCh38
rs550718609 Chr.1: 43150942-43150942 on Build GRCh38
rs759979437 Chr.1: 43150715-43150715 on Build GRCh38
rs374667425 Chr.1: 43150804-43150804 on Build GRCh38
rs761200975 Chr.1: 43150726-43150726 on Build GRCh38
rs779756811 Chr.1: 43150789-43150789 on Build GRCh38
rs116718141 Chr.1: 43151046-43151046 on Build GRCh38
rs201473714 Chr.1: 43151007-43151007 on Build GRCh38
rs965631045 Chr.1: 43150895-43150895 on Build GRCh38
rs796683459 Chr.1: 43151052-43151052 on Build GRCh38
rs769466380 Chr.1: 43150711-43150711 on Build GRCh38
rs975143459 Chr.1: 43150709-43150709 on Build GRCh38
rs79846647 Chr.1: 43150798-43150798 on Build GRCh38
rs975674586 Chr.1: 43150916-43150916 on Build GRCh38
rs994833166 Chr.1: 43150824-43150824 on Build GRCh38
rs934491605 Chr.1: 43151001-43151001 on Build GRCh38
rs1057017586 Chr.1: 43151006-43151007 on Build GRCh38
rs1037875892 Chr.1: 43151059-43151059 on Build GRCh38
rs886678022 Chr.1: 43150922-43150922 on Build GRCh38
rs1028025504 Chr.1: 43150983-43150983 on Build GRCh38
rs528466646 Chr.1: 43150808-43150808 on Build GRCh38
rs369016239 Chr.1: 43150700-43150700 on Build GRCh38
rs779224793 Chr.1: 43150757-43150757 on Build GRCh38
rs768566781 Chr.1: 43150794-43150794 on Build GRCh38
rs765170895 Chr.1: 43150837-43150837 on Build GRCh38
rs753552458 Chr.1: 43150749-43150750 on Build GRCh38
rs78945945 Chr.1: 43151026-43151026 on Build GRCh38
rs1019766552 Chr.1: 43151084-43151084 on Build GRCh38
rs375660558 Chr.1: 43150793-43150793 on Build GRCh38
rs759147262 Chr.1: 43150854-43150854 on Build GRCh38
rs965073512 Chr.1: 43150703-43150703 on Build GRCh38
rs774619898 Chr.1: 43150714-43150714 on Build GRCh38
rs753273720 Chr.1: 43150725-43150725 on Build GRCh38
rs1057087540 Chr.1: 43150719-43150719 on Build GRCh38
rs373578024 Chr.1: 43150827-43150827 on Build GRCh38
rs532860301 Chr.1: 43150986-43150986 on Build GRCh38
rs917048459 Chr.1: 43151054-43151054 on Build GRCh38
rs968473216 Chr.1: 43150998-43150998 on Build GRCh38
rs531110764 Chr.1: 43150839-43150839 on Build GRCh38
rs758225215 Chr.1: 43150784-43150784 on Build GRCh38
rs76650845 Chr.1: 43151025-43151025 on Build GRCh38
rs189605677 Chr.1: 43150993-43150993 on Build GRCh38
rs368847619 Chr.1: 43150817-43150817 on Build GRCh38
rs41269531 Chr.1: 43151021-43151021 on Build GRCh38
rs758544218 Chr.1: 43150844-43150844 on Build GRCh38
rs964853989 Chr.1: 43150976-43150976 on Build GRCh38
rs753942666 Chr.1: 43150749-43150749 on Build GRCh38
rs147138163 Chr.1: 43151020-43151020 on Build GRCh38
rs541897355 Chr.1: 43150814-43150814 on Build GRCh38
rs16830421 Chr.1: 43150975-43150975 on Build GRCh38
rs574795885 Chr.1: 43150767-43150767 on Build GRCh38
rs372936834 Chr.1: 43150707-43150707 on Build GRCh38
rs924542957 Chr.1: 43150750-43150750 on Build GRCh38
rs898835194 Chr.1: 43151035-43151035 on Build GRCh38
rs577373724 Chr.1: 43150744-43150744 on Build GRCh38
rs1009583780 Chr.1: 43150959-43150959 on Build GRCh38
rs75150344 Chr.1: 43150721-43150721 on Build GRCh38
rs921305305 Chr.1: 43150951-43150951 on Build GRCh38
rs772942637 Chr.1: 43150803-43150803 on Build GRCh38

View Full Product Details