Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 581918-582437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSG
Entrez Gene ID
682
Gene Name
basigin (Ok blood group)
Gene Aliases
5F7, CD147, EMMPRIN, OK, TCSF
UniGene
Hs.501293
Gene Chromosome Location
Chr.19: 571277-583493 on Build GRCh38
Transcripts
NM_001322243.1 NM_001728.3 NM_198589.2 NM_198590.2 NM_198591.2 XM_017027173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3540522 c.1070-1G>A p.? Chr.19: 582305-582305 on Build GRCh38
1680445 c.1076A>G p.D359G Chr.19: 582312-582312 on Build GRCh38
1680446 c.536A>G p.D179G Chr.19: 582312-582312 on Build GRCh38
5589844 c.1092C>T p.P364P Chr.19: 582328-582328 on Build GRCh38
3540523 c.530-1G>A p.? Chr.19: 582305-582305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753155859 Chr.19: 582276-582276 on Build GRCh38
rs767724482 Chr.19: 582368-582368 on Build GRCh38
rs146784114 Chr.19: 582314-582314 on Build GRCh38
rs537512067 Chr.19: 582140-582140 on Build GRCh38
rs543483161 Chr.19: 582339-582339 on Build GRCh38
rs935256511 Chr.19: 582153-582153 on Build GRCh38
rs1054255143 Chr.19: 582254-582254 on Build GRCh38
rs1026754394 Chr.19: 582172-582172 on Build GRCh38
rs937479249 Chr.19: 581945-581945 on Build GRCh38
rs56067110 Chr.19: 582006-582006 on Build GRCh38
rs781097295 Chr.19: 582296-582296 on Build GRCh38
rs374164143 Chr.19: 582369-582369 on Build GRCh38
rs369414573 Chr.19: 582357-582357 on Build GRCh38
rs757345514 Chr.19: 582293-582293 on Build GRCh38
rs774585852 Chr.19: 582297-582299 on Build GRCh38
rs542552123 Chr.19: 582245-582245 on Build GRCh38
rs1027138204 Chr.19: 582324-582324 on Build GRCh38
rs527846123 Chr.19: 581969-581969 on Build GRCh38
rs772819177 Chr.19: 582272-582272 on Build GRCh38
rs779194178 Chr.19: 582057-582057 on Build GRCh38
rs957304161 Chr.19: 582005-582005 on Build GRCh38
rs981083858 Chr.19: 582022-582022 on Build GRCh38
rs780058910 Chr.19: 582380-582380 on Build GRCh38
rs776558092 Chr.19: 582352-582370 on Build GRCh38
rs28992487 Chr.19: 582092-582092 on Build GRCh38
rs202135216 Chr.19: 582292-582292 on Build GRCh38
rs903127527 Chr.19: 582126-582126 on Build GRCh38
rs564651283 Chr.19: 581989-581989 on Build GRCh38
rs779976854 Chr.19: 582313-582313 on Build GRCh38
rs531691645 Chr.19: 582001-582001 on Build GRCh38
rs529207268 Chr.19: 582363-582363 on Build GRCh38
rs28992485 Chr.19: 581974-581974 on Build GRCh38
rs369856442 Chr.19: 582268-582268 on Build GRCh38
rs991163862 Chr.19: 582166-582166 on Build GRCh38
rs765810022 Chr.19: 582349-582349 on Build GRCh38
rs772609315 Chr.19: 582271-582273 on Build GRCh38
rs756216375 Chr.19: 582370-582370 on Build GRCh38
rs200086920 Chr.19: 582290-582290 on Build GRCh38
rs568077701 Chr.19: 582013-582013 on Build GRCh38
rs958436387 Chr.19: 581953-581953 on Build GRCh38
rs995995043 Chr.19: 582000-582000 on Build GRCh38
rs572647656 Chr.19: 582275-582275 on Build GRCh38
rs372918193 Chr.19: 582285-582285 on Build GRCh38
rs760173898 Chr.19: 582345-582345 on Build GRCh38
rs773186872 Chr.19: 582227-582227 on Build GRCh38
rs899947991 Chr.19: 581994-581994 on Build GRCh38
rs574442476 Chr.19: 582163-582163 on Build GRCh38
rs952456664 Chr.19: 582081-582081 on Build GRCh38
rs887069304 Chr.19: 581968-581968 on Build GRCh38
rs1041091347 Chr.19: 582074-582074 on Build GRCh38
rs375929078 Chr.19: 582265-582265 on Build GRCh38
rs921232937 Chr.19: 582236-582236 on Build GRCh38
rs931595588 Chr.19: 581957-581957 on Build GRCh38
rs1047049137 Chr.19: 581961-581961 on Build GRCh38
rs558467385 Chr.19: 582114-582114 on Build GRCh38
rs1055916755 Chr.19: 581947-581947 on Build GRCh38
rs536000280 Chr.19: 582145-582145 on Build GRCh38
rs1023777597 Chr.19: 582045-582045 on Build GRCh38
rs943911305 Chr.19: 582093-582093 on Build GRCh38
rs554598015 Chr.19: 582234-582234 on Build GRCh38
rs1049556694 Chr.19: 581984-581984 on Build GRCh38
rs958471897 Chr.19: 582027-582027 on Build GRCh38
rs908808241 Chr.19: 582070-582070 on Build GRCh38
rs553637912 Chr.19: 582175-582175 on Build GRCh38
rs547374902 Chr.19: 582365-582365 on Build GRCh38
rs538375679 Chr.19: 582353-582353 on Build GRCh38
rs532703227 Chr.19: 582071-582071 on Build GRCh38
rs761617952 Chr.19: 582263-582263 on Build GRCh38
rs764506893 Chr.19: 582289-582289 on Build GRCh38
rs901934467 Chr.19: 582004-582004 on Build GRCh38
rs772760933 Chr.19: 582341-582341 on Build GRCh38
rs989523686 Chr.19: 582408-582408 on Build GRCh38
rs28992483 Chr.19: 581944-581944 on Build GRCh38
rs1015114957 Chr.19: 582232-582232 on Build GRCh38
rs1053882201 Chr.19: 582082-582082 on Build GRCh38
rs770770103 Chr.19: 582308-582310 on Build GRCh38
rs576495399 Chr.19: 582329-582329 on Build GRCh38
rs556218809 Chr.19: 582154-582154 on Build GRCh38
rs200237260 Chr.19: 582352-582352 on Build GRCh38
rs191110653 Chr.19: 582137-582137 on Build GRCh38
rs28992484 Chr.19: 581956-581956 on Build GRCh38
rs1035357491 Chr.19: 582160-582160 on Build GRCh38
rs768559835 Chr.19: 582317-582317 on Build GRCh38
rs920121525 Chr.19: 582056-582056 on Build GRCh38
rs908768792 Chr.19: 582205-582205 on Build GRCh38
rs923786354 Chr.19: 582299-582299 on Build GRCh38
rs774143658 Chr.19: 582262-582262 on Build GRCh38
rs928372831 Chr.19: 582024-582024 on Build GRCh38
rs944312447 Chr.19: 582233-582233 on Build GRCh38
rs889618506 Chr.19: 581993-581993 on Build GRCh38
rs894105830 Chr.19: 582316-582316 on Build GRCh38
rs1000016233 Chr.19: 581946-581946 on Build GRCh38
rs753545807 Chr.19: 582273-582273 on Build GRCh38
rs28992486 Chr.19: 582077-582077 on Build GRCh38
rs970932596 Chr.19: 581966-581966 on Build GRCh38
rs748132503 Chr.19: 582115-582115 on Build GRCh38
rs931069639 Chr.19: 581982-581982 on Build GRCh38
rs571614464 Chr.19: 582049-582049 on Build GRCh38
rs983000267 Chr.19: 582178-582178 on Build GRCh38
rs932759632 Chr.19: 582080-582080 on Build GRCh38
rs972876619 Chr.19: 581973-581973 on Build GRCh38
rs529959881 Chr.19: 582016-582016 on Build GRCh38
rs4919862 Chr.19: 582253-582253 on Build GRCh38
rs774338462 Chr.19: 582326-582326 on Build GRCh38
rs1047739105 Chr.19: 582068-582068 on Build GRCh38
rs112152563 Chr.19: 582310-582310 on Build GRCh38
rs1003743321 Chr.19: 582226-582226 on Build GRCh38

View Full Product Details