Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1277042-1277478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf24
Entrez Gene ID
55009
Gene Name
chromosome 19 open reading frame 24
Gene Aliases
-
UniGene
Hs.591383
Gene Chromosome Location
Chr.19: 1275521-1279244 on Build GRCh38
Transcripts
NM_017914.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438617 c.375G>C p.V125V Chr.19: 1277276-1277276 on Build GRCh38
7269759 c.386G>A p.R129Q Chr.19: 1277287-1277287 on Build GRCh38
3764698 c.363C>T p.D121D Chr.19: 1277264-1277264 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001539687 Chr.19: 1277183-1277183 on Build GRCh38
rs757881896 Chr.19: 1277140-1277140 on Build GRCh38
rs896272846 Chr.19: 1277266-1277267 on Build GRCh38
rs954284705 Chr.19: 1277086-1277086 on Build GRCh38
rs150667555 Chr.19: 1277382-1277382 on Build GRCh38
rs553277632 Chr.19: 1277252-1277252 on Build GRCh38
rs777728540 Chr.19: 1277182-1277182 on Build GRCh38
rs757292 Chr.19: 1277276-1277276 on Build GRCh38
rs1004196885 Chr.19: 1277205-1277205 on Build GRCh38
rs565357941 Chr.19: 1277123-1277123 on Build GRCh38
rs183439478 Chr.19: 1277369-1277369 on Build GRCh38
rs1004653609 Chr.19: 1277449-1277449 on Build GRCh38
rs566677899 Chr.19: 1277261-1277261 on Build GRCh38
rs780887692 Chr.19: 1277168-1277168 on Build GRCh38
rs112664514 Chr.19: 1277264-1277264 on Build GRCh38
rs927717695 Chr.19: 1277153-1277153 on Build GRCh38
rs910738562 Chr.19: 1277089-1277089 on Build GRCh38
rs531820089 Chr.19: 1277340-1277340 on Build GRCh38
rs779504906 Chr.19: 1277145-1277145 on Build GRCh38
rs1048944295 Chr.19: 1277198-1277198 on Build GRCh38
rs976942308 Chr.19: 1277084-1277084 on Build GRCh38
rs984956917 Chr.19: 1277087-1277087 on Build GRCh38
rs569686123 Chr.19: 1277206-1277206 on Build GRCh38
rs1017747717 Chr.19: 1277335-1277335 on Build GRCh38
rs200278776 Chr.19: 1277298-1277298 on Build GRCh38
rs770951244 Chr.19: 1277189-1277189 on Build GRCh38
rs934669063 Chr.19: 1277156-1277156 on Build GRCh38
rs80150968 Chr.19: 1277120-1277120 on Build GRCh38
rs952676169 Chr.19: 1277064-1277064 on Build GRCh38
rs769499794 Chr.19: 1277179-1277179 on Build GRCh38
rs564467817 Chr.19: 1277286-1277286 on Build GRCh38
rs767743867 Chr.19: 1277170-1277170 on Build GRCh38
rs529418775 Chr.19: 1277414-1277414 on Build GRCh38
rs555528240 Chr.19: 1277219-1277219 on Build GRCh38
rs138739305 Chr.19: 1277210-1277210 on Build GRCh38
rs561922263 Chr.19: 1277391-1277391 on Build GRCh38
rs993159420 Chr.19: 1277275-1277275 on Build GRCh38
rs983291757 Chr.19: 1277067-1277067 on Build GRCh38
rs1030371583 Chr.19: 1277379-1277379 on Build GRCh38
rs965248360 Chr.19: 1277338-1277338 on Build GRCh38
rs755831014 Chr.19: 1277331-1277331 on Build GRCh38
rs551471438 Chr.19: 1277236-1277236 on Build GRCh38
rs777089956 Chr.19: 1277251-1277251 on Build GRCh38
rs751062801 Chr.19: 1277272-1277272 on Build GRCh38
rs539011382 Chr.19: 1277157-1277157 on Build GRCh38
rs774563106 Chr.19: 1277194-1277194 on Build GRCh38
rs145899521 Chr.19: 1277085-1277085 on Build GRCh38
rs551682863 Chr.19: 1277240-1277240 on Build GRCh38
rs913800949 Chr.19: 1277177-1277177 on Build GRCh38
rs1015664082 Chr.19: 1277069-1277069 on Build GRCh38
rs939221091 Chr.19: 1277161-1277161 on Build GRCh38
rs1028498053 Chr.19: 1277232-1277232 on Build GRCh38
rs72979542 Chr.19: 1277071-1277071 on Build GRCh38
rs557576284 Chr.19: 1277199-1277199 on Build GRCh38
rs765790295 Chr.19: 1277265-1277265 on Build GRCh38
rs989692746 Chr.19: 1277104-1277104 on Build GRCh38
rs1048988827 Chr.19: 1277426-1277426 on Build GRCh38
rs751118010 Chr.19: 1277147-1277147 on Build GRCh38
rs749169747 Chr.19: 1277341-1277341 on Build GRCh38
rs1006896715 Chr.19: 1277334-1277334 on Build GRCh38
rs752325836 Chr.19: 1277302-1277302 on Build GRCh38
rs750522767 Chr.19: 1277211-1277211 on Build GRCh38
rs757293 Chr.19: 1277428-1277428 on Build GRCh38
rs574018765 Chr.19: 1277220-1277220 on Build GRCh38
rs771242963 Chr.19: 1277139-1277140 on Build GRCh38
rs752503228 Chr.19: 1277311-1277311 on Build GRCh38
rs964796581 Chr.19: 1277098-1277098 on Build GRCh38

View Full Product Details