Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1425800-1426310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAZAP1
Entrez Gene ID
26528
Gene Name
DAZ associated protein 1
Gene Aliases
-
UniGene
Hs.222510
Gene Chromosome Location
Chr.19: 1407569-1435687 on Build GRCh38
Transcripts
NM_018959.3 NM_170711.2 XM_005259531.3 XM_005259534.3 XM_005259535.3 XM_005259536.3 XM_011527904.2 XM_011527905.2 XM_011527906.2 XM_011527907.2 XM_011527908.2 XM_011527909.2 XM_011527910.2 XM_017026581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640138 c.546+2T>C p.? Chr.19: 1425962-1425962 on Build GRCh38
6640127 c.529G>A p.D177N Chr.19: 1425943-1425943 on Build GRCh38
6640126 c.529G>A p.D177N Chr.19: 1425943-1425943 on Build GRCh38
7127244 c.513C>A p.V171V Chr.19: 1425927-1425927 on Build GRCh38
6346615 c.464-6G>T p.? Chr.19: 1425872-1425872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115556085 Chr.19: 1426169-1426169 on Build GRCh38
rs552249754 Chr.19: 1426170-1426170 on Build GRCh38
rs781295069 Chr.19: 1426259-1426259 on Build GRCh38
rs531321403 Chr.19: 1426196-1426196 on Build GRCh38
rs925981997 Chr.19: 1426099-1426099 on Build GRCh38
rs987718576 Chr.19: 1426250-1426250 on Build GRCh38
rs569256794 Chr.19: 1426244-1426244 on Build GRCh38
rs956497677 Chr.19: 1426190-1426190 on Build GRCh38
rs575231515 Chr.19: 1426088-1426088 on Build GRCh38
rs755221846 Chr.19: 1425829-1425829 on Build GRCh38
rs537859363 Chr.19: 1426235-1426235 on Build GRCh38
rs540332721 Chr.19: 1426040-1426040 on Build GRCh38
rs754739773 Chr.19: 1426257-1426257 on Build GRCh38
rs1000778181 Chr.19: 1426156-1426156 on Build GRCh38
rs946150793 Chr.19: 1426081-1426081 on Build GRCh38
rs930144632 Chr.19: 1426048-1426048 on Build GRCh38
rs1020496496 Chr.19: 1426267-1426267 on Build GRCh38
rs544311974 Chr.19: 1426061-1426061 on Build GRCh38
rs547142760 Chr.19: 1426271-1426271 on Build GRCh38
rs764782717 Chr.19: 1426058-1426058 on Build GRCh38
rs189439378 Chr.19: 1426151-1426151 on Build GRCh38
rs770268589 Chr.19: 1425849-1425849 on Build GRCh38
rs564066893 Chr.19: 1426182-1426182 on Build GRCh38
rs996217831 Chr.19: 1426218-1426219 on Build GRCh38
rs748471404 Chr.19: 1425846-1425846 on Build GRCh38
rs781482303 Chr.19: 1425844-1425844 on Build GRCh38
rs577156666 Chr.19: 1426046-1426046 on Build GRCh38
rs779731182 Chr.19: 1426201-1426201 on Build GRCh38
rs962947414 Chr.19: 1426270-1426270 on Build GRCh38
rs370042829 Chr.19: 1426012-1426012 on Build GRCh38
rs372564307 Chr.19: 1426253-1426254 on Build GRCh38
rs535370361 Chr.19: 1426243-1426243 on Build GRCh38
rs890889195 Chr.19: 1426087-1426087 on Build GRCh38
rs937300335 Chr.19: 1426102-1426102 on Build GRCh38
rs903764913 Chr.19: 1426152-1426152 on Build GRCh38
rs1038543664 Chr.19: 1426015-1426015 on Build GRCh38
rs1050828859 Chr.19: 1426070-1426070 on Build GRCh38
rs1028646221 Chr.19: 1426243-1426244 on Build GRCh38
rs1046256214 Chr.19: 1426150-1426150 on Build GRCh38
rs563173405 Chr.19: 1426068-1426068 on Build GRCh38
rs949317555 Chr.19: 1426141-1426141 on Build GRCh38
rs549540976 Chr.19: 1426212-1426212 on Build GRCh38
rs941616925 Chr.19: 1425836-1425836 on Build GRCh38
rs1015930513 Chr.19: 1426189-1426190 on Build GRCh38
rs745368158 Chr.19: 1425986-1425986 on Build GRCh38
rs558831610 Chr.19: 1426044-1426044 on Build GRCh38
rs971914677 Chr.19: 1426204-1426204 on Build GRCh38
rs200309834 Chr.19: 1425869-1425869 on Build GRCh38
rs1055068290 Chr.19: 1426104-1426104 on Build GRCh38
rs117428133 Chr.19: 1426112-1426112 on Build GRCh38

View Full Product Details