Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1457849-1458345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC2
Entrez Gene ID
10297
Gene Name
APC2, WNT signaling pathway regulator
Gene Aliases
APCL
UniGene
Hs.446376
Gene Chromosome Location
Chr.19: 1446268-1473244 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296739 c.1211C>T p.P404L Chr.19: 1457968-1457968 on Build GRCh38
6371451 c.1283G>A p.R428H Chr.19: 1458040-1458040 on Build GRCh38
6270354 c.1247G>T p.C416F Chr.19: 1458004-1458004 on Build GRCh38
5368376 c.1301T>A p.L434Q Chr.19: 1458058-1458058 on Build GRCh38
6384914 c.1261C>A p.L421M Chr.19: 1458018-1458018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992414319 Chr.19: 1458201-1458201 on Build GRCh38
rs909311662 Chr.19: 1458239-1458239 on Build GRCh38
rs140141232 Chr.19: 1458289-1458289 on Build GRCh38
rs374392097 Chr.19: 1457904-1457904 on Build GRCh38
rs188279084 Chr.19: 1457986-1457986 on Build GRCh38
rs150835778 Chr.19: 1457983-1457983 on Build GRCh38
rs375692529 Chr.19: 1457922-1457922 on Build GRCh38
rs374455383 Chr.19: 1457940-1457940 on Build GRCh38
rs1053796983 Chr.19: 1458188-1458188 on Build GRCh38
rs1029945125 Chr.19: 1458112-1458112 on Build GRCh38
rs754970632 Chr.19: 1458005-1458005 on Build GRCh38
rs576515497 Chr.19: 1458103-1458103 on Build GRCh38
rs146114138 Chr.19: 1458043-1458043 on Build GRCh38
rs73921709 Chr.19: 1458126-1458126 on Build GRCh38
rs143064262 Chr.19: 1458039-1458039 on Build GRCh38
rs946348456 Chr.19: 1457919-1457919 on Build GRCh38
rs141413463 Chr.19: 1457968-1457968 on Build GRCh38
rs1009737272 Chr.19: 1457960-1457960 on Build GRCh38
rs374792492 Chr.19: 1458056-1458056 on Build GRCh38
rs12005 Chr.19: 1457876-1457876 on Build GRCh38
rs767995316 Chr.19: 1458077-1458077 on Build GRCh38
rs751535159 Chr.19: 1457984-1457984 on Build GRCh38
rs370654661 Chr.19: 1458027-1458027 on Build GRCh38
rs562185980 Chr.19: 1458124-1458124 on Build GRCh38
rs756489747 Chr.19: 1457973-1457973 on Build GRCh38
rs151160268 Chr.19: 1458177-1458177 on Build GRCh38
rs757809712 Chr.19: 1458092-1458092 on Build GRCh38
rs117105828 Chr.19: 1458162-1458162 on Build GRCh38
rs181292388 Chr.19: 1458227-1458227 on Build GRCh38
rs760056307 Chr.19: 1457926-1457926 on Build GRCh38
rs868280811 Chr.19: 1458046-1458046 on Build GRCh38
rs868027511 Chr.19: 1458042-1458042 on Build GRCh38
rs778278037 Chr.19: 1458040-1458040 on Build GRCh38
rs202079646 Chr.19: 1457893-1457894 on Build GRCh38
rs768496655 Chr.19: 1458294-1458294 on Build GRCh38
rs138304573 Chr.19: 1458013-1458013 on Build GRCh38
rs559375014 Chr.19: 1458146-1458146 on Build GRCh38
rs112680150 Chr.19: 1457898-1457898 on Build GRCh38
rs868200980 Chr.19: 1457896-1457896 on Build GRCh38
rs974955543 Chr.19: 1458171-1458171 on Build GRCh38
rs754467372 Chr.19: 1458076-1458076 on Build GRCh38
rs781447236 Chr.19: 1457949-1457956 on Build GRCh38
rs1054170018 Chr.19: 1457913-1457913 on Build GRCh38
rs967761601 Chr.19: 1458158-1458158 on Build GRCh38
rs565711332 Chr.19: 1458260-1458260 on Build GRCh38
rs537253505 Chr.19: 1457889-1457889 on Build GRCh38
rs776265391 Chr.19: 1458102-1458102 on Build GRCh38
rs750782191 Chr.19: 1458090-1458091 on Build GRCh38
rs887801417 Chr.19: 1458006-1458006 on Build GRCh38
rs557991091 Chr.19: 1458280-1458280 on Build GRCh38
rs1033459450 Chr.19: 1458086-1458086 on Build GRCh38
rs950359429 Chr.19: 1458143-1458143 on Build GRCh38
rs942084716 Chr.19: 1458259-1458259 on Build GRCh38
rs71174372 Chr.19: 1457898-1457899 on Build GRCh38
rs776405778 Chr.19: 1458311-1458311 on Build GRCh38
rs753368242 Chr.19: 1457937-1457937 on Build GRCh38
rs72983582 Chr.19: 1458271-1458271 on Build GRCh38
rs530752200 Chr.19: 1458187-1458187 on Build GRCh38
rs915282520 Chr.19: 1458206-1458206 on Build GRCh38
rs913600195 Chr.19: 1457906-1457906 on Build GRCh38
rs761470970 Chr.19: 1457941-1457941 on Build GRCh38
rs549964908 Chr.19: 1457905-1457905 on Build GRCh38
rs866008886 Chr.19: 1458032-1458032 on Build GRCh38
rs867067028 Chr.19: 1458054-1458054 on Build GRCh38
rs911797896 Chr.19: 1458149-1458149 on Build GRCh38
rs547498958 Chr.19: 1458236-1458236 on Build GRCh38
rs140139305 Chr.19: 1458053-1458053 on Build GRCh38
rs368556143 Chr.19: 1457997-1457997 on Build GRCh38
rs568530097 Chr.19: 1457981-1457981 on Build GRCh38
rs987883589 Chr.19: 1457901-1457901 on Build GRCh38
rs1039446337 Chr.19: 1458265-1458265 on Build GRCh38
rs774681407 Chr.19: 1458065-1458065 on Build GRCh38
rs781334434 Chr.19: 1458012-1458012 on Build GRCh38
rs779686435 Chr.19: 1458228-1458228 on Build GRCh38
rs765006967 Chr.19: 1457952-1457952 on Build GRCh38
rs113990038 Chr.19: 1457895-1457895 on Build GRCh38
rs558054376 Chr.19: 1458081-1458081 on Build GRCh38
rs899754120 Chr.19: 1458087-1458087 on Build GRCh38
rs867374414 Chr.19: 1458315-1458315 on Build GRCh38
rs142105979 Chr.19: 1458125-1458125 on Build GRCh38
rs867828116 Chr.19: 1457994-1457994 on Build GRCh38
rs758297254 Chr.19: 1457969-1457969 on Build GRCh38
rs756509712 Chr.19: 1458097-1458098 on Build GRCh38
rs374378343 Chr.19: 1457895-1457896 on Build GRCh38
rs979517366 Chr.19: 1458169-1458169 on Build GRCh38
rs115085389 Chr.19: 1458290-1458290 on Build GRCh38
rs376651213 Chr.19: 1458304-1458304 on Build GRCh38
rs371111262 Chr.19: 1457928-1457928 on Build GRCh38
rs2656876 Chr.19: 1458181-1458181 on Build GRCh38
rs929427408 Chr.19: 1457993-1457993 on Build GRCh38
rs10402084 Chr.19: 1458178-1458178 on Build GRCh38
rs544005584 Chr.19: 1458122-1458122 on Build GRCh38
rs868519009 Chr.19: 1458031-1458031 on Build GRCh38
rs12974445 Chr.19: 1458212-1458212 on Build GRCh38
rs750288797 Chr.19: 1457956-1457956 on Build GRCh38
rs2656875 Chr.19: 1458113-1458113 on Build GRCh38
rs955181183 Chr.19: 1458121-1458121 on Build GRCh38
rs576210142 Chr.19: 1457885-1457885 on Build GRCh38
rs549155915 Chr.19: 1458200-1458200 on Build GRCh38

View Full Product Details