Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2108596-2109099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3D1
Entrez Gene ID
8943
Gene Name
adaptor related protein complex 3 delta 1 subunit
Gene Aliases
ADTD, hBLVR
UniGene
Hs.512815
Gene Chromosome Location
Chr.19: 2100988-2151620 on Build GRCh38
Transcripts
NM_001261826.1 NM_003938.6 XM_006722932.2 XM_017027422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283443 c.3358delG p.V1120fs*1 Chr.19: 2108695-2108695 on Build GRCh38
3378669 c.3286+1G>A p.? Chr.19: 2109085-2109085 on Build GRCh38
3378670 c.3163+1G>A p.? Chr.19: 2109085-2109085 on Build GRCh38
7153884 c.3158T>C p.F1053S Chr.19: 2109091-2109091 on Build GRCh38
5958122 c.3490T>C p.S1164P Chr.19: 2108749-2108749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995610582 Chr.19: 2108823-2108823 on Build GRCh38
rs772733833 Chr.19: 2108676-2108676 on Build GRCh38
rs898227406 Chr.19: 2108797-2108797 on Build GRCh38
rs373771967 Chr.19: 2109038-2109038 on Build GRCh38
rs555478575 Chr.19: 2108665-2108665 on Build GRCh38
rs776751558 Chr.19: 2109068-2109068 on Build GRCh38
rs375487362 Chr.19: 2108744-2108744 on Build GRCh38
rs371867820 Chr.19: 2108775-2108775 on Build GRCh38
rs745974282 Chr.19: 2108760-2108760 on Build GRCh38
rs953880721 Chr.19: 2108890-2108890 on Build GRCh38
rs137960645 Chr.19: 2108753-2108753 on Build GRCh38
rs756763892 Chr.19: 2109043-2109043 on Build GRCh38
rs1055928933 Chr.19: 2108771-2108771 on Build GRCh38
rs767309496 Chr.19: 2108723-2108723 on Build GRCh38
rs759312388 Chr.19: 2108717-2108717 on Build GRCh38
rs986663872 Chr.19: 2108627-2108627 on Build GRCh38
rs56123473 Chr.19: 2108930-2108930 on Build GRCh38
rs553536480 Chr.19: 2108741-2108741 on Build GRCh38
rs891746338 Chr.19: 2108653-2108653 on Build GRCh38
rs924954140 Chr.19: 2108969-2108969 on Build GRCh38
rs770466874 Chr.19: 2108777-2108777 on Build GRCh38
rs776696712 Chr.19: 2108816-2108816 on Build GRCh38
rs56150334 Chr.19: 2109072-2109072 on Build GRCh38
rs945037191 Chr.19: 2108644-2108644 on Build GRCh38
rs760479001 Chr.19: 2108802-2108802 on Build GRCh38
rs761145521 Chr.19: 2108956-2108956 on Build GRCh38
rs912446498 Chr.19: 2108918-2108918 on Build GRCh38
rs763260543 Chr.19: 2108862-2108862 on Build GRCh38
rs545273657 Chr.19: 2108757-2108757 on Build GRCh38
rs758778127 Chr.19: 2108647-2108647 on Build GRCh38
rs986632675 Chr.19: 2108917-2108917 on Build GRCh38
rs775335875 Chr.19: 2108727-2108727 on Build GRCh38
rs750654170 Chr.19: 2108743-2108743 on Build GRCh38
rs557215176 Chr.19: 2108798-2108798 on Build GRCh38
rs569445279 Chr.19: 2108617-2108617 on Build GRCh38
rs2072306 Chr.19: 2109020-2109020 on Build GRCh38
rs765330856 Chr.19: 2108833-2108833 on Build GRCh38
rs545336556 Chr.19: 2108936-2108936 on Build GRCh38
rs564668060 Chr.19: 2109023-2109023 on Build GRCh38
rs936436375 Chr.19: 2108661-2108661 on Build GRCh38
rs1004479532 Chr.19: 2109027-2109027 on Build GRCh38
rs191082490 Chr.19: 2109039-2109039 on Build GRCh38
rs755513592 Chr.19: 2108755-2108755 on Build GRCh38
rs535135693 Chr.19: 2108740-2108740 on Build GRCh38
rs575460608 Chr.19: 2108841-2108841 on Build GRCh38
rs537411375 Chr.19: 2108645-2108645 on Build GRCh38
rs945977676 Chr.19: 2108968-2108968 on Build GRCh38
rs760710224 Chr.19: 2108728-2108728 on Build GRCh38
rs772395600 Chr.19: 2108765-2108765 on Build GRCh38
rs542564084 Chr.19: 2108892-2108892 on Build GRCh38
rs779123929 Chr.19: 2108637-2108637 on Build GRCh38
rs903562988 Chr.19: 2108655-2108655 on Build GRCh38
rs1045933924 Chr.19: 2109014-2109014 on Build GRCh38
rs149059405 Chr.19: 2109040-2109040 on Build GRCh38
rs528142321 Chr.19: 2109018-2109018 on Build GRCh38
rs745508810 Chr.19: 2108779-2108779 on Build GRCh38
rs747485603 Chr.19: 2108672-2108672 on Build GRCh38
rs768567974 Chr.19: 2109067-2109067 on Build GRCh38
rs539688268 Chr.19: 2108662-2108662 on Build GRCh38
rs778467175 Chr.19: 2108778-2108778 on Build GRCh38
rs974547696 Chr.19: 2108812-2108812 on Build GRCh38
rs567719709 Chr.19: 2108677-2108677 on Build GRCh38
rs916368434 Chr.19: 2108992-2108992 on Build GRCh38
rs771574068 Chr.19: 2108795-2108795 on Build GRCh38
rs1007977379 Chr.19: 2109054-2109054 on Build GRCh38
rs780447563 Chr.19: 2108748-2108748 on Build GRCh38
rs761866778 Chr.19: 2108828-2108828 on Build GRCh38
rs769345291 Chr.19: 2108675-2108675 on Build GRCh38
rs962795468 Chr.19: 2108801-2108801 on Build GRCh38
rs948831094 Chr.19: 2109012-2109012 on Build GRCh38
rs753972117 Chr.19: 2108732-2108732 on Build GRCh38
rs761288715 Chr.19: 2108658-2108658 on Build GRCh38
rs768727659 Chr.19: 2108803-2108803 on Build GRCh38
rs1019137106 Chr.19: 2108929-2108929 on Build GRCh38
rs1037312797 Chr.19: 2109030-2109030 on Build GRCh38
rs1045597360 Chr.19: 2108979-2108979 on Build GRCh38
rs764423271 Chr.19: 2109006-2109006 on Build GRCh38
rs922038046 Chr.19: 2108932-2108932 on Build GRCh38
rs770498955 Chr.19: 2108678-2108678 on Build GRCh38
rs936379735 Chr.19: 2108983-2108983 on Build GRCh38
rs1055280657 Chr.19: 2108990-2108990 on Build GRCh38
rs950983977 Chr.19: 2108886-2108886 on Build GRCh38
rs746859077 Chr.19: 2109057-2109057 on Build GRCh38
rs974919772 Chr.19: 2108931-2108931 on Build GRCh38
rs573498476 Chr.19: 2109037-2109037 on Build GRCh38
rs377720238 Chr.19: 2108646-2108646 on Build GRCh38
rs990146757 Chr.19: 2108950-2108950 on Build GRCh38
rs771758918 Chr.19: 2109050-2109050 on Build GRCh38
rs1005103987 Chr.19: 2108895-2108895 on Build GRCh38
rs779794299 Chr.19: 2109055-2109055 on Build GRCh38
rs200642842 Chr.19: 2109032-2109032 on Build GRCh38
rs749841187 Chr.19: 2109044-2109044 on Build GRCh38

View Full Product Details