Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2111559-2112051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3D1
Entrez Gene ID
8943
Gene Name
adaptor related protein complex 3 delta 1 subunit
Gene Aliases
ADTD, hBLVR
UniGene
Hs.512815
Gene Chromosome Location
Chr.19: 2100988-2151620 on Build GRCh38
Transcripts
NM_001261826.1 NM_003938.6 XM_006722932.2 XM_017027422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3422584 c.2850G>T p.K950N Chr.19: 2111766-2111766 on Build GRCh38
3422582 c.2664G>T p.K888N Chr.19: 2111766-2111766 on Build GRCh38
6390093 c.2509A>G p.R837G Chr.19: 2111798-2111798 on Build GRCh38
4280423 c.2582C>T p.A861V Chr.19: 2111725-2111725 on Build GRCh38
6390094 c.2818A>G p.R940G Chr.19: 2111798-2111798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768152921 Chr.19: 2111819-2111819 on Build GRCh38
rs530539281 Chr.19: 2111700-2111700 on Build GRCh38
rs771037677 Chr.19: 2111839-2111839 on Build GRCh38
rs757218055 Chr.19: 2111648-2111648 on Build GRCh38
rs762951857 Chr.19: 2111779-2111779 on Build GRCh38
rs760993312 Chr.19: 2111861-2111861 on Build GRCh38
rs1002186652 Chr.19: 2111849-2111849 on Build GRCh38
rs779572255 Chr.19: 2111744-2111744 on Build GRCh38
rs929800666 Chr.19: 2111956-2111956 on Build GRCh38
rs372236052 Chr.19: 2111780-2111780 on Build GRCh38
rs534854114 Chr.19: 2111960-2111960 on Build GRCh38
rs1046406337 Chr.19: 2111835-2111835 on Build GRCh38
rs55759847 Chr.19: 2111662-2111662 on Build GRCh38
rs79777473 Chr.19: 2111911-2111911 on Build GRCh38
rs925756556 Chr.19: 2111808-2111808 on Build GRCh38
rs1016664349 Chr.19: 2111927-2111927 on Build GRCh38
rs757804607 Chr.19: 2111811-2111811 on Build GRCh38
rs375904659 Chr.19: 2111762-2111762 on Build GRCh38
rs150982298 Chr.19: 2111715-2111715 on Build GRCh38
rs780879347 Chr.19: 2111822-2111822 on Build GRCh38
rs767889885 Chr.19: 2111784-2111784 on Build GRCh38
rs899071701 Chr.19: 2111895-2111895 on Build GRCh38
rs778205414 Chr.19: 2111732-2111732 on Build GRCh38
rs969338426 Chr.19: 2111705-2111705 on Build GRCh38
rs750518726 Chr.19: 2111649-2111649 on Build GRCh38
rs772824283 Chr.19: 2111769-2111769 on Build GRCh38
rs765802494 Chr.19: 2111877-2111877 on Build GRCh38
rs764236512 Chr.19: 2111790-2111792 on Build GRCh38
rs778027207 Chr.19: 2111847-2111847 on Build GRCh38
rs762602190 Chr.19: 2111836-2111836 on Build GRCh38
rs754969586 Chr.19: 2111913-2111914 on Build GRCh38
rs1025360562 Chr.19: 2111696-2111696 on Build GRCh38
rs766493938 Chr.19: 2111716-2111716 on Build GRCh38
rs1052554704 Chr.19: 2111668-2111668 on Build GRCh38
rs752956946 Chr.19: 2111794-2111794 on Build GRCh38
rs779370812 Chr.19: 2111814-2111814 on Build GRCh38
rs747848580 Chr.19: 2111827-2111827 on Build GRCh38
rs1013988288 Chr.19: 2111607-2111607 on Build GRCh38
rs775206068 Chr.19: 2111643-2111643 on Build GRCh38
rs751622663 Chr.19: 2111717-2111717 on Build GRCh38
rs201826090 Chr.19: 2111701-2111701 on Build GRCh38
rs751708491 Chr.19: 2111781-2111781 on Build GRCh38
rs150110874 Chr.19: 2112034-2112034 on Build GRCh38
rs367708267 Chr.19: 2111724-2111724 on Build GRCh38
rs1020689642 Chr.19: 2111993-2111993 on Build GRCh38
rs529475787 Chr.19: 2111719-2111719 on Build GRCh38
rs556612715 Chr.19: 2111818-2111818 on Build GRCh38
rs949513143 Chr.19: 2112003-2112003 on Build GRCh38
rs750369269 Chr.19: 2111702-2111702 on Build GRCh38
rs538222861 Chr.19: 2111965-2111965 on Build GRCh38
rs763068481 Chr.19: 2111760-2111762 on Build GRCh38
rs950907842 Chr.19: 2111624-2111624 on Build GRCh38
rs978384269 Chr.19: 2111957-2111957 on Build GRCh38
rs370011282 Chr.19: 2111776-2111776 on Build GRCh38
rs199976556 Chr.19: 2111799-2111799 on Build GRCh38
rs1025395190 Chr.19: 2111703-2111703 on Build GRCh38
rs78496920 Chr.19: 2111625-2111625 on Build GRCh38
rs757550445 Chr.19: 2111903-2111903 on Build GRCh38
rs892592454 Chr.19: 2111824-2111824 on Build GRCh38
rs774809764 Chr.19: 2111669-2111669 on Build GRCh38
rs768347532 Chr.19: 2111752-2111752 on Build GRCh38
rs770444809 Chr.19: 2111660-2111660 on Build GRCh38
rs74967342 Chr.19: 2111875-2111875 on Build GRCh38
rs904410548 Chr.19: 2111657-2111657 on Build GRCh38
rs755192246 Chr.19: 2111654-2111654 on Build GRCh38
rs1005247097 Chr.19: 2111923-2111923 on Build GRCh38
rs551488706 Chr.19: 2111834-2111834 on Build GRCh38
rs376574497 Chr.19: 2111789-2111789 on Build GRCh38
rs539498849 Chr.19: 2111585-2111585 on Build GRCh38
rs969703151 Chr.19: 2111614-2111614 on Build GRCh38
rs559730562 Chr.19: 2111655-2111655 on Build GRCh38
rs751766280 Chr.19: 2111628-2111628 on Build GRCh38
rs759749911 Chr.19: 2111845-2111845 on Build GRCh38
rs555622746 Chr.19: 2111645-2111645 on Build GRCh38
rs754115647 Chr.19: 2111805-2111805 on Build GRCh38
rs113189606 Chr.19: 2111831-2111831 on Build GRCh38
rs573877797 Chr.19: 2111934-2111934 on Build GRCh38
rs544684683 Chr.19: 2111658-2111658 on Build GRCh38
rs769262667 Chr.19: 2111829-2111829 on Build GRCh38
rs760328728 Chr.19: 2111672-2111672 on Build GRCh38
rs775695925 Chr.19: 2111656-2111656 on Build GRCh38
rs771279999 Chr.19: 2111742-2111742 on Build GRCh38
rs754278669 Chr.19: 2111872-2111872 on Build GRCh38
rs577860175 Chr.19: 2111977-2111977 on Build GRCh38
rs544959152 Chr.19: 2112022-2112022 on Build GRCh38
rs746474101 Chr.19: 2111747-2111747 on Build GRCh38
rs776237140 Chr.19: 2111758-2111758 on Build GRCh38
rs763697220 Chr.19: 2111673-2111673 on Build GRCh38
rs201127154 Chr.19: 2111804-2111804 on Build GRCh38
rs980601406 Chr.19: 2111618-2111618 on Build GRCh38
rs142590416 Chr.19: 2111596-2111596 on Build GRCh38
rs2074959 Chr.19: 2111650-2111650 on Build GRCh38
rs919045163 Chr.19: 2111736-2111736 on Build GRCh38
rs764173590 Chr.19: 2111871-2111871 on Build GRCh38
rs547293900 Chr.19: 2111633-2111633 on Build GRCh38
rs371484246 Chr.19: 2111761-2111761 on Build GRCh38
rs776432599 Chr.19: 2111680-2111680 on Build GRCh38
rs914026703 Chr.19: 2111983-2111983 on Build GRCh38
rs372796139 Chr.19: 2111586-2111586 on Build GRCh38
rs967426921 Chr.19: 2112011-2112012 on Build GRCh38
rs776351872 Chr.19: 2111874-2111875 on Build GRCh38
rs749819797 Chr.19: 2111735-2111735 on Build GRCh38
rs540094467 Chr.19: 2111914-2111915 on Build GRCh38
rs988331051 Chr.19: 2111979-2111979 on Build GRCh38
rs913314681 Chr.19: 2111652-2111652 on Build GRCh38
rs554350898 Chr.19: 2111646-2111646 on Build GRCh38
rs368249548 Chr.19: 2111665-2111665 on Build GRCh38
rs775648858 Chr.19: 2111848-2111848 on Build GRCh38
rs547683806 Chr.19: 2111725-2111725 on Build GRCh38
rs757532166 Chr.19: 2111876-2111876 on Build GRCh38
rs978836886 Chr.19: 2112027-2112030 on Build GRCh38
rs567891146 Chr.19: 2111950-2111950 on Build GRCh38
rs996307463 Chr.19: 2111964-2111964 on Build GRCh38
rs555924373 Chr.19: 2111916-2111916 on Build GRCh38
rs570107821 Chr.19: 2111907-2111907 on Build GRCh38
rs747220969 Chr.19: 2111651-2111651 on Build GRCh38
rs773744394 Chr.19: 2111661-2111661 on Build GRCh38
rs114693097 Chr.19: 2111656-2111656 on Build GRCh38
rs543559100 Chr.19: 2111630-2111630 on Build GRCh38

View Full Product Details